Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8389411-8389825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX3
Entrez Gene ID
8310
Gene Name
acyl-CoA oxidase 3, pristanoyl
Gene Aliases
-
UniGene
Hs.479122
Gene Chromosome Location
Chr.4: 8366263-8440725 on Build GRCh38
Transcripts
NM_001101667.1 NM_003501.2 XM_005248011.4 XM_005248013.3 XM_011513565.2 XM_011513566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1431411 c.1336C>A p.P446T Chr.4: 8389699-8389699 on Build GRCh38
5679111 c.1344C>T p.C448C Chr.4: 8389691-8389691 on Build GRCh38
3826338 c.1324G>A p.D442N Chr.4: 8389711-8389711 on Build GRCh38
6352704 c.1404C>G p.L468L Chr.4: 8389631-8389631 on Build GRCh38
5736904 c.1306C>T p.R436W Chr.4: 8389729-8389729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750134109 Chr.4: 8389741-8389741 on Build GRCh38
rs745464321 Chr.4: 8389669-8389669 on Build GRCh38
rs769769967 Chr.4: 8389561-8389561 on Build GRCh38
rs757998953 Chr.4: 8389559-8389559 on Build GRCh38
rs543543995 Chr.4: 8389798-8389798 on Build GRCh38
rs768706873 Chr.4: 8389695-8389695 on Build GRCh38
rs57692256 Chr.4: 8389524-8389524 on Build GRCh38
rs372482219 Chr.4: 8389583-8389583 on Build GRCh38
rs1053952861 Chr.4: 8389589-8389589 on Build GRCh38
rs116406348 Chr.4: 8389631-8389631 on Build GRCh38
rs992727737 Chr.4: 8389720-8389720 on Build GRCh38
rs754384280 Chr.4: 8389643-8389643 on Build GRCh38
rs369257303 Chr.4: 8389762-8389762 on Build GRCh38
rs760103634 Chr.4: 8389727-8389727 on Build GRCh38
rs144215776 Chr.4: 8389723-8389723 on Build GRCh38
rs768903237 Chr.4: 8389614-8389614 on Build GRCh38
rs373434833 Chr.4: 8389765-8389765 on Build GRCh38
rs961314093 Chr.4: 8389511-8389511 on Build GRCh38
rs914039449 Chr.4: 8389794-8389794 on Build GRCh38
rs377178118 Chr.4: 8389666-8389666 on Build GRCh38
rs368490039 Chr.4: 8389461-8389461 on Build GRCh38
rs745471901 Chr.4: 8389627-8389627 on Build GRCh38
rs201011526 Chr.4: 8389590-8389590 on Build GRCh38
rs770604062 Chr.4: 8389605-8389605 on Build GRCh38
rs1030027155 Chr.4: 8389619-8389619 on Build GRCh38
rs536521584 Chr.4: 8389684-8389684 on Build GRCh38
rs756596645 Chr.4: 8389662-8389662 on Build GRCh38
rs750792736 Chr.4: 8389582-8389582 on Build GRCh38
rs918041682 Chr.4: 8389743-8389743 on Build GRCh38
rs766899375 Chr.4: 8389737-8389737 on Build GRCh38
rs762070735 Chr.4: 8389574-8389574 on Build GRCh38
rs755457059 Chr.4: 8389645-8389645 on Build GRCh38
rs780917248 Chr.4: 8389606-8389606 on Build GRCh38
rs776969752 Chr.4: 8389726-8389726 on Build GRCh38
rs371322956 Chr.4: 8389713-8389713 on Build GRCh38
rs747995279 Chr.4: 8389771-8389771 on Build GRCh38
rs762003746 Chr.4: 8389624-8389624 on Build GRCh38
rs552214471 Chr.4: 8389594-8389594 on Build GRCh38
rs985134972 Chr.4: 8389438-8389438 on Build GRCh38
rs112332730 Chr.4: 8389655-8389655 on Build GRCh38
rs767781429 Chr.4: 8389627-8389627 on Build GRCh38
rs1028393578 Chr.4: 8389628-8389628 on Build GRCh38
rs1017955789 Chr.4: 8389648-8389648 on Build GRCh38
rs749294098 Chr.4: 8389685-8389685 on Build GRCh38
rs773513363 Chr.4: 8389716-8389716 on Build GRCh38
rs532690394 Chr.4: 8389473-8389473 on Build GRCh38
rs774557016 Chr.4: 8389617-8389617 on Build GRCh38
rs992904936 Chr.4: 8389518-8389518 on Build GRCh38
rs773144653 Chr.4: 8389773-8389773 on Build GRCh38
rs935470341 Chr.4: 8389477-8389477 on Build GRCh38
rs1056776904 Chr.4: 8389484-8389484 on Build GRCh38
rs745684291 Chr.4: 8389607-8389607 on Build GRCh38
rs1028130468 Chr.4: 8389455-8389455 on Build GRCh38
rs887751859 Chr.4: 8389535-8389535 on Build GRCh38
rs775505972 Chr.4: 8389562-8389562 on Build GRCh38
rs368543216 Chr.4: 8389670-8389670 on Build GRCh38
rs572332752 Chr.4: 8389659-8389659 on Build GRCh38
rs774360716 Chr.4: 8389702-8389702 on Build GRCh38
rs979202117 Chr.4: 8389691-8389691 on Build GRCh38
rs572063146 Chr.4: 8389770-8389770 on Build GRCh38
rs144357499 Chr.4: 8389474-8389474 on Build GRCh38
rs114372559 Chr.4: 8389571-8389571 on Build GRCh38
rs149402565 Chr.4: 8389611-8389611 on Build GRCh38
rs146052311 Chr.4: 8389729-8389729 on Build GRCh38
rs375898726 Chr.4: 8389718-8389718 on Build GRCh38
rs916917868 Chr.4: 8389721-8389721 on Build GRCh38
rs766688949 Chr.4: 8389635-8389635 on Build GRCh38
rs747316012 Chr.4: 8389779-8389779 on Build GRCh38
rs755811635 Chr.4: 8389752-8389752 on Build GRCh38
rs36055234 Chr.4: 8389438-8389439 on Build GRCh38
rs780059756 Chr.4: 8389760-8389760 on Build GRCh38
rs113578128 Chr.4: 8389613-8389613 on Build GRCh38
rs748272279 Chr.4: 8389703-8389703 on Build GRCh38
rs767825276 Chr.4: 8389580-8389580 on Build GRCh38
rs199993833 Chr.4: 8389728-8389728 on Build GRCh38
rs1045076752 Chr.4: 8389531-8389531 on Build GRCh38
rs763142029 Chr.4: 8389570-8389570 on Build GRCh38
rs201816216 Chr.4: 8389612-8389612 on Build GRCh38
rs530316561 Chr.4: 8389548-8389548 on Build GRCh38
rs757933599 Chr.4: 8389587-8389587 on Build GRCh38
rs945633258 Chr.4: 8389796-8389796 on Build GRCh38
rs1036444751 Chr.4: 8389789-8389789 on Build GRCh38

View Full Product Details