Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 13569409-13569910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOD1L1
Entrez Gene ID
259282
Gene Name
biorientation of chromosomes in cell division 1 like 1
Gene Aliases
BOD1L, FAM44A
UniGene
Hs.744935
Gene Chromosome Location
Chr.4: 13568738-13627721 on Build GRCh38
Transcripts
NM_148894.2 XM_005248150.3 XM_005248151.3 XM_006713958.3 XM_011513827.2 XM_011513829.2 XM_011513830.2 XM_017008009.1 XM_017008010.1

SNP Detail

SNP ID Chromosome Location
rs904147431 Chr.4: 13569741-13569741 on Build GRCh38
rs1001613681 Chr.4: 13569769-13569769 on Build GRCh38
rs904995173 Chr.4: 13569755-13569755 on Build GRCh38
rs1051578132 Chr.4: 13569634-13569634 on Build GRCh38
rs1014538168 Chr.4: 13569800-13569800 on Build GRCh38
rs951568720 Chr.4: 13569819-13569819 on Build GRCh38
rs543192014 Chr.4: 13569845-13569845 on Build GRCh38
rs529048640 Chr.4: 13569521-13569521 on Build GRCh38
rs908575318 Chr.4: 13569646-13569646 on Build GRCh38
rs900409187 Chr.4: 13569605-13569605 on Build GRCh38
rs986847371 Chr.4: 13569624-13569624 on Build GRCh38
rs942699502 Chr.4: 13569542-13569542 on Build GRCh38
rs984131420 Chr.4: 13569839-13569839 on Build GRCh38
rs558175713 Chr.4: 13569498-13569498 on Build GRCh38
rs759143713 Chr.4: 13569440-13569440 on Build GRCh38
rs112534632 Chr.4: 13569687-13569687 on Build GRCh38
rs560497845 Chr.4: 13569609-13569609 on Build GRCh38
rs960552529 Chr.4: 13569799-13569799 on Build GRCh38
rs751954616 Chr.4: 13569716-13569716 on Build GRCh38
rs909978391 Chr.4: 13569843-13569843 on Build GRCh38
rs560900038 Chr.4: 13569686-13569686 on Build GRCh38
rs1056134783 Chr.4: 13569790-13569790 on Build GRCh38
rs930050936 Chr.4: 13569521-13569521 on Build GRCh38
rs150716450 Chr.4: 13569520-13569521 on Build GRCh38
rs1009218981 Chr.4: 13569846-13569846 on Build GRCh38
rs975595929 Chr.4: 13569577-13569577 on Build GRCh38
rs753246693 Chr.4: 13569520-13569520 on Build GRCh38
rs745508973 Chr.4: 13569700-13569700 on Build GRCh38
rs758885393 Chr.4: 13569533-13569533 on Build GRCh38
rs1044923176 Chr.4: 13569728-13569728 on Build GRCh38
rs34402437 Chr.4: 13569624-13569625 on Build GRCh38
rs955461716 Chr.4: 13569620-13569620 on Build GRCh38
rs920508993 Chr.4: 13569709-13569709 on Build GRCh38
rs892171811 Chr.4: 13569818-13569818 on Build GRCh38
rs112053136 Chr.4: 13569826-13569826 on Build GRCh38
rs930525924 Chr.4: 13569711-13569711 on Build GRCh38
rs939112757 Chr.4: 13569780-13569780 on Build GRCh38
rs542230399 Chr.4: 13569543-13569543 on Build GRCh38
rs1028468739 Chr.4: 13569601-13569601 on Build GRCh38
rs968646694 Chr.4: 13569547-13569547 on Build GRCh38
rs188708986 Chr.4: 13569534-13569534 on Build GRCh38
rs1034061130 Chr.4: 13569791-13569791 on Build GRCh38
rs562956823 Chr.4: 13569869-13569869 on Build GRCh38
rs964195401 Chr.4: 13569853-13569853 on Build GRCh38
rs933291167 Chr.4: 13569618-13569618 on Build GRCh38

View Full Product Details