Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 13569703-13570241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOD1L1
Entrez Gene ID
259282
Gene Name
biorientation of chromosomes in cell division 1 like 1
Gene Aliases
BOD1L, FAM44A
UniGene
Hs.744935
Gene Chromosome Location
Chr.4: 13568738-13627721 on Build GRCh38
Transcripts
NM_148894.2 XM_005248150.3 XM_005248151.3 XM_006713958.3 XM_011513827.2 XM_011513829.2 XM_011513830.2 XM_017008009.1 XM_017008010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444951 c.9092C>A p.P3031H Chr.4: 13570075-13570075 on Build GRCh38
5731246 c.9048A>G p.T3016T Chr.4: 13570119-13570119 on Build GRCh38
256410 c.9146C>T p.A3049V Chr.4: 13570021-13570021 on Build GRCh38
6099322 c.9136G>T p.V3046L Chr.4: 13570031-13570031 on Build GRCh38
1051263 c.9095G>T p.G3032V Chr.4: 13570072-13570072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374872080 Chr.4: 13570081-13570081 on Build GRCh38
rs780264217 Chr.4: 13569999-13569999 on Build GRCh38
rs904995173 Chr.4: 13569755-13569755 on Build GRCh38
rs543192014 Chr.4: 13569845-13569845 on Build GRCh38
rs771487493 Chr.4: 13570116-13570116 on Build GRCh38
rs778357205 Chr.4: 13570113-13570113 on Build GRCh38
rs984131420 Chr.4: 13569839-13569839 on Build GRCh38
rs757047278 Chr.4: 13569973-13569973 on Build GRCh38
rs960552529 Chr.4: 13569799-13569799 on Build GRCh38
rs535079554 Chr.4: 13569989-13569989 on Build GRCh38
rs762546163 Chr.4: 13570023-13570023 on Build GRCh38
rs1023001310 Chr.4: 13569897-13569897 on Build GRCh38
rs530629740 Chr.4: 13569910-13569910 on Build GRCh38
rs755111547 Chr.4: 13570003-13570003 on Build GRCh38
rs1056134783 Chr.4: 13569790-13569790 on Build GRCh38
rs1009218981 Chr.4: 13569846-13569846 on Build GRCh38
rs760461676 Chr.4: 13570145-13570145 on Build GRCh38
rs866301545 Chr.4: 13570093-13570093 on Build GRCh38
rs1049999965 Chr.4: 13570167-13570167 on Build GRCh38
rs747400099 Chr.4: 13570020-13570020 on Build GRCh38
rs181759640 Chr.4: 13570153-13570153 on Build GRCh38
rs1017142186 Chr.4: 13570147-13570147 on Build GRCh38
rs181404914 Chr.4: 13569940-13569940 on Build GRCh38
rs761607299 Chr.4: 13570066-13570066 on Build GRCh38
rs892171811 Chr.4: 13569818-13569818 on Build GRCh38
rs112053136 Chr.4: 13569826-13569826 on Build GRCh38
rs372133509 Chr.4: 13570031-13570031 on Build GRCh38
rs747364083 Chr.4: 13570115-13570115 on Build GRCh38
rs930580122 Chr.4: 13569990-13569990 on Build GRCh38
rs939112757 Chr.4: 13569780-13569780 on Build GRCh38
rs750122596 Chr.4: 13570085-13570085 on Build GRCh38
rs756288959 Chr.4: 13569993-13569993 on Build GRCh38
rs981062362 Chr.4: 13569992-13569992 on Build GRCh38
rs964195401 Chr.4: 13569853-13569853 on Build GRCh38
rs922651906 Chr.4: 13569944-13569944 on Build GRCh38
rs904147431 Chr.4: 13569741-13569741 on Build GRCh38
rs1001613681 Chr.4: 13569769-13569769 on Build GRCh38
rs1014538168 Chr.4: 13569800-13569800 on Build GRCh38
rs554504093 Chr.4: 13570205-13570205 on Build GRCh38
rs951568720 Chr.4: 13569819-13569819 on Build GRCh38
rs913642892 Chr.4: 13570103-13570103 on Build GRCh38
rs547228920 Chr.4: 13569952-13569952 on Build GRCh38
rs768378970 Chr.4: 13570124-13570124 on Build GRCh38
rs369852170 Chr.4: 13570097-13570097 on Build GRCh38
rs199706682 Chr.4: 13570027-13570027 on Build GRCh38
rs369566334 Chr.4: 13570016-13570016 on Build GRCh38
rs755025704 Chr.4: 13570098-13570098 on Build GRCh38
rs758760550 Chr.4: 13570109-13570109 on Build GRCh38
rs767522018 Chr.4: 13570067-13570067 on Build GRCh38
rs533285916 Chr.4: 13569943-13569943 on Build GRCh38
rs550716949 Chr.4: 13569911-13569911 on Build GRCh38
rs1001317961 Chr.4: 13570036-13570036 on Build GRCh38
rs546476024 Chr.4: 13569948-13569948 on Build GRCh38
rs773695033 Chr.4: 13570135-13570135 on Build GRCh38
rs904121018 Chr.4: 13569900-13569900 on Build GRCh38
rs779589902 Chr.4: 13570046-13570046 on Build GRCh38
rs909978391 Chr.4: 13569843-13569843 on Build GRCh38
rs534530395 Chr.4: 13570179-13570179 on Build GRCh38
rs773262543 Chr.4: 13570139-13570139 on Build GRCh38
rs891322139 Chr.4: 13570185-13570185 on Build GRCh38
rs757557330 Chr.4: 13569906-13569906 on Build GRCh38
rs184156653 Chr.4: 13570012-13570012 on Build GRCh38
rs1008385457 Chr.4: 13570188-13570188 on Build GRCh38
rs769512613 Chr.4: 13570021-13570021 on Build GRCh38
rs919897456 Chr.4: 13569985-13569985 on Build GRCh38
rs774325271 Chr.4: 13570063-13570063 on Build GRCh38
rs753653472 Chr.4: 13570171-13570171 on Build GRCh38
rs779473224 Chr.4: 13570008-13570008 on Build GRCh38
rs200736085 Chr.4: 13569974-13569974 on Build GRCh38
rs779820610 Chr.4: 13570122-13570122 on Build GRCh38
rs201304977 Chr.4: 13570138-13570138 on Build GRCh38
rs1044923176 Chr.4: 13569728-13569728 on Build GRCh38
rs778806502 Chr.4: 13570104-13570104 on Build GRCh38
rs752849745 Chr.4: 13570105-13570105 on Build GRCh38
rs749312096 Chr.4: 13570002-13570002 on Build GRCh38
rs755867029 Chr.4: 13569961-13569961 on Build GRCh38
rs761391897 Chr.4: 13570137-13570137 on Build GRCh38
rs759711489 Chr.4: 13570174-13570174 on Build GRCh38
rs147028451 Chr.4: 13570096-13570096 on Build GRCh38
rs781532571 Chr.4: 13569907-13569907 on Build GRCh38
rs765498283 Chr.4: 13570176-13570176 on Build GRCh38
rs748999437 Chr.4: 13570123-13570123 on Build GRCh38
rs1034061130 Chr.4: 13569791-13569791 on Build GRCh38
rs562956823 Chr.4: 13569869-13569869 on Build GRCh38

View Full Product Details