Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 13582120-13582634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOD1L1
Entrez Gene ID
259282
Gene Name
biorientation of chromosomes in cell division 1 like 1
Gene Aliases
BOD1L, FAM44A
UniGene
Hs.744935
Gene Chromosome Location
Chr.4: 13568738-13627721 on Build GRCh38
Transcripts
NM_148894.2 XM_005248150.3 XM_005248151.3 XM_006713958.3 XM_011513827.2 XM_011513829.2 XM_011513830.2 XM_017008009.1 XM_017008010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4823271 c.8587C>T p.Q2863* Chr.4: 13582242-13582242 on Build GRCh38
3409060 c.8557G>A p.E2853K Chr.4: 13582272-13582272 on Build GRCh38
4138048 c.8521G>C p.E2841Q Chr.4: 13582308-13582308 on Build GRCh38
149649 c.8536G>A p.E2846K Chr.4: 13582293-13582293 on Build GRCh38
3010642 c.8545G>T p.G2849C Chr.4: 13582284-13582284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745466640 Chr.4: 13582204-13582204 on Build GRCh38
rs759508555 Chr.4: 13582604-13582604 on Build GRCh38
rs114343018 Chr.4: 13582270-13582270 on Build GRCh38
rs758993354 Chr.4: 13582293-13582293 on Build GRCh38
rs753805967 Chr.4: 13582268-13582268 on Build GRCh38
rs968685142 Chr.4: 13582560-13582560 on Build GRCh38
rs755770417 Chr.4: 13582317-13582317 on Build GRCh38
rs766597027 Chr.4: 13582265-13582265 on Build GRCh38
rs750241713 Chr.4: 13582260-13582260 on Build GRCh38
rs748743551 Chr.4: 13582330-13582330 on Build GRCh38
rs959582579 Chr.4: 13582599-13582599 on Build GRCh38
rs553252507 Chr.4: 13582611-13582611 on Build GRCh38
rs980808588 Chr.4: 13582581-13582581 on Build GRCh38
rs1028873278 Chr.4: 13582384-13582384 on Build GRCh38
rs779813228 Chr.4: 13582325-13582325 on Build GRCh38
rs764145811 Chr.4: 13582236-13582238 on Build GRCh38
rs17745075 Chr.4: 13582484-13582484 on Build GRCh38
rs573510116 Chr.4: 13582340-13582340 on Build GRCh38
rs772031641 Chr.4: 13582350-13582350 on Build GRCh38
rs141975549 Chr.4: 13582310-13582310 on Build GRCh38
rs1030928581 Chr.4: 13582423-13582423 on Build GRCh38
rs577854208 Chr.4: 13582182-13582182 on Build GRCh38
rs201668743 Chr.4: 13582287-13582287 on Build GRCh38
rs184233130 Chr.4: 13582592-13582592 on Build GRCh38
rs926544470 Chr.4: 13582587-13582587 on Build GRCh38
rs780967935 Chr.4: 13582199-13582199 on Build GRCh38
rs552370622 Chr.4: 13582596-13582596 on Build GRCh38
rs559616475 Chr.4: 13582388-13582388 on Build GRCh38
rs575708586 Chr.4: 13582456-13582456 on Build GRCh38
rs577033753 Chr.4: 13582276-13582276 on Build GRCh38
rs13134162 Chr.4: 13582528-13582528 on Build GRCh38
rs962126244 Chr.4: 13582469-13582469 on Build GRCh38
rs961741744 Chr.4: 13582203-13582203 on Build GRCh38
rs1056625648 Chr.4: 13582303-13582303 on Build GRCh38
rs529906844 Chr.4: 13582432-13582432 on Build GRCh38
rs776948890 Chr.4: 13582360-13582360 on Build GRCh38
rs536790610 Chr.4: 13582220-13582220 on Build GRCh38
rs376375766 Chr.4: 13582221-13582221 on Build GRCh38
rs953256858 Chr.4: 13582428-13582428 on Build GRCh38
rs774118037 Chr.4: 13582276-13582277 on Build GRCh38
rs774415200 Chr.4: 13582238-13582238 on Build GRCh38
rs188564359 Chr.4: 13582600-13582600 on Build GRCh38
rs761482534 Chr.4: 13582255-13582255 on Build GRCh38
rs980551733 Chr.4: 13582586-13582586 on Build GRCh38
rs768252476 Chr.4: 13582336-13582336 on Build GRCh38
rs900773515 Chr.4: 13582387-13582387 on Build GRCh38
rs564529155 Chr.4: 13582555-13582555 on Build GRCh38
rs745945591 Chr.4: 13582157-13582157 on Build GRCh38
rs13139564 Chr.4: 13582529-13582529 on Build GRCh38
rs967420899 Chr.4: 13582370-13582382 on Build GRCh38
rs548195967 Chr.4: 13582171-13582171 on Build GRCh38
rs778241330 Chr.4: 13582309-13582309 on Build GRCh38
rs760480201 Chr.4: 13582354-13582354 on Build GRCh38
rs370508182 Chr.4: 13582264-13582264 on Build GRCh38
rs115357951 Chr.4: 13582370-13582370 on Build GRCh38
rs539609536 Chr.4: 13582286-13582286 on Build GRCh38
rs766578654 Chr.4: 13582359-13582359 on Build GRCh38
rs1021544530 Chr.4: 13582551-13582551 on Build GRCh38
rs768463470 Chr.4: 13582237-13582237 on Build GRCh38
rs1010276780 Chr.4: 13582524-13582524 on Build GRCh38
rs951755040 Chr.4: 13582573-13582573 on Build GRCh38
rs748071679 Chr.4: 13582344-13582344 on Build GRCh38
rs867644240 Chr.4: 13582244-13582244 on Build GRCh38
rs1018721102 Chr.4: 13582445-13582445 on Build GRCh38
rs17806907 Chr.4: 13582214-13582214 on Build GRCh38
rs767351383 Chr.4: 13582258-13582258 on Build GRCh38
rs375028832 Chr.4: 13582352-13582352 on Build GRCh38
rs762598204 Chr.4: 13582208-13582211 on Build GRCh38

View Full Product Details