Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 13587361-13587862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOD1L1
Entrez Gene ID
259282
Gene Name
biorientation of chromosomes in cell division 1 like 1
Gene Aliases
BOD1L, FAM44A
UniGene
Hs.744935
Gene Chromosome Location
Chr.4: 13568738-13627721 on Build GRCh38
Transcripts
NM_148894.2 XM_005248150.3 XM_005248151.3 XM_006713958.3 XM_011513827.2 XM_011513829.2 XM_011513830.2 XM_017008009.1 XM_017008010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3767699 c.8323C>T p.Q2775* Chr.4: 13587729-13587729 on Build GRCh38
4428701 c.8312G>C p.R2771T Chr.4: 13587740-13587740 on Build GRCh38
1427050 c.8285A>C p.E2762A Chr.4: 13587767-13587767 on Build GRCh38
7425591 c.8311A>G p.R2771G Chr.4: 13587741-13587741 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035186820 Chr.4: 13587473-13587473 on Build GRCh38
rs947070962 Chr.4: 13587827-13587827 on Build GRCh38
rs755758957 Chr.4: 13587685-13587690 on Build GRCh38
rs960094237 Chr.4: 13587790-13587790 on Build GRCh38
rs373545205 Chr.4: 13587777-13587777 on Build GRCh38
rs919377560 Chr.4: 13587490-13587490 on Build GRCh38
rs905403150 Chr.4: 13587668-13587668 on Build GRCh38
rs969641617 Chr.4: 13587449-13587449 on Build GRCh38
rs116819851 Chr.4: 13587429-13587429 on Build GRCh38
rs1022396867 Chr.4: 13587662-13587662 on Build GRCh38
rs900737692 Chr.4: 13587545-13587545 on Build GRCh38
rs113831797 Chr.4: 13587522-13587522 on Build GRCh38
rs145860949 Chr.4: 13587459-13587459 on Build GRCh38
rs749980966 Chr.4: 13587794-13587794 on Build GRCh38
rs763729994 Chr.4: 13587762-13587762 on Build GRCh38
rs1004967813 Chr.4: 13587596-13587596 on Build GRCh38
rs909774952 Chr.4: 13587547-13587547 on Build GRCh38
rs536595552 Chr.4: 13587506-13587506 on Build GRCh38
rs771428809 Chr.4: 13587739-13587739 on Build GRCh38
rs942639785 Chr.4: 13587571-13587571 on Build GRCh38
rs761363500 Chr.4: 13587768-13587768 on Build GRCh38
rs963380912 Chr.4: 13587643-13587643 on Build GRCh38
rs572131384 Chr.4: 13587701-13587701 on Build GRCh38
rs777058653 Chr.4: 13587466-13587466 on Build GRCh38
rs775640014 Chr.4: 13587615-13587615 on Build GRCh38
rs962274756 Chr.4: 13587551-13587551 on Build GRCh38
rs772087503 Chr.4: 13587692-13587692 on Build GRCh38
rs1026387590 Chr.4: 13587758-13587758 on Build GRCh38
rs149555351 Chr.4: 13587803-13587803 on Build GRCh38
rs773913832 Chr.4: 13587767-13587767 on Build GRCh38
rs912942434 Chr.4: 13587816-13587816 on Build GRCh38
rs996760749 Chr.4: 13587548-13587548 on Build GRCh38
rs759950314 Chr.4: 13587741-13587741 on Build GRCh38
rs994098094 Chr.4: 13587752-13587752 on Build GRCh38
rs901022140 Chr.4: 13587595-13587595 on Build GRCh38
rs749100705 Chr.4: 13587658-13587658 on Build GRCh38
rs558389339 Chr.4: 13587669-13587669 on Build GRCh38
rs369354154 Chr.4: 13587679-13587679 on Build GRCh38
rs555188941 Chr.4: 13587835-13587835 on Build GRCh38
rs10939491 Chr.4: 13587438-13587438 on Build GRCh38
rs781189767 Chr.4: 13587574-13587574 on Build GRCh38
rs776920790 Chr.4: 13587740-13587740 on Build GRCh38
rs763585968 Chr.4: 13587684-13587685 on Build GRCh38
rs752261303 Chr.4: 13587656-13587656 on Build GRCh38
rs114602753 Chr.4: 13587542-13587542 on Build GRCh38
rs980365346 Chr.4: 13587754-13587754 on Build GRCh38
rs747430912 Chr.4: 13587735-13587735 on Build GRCh38
rs386671909 Chr.4: 13587428-13587429 on Build GRCh38
rs1002463016 Chr.4: 13587670-13587670 on Build GRCh38
rs900907889 Chr.4: 13587392-13587392 on Build GRCh38
rs1052172002 Chr.4: 13587637-13587637 on Build GRCh38
rs748258014 Chr.4: 13587683-13587683 on Build GRCh38
rs938425921 Chr.4: 13587481-13587481 on Build GRCh38
rs538592470 Chr.4: 13587647-13587647 on Build GRCh38
rs6838908 Chr.4: 13587588-13587588 on Build GRCh38
rs985227521 Chr.4: 13587543-13587543 on Build GRCh38
rs57205020 Chr.4: 13587484-13587484 on Build GRCh38
rs1022531835 Chr.4: 13587446-13587446 on Build GRCh38
rs1015375039 Chr.4: 13587611-13587611 on Build GRCh38
rs10939490 Chr.4: 13587390-13587390 on Build GRCh38
rs997893889 Chr.4: 13587420-13587420 on Build GRCh38
rs77337617 Chr.4: 13587428-13587428 on Build GRCh38
rs759787721 Chr.4: 13587485-13587485 on Build GRCh38

View Full Product Details