Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15008213-15008725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB2
Entrez Gene ID
132864
Gene Name
cytoplasmic polyadenylation element binding protein 2
Gene Aliases
CPE-BP2, CPEB-2, hCPEB-2
UniGene
Hs.656937
Gene Chromosome Location
Chr.4: 15002451-15070153 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Gene Symbol
LOC101929095
Entrez Gene ID
101929095
Gene Name
uncharacterized LOC101929095
Gene Aliases
-
UniGene
Hs.656908
Gene Chromosome Location
Chr.4: 15004942-15427914 on Build GRCh38
Transcripts
NM_001177381.1 NM_001177382.1 NM_001177383.1 NM_001177384.1 NM_182485.2 NM_182646.2 XM_005248135.3 XM_011513777.2 XM_011513778.2 XM_011513779.1 XM_017007733.1 XM_017007734.1 NR_125911.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1685137 c.1961A>G p.Q654R Chr.4: 15008354-15008354 on Build GRCh38
6400799 c.674C>A p.S225* Chr.4: 15008378-15008378 on Build GRCh38
6634516 c.693C>A p.C231* Chr.4: 15008397-15008397 on Build GRCh38
6371630 c.2014G>A p.G672R Chr.4: 15008407-15008407 on Build GRCh38
6634515 c.2004C>A p.C668* Chr.4: 15008397-15008397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048677663 Chr.4: 15008284-15008284 on Build GRCh38
rs919005099 Chr.4: 15008276-15008276 on Build GRCh38
rs943285004 Chr.4: 15008502-15008502 on Build GRCh38
rs892658814 Chr.4: 15008676-15008676 on Build GRCh38
rs768939954 Chr.4: 15008326-15008326 on Build GRCh38
rs757550324 Chr.4: 15008302-15008302 on Build GRCh38
rs780150401 Chr.4: 15008460-15008460 on Build GRCh38
rs572067309 Chr.4: 15008510-15008510 on Build GRCh38
rs901639460 Chr.4: 15008258-15008258 on Build GRCh38
rs772097287 Chr.4: 15008362-15008362 on Build GRCh38
rs1037792115 Chr.4: 15008597-15008597 on Build GRCh38
rs960947184 Chr.4: 15008499-15008499 on Build GRCh38
rs565976876 Chr.4: 15008513-15008513 on Build GRCh38
rs886145325 Chr.4: 15008275-15008275 on Build GRCh38
rs751209038 Chr.4: 15008440-15008440 on Build GRCh38
rs34393704 Chr.4: 15008668-15008669 on Build GRCh38
rs182244663 Chr.4: 15008243-15008243 on Build GRCh38
rs933139773 Chr.4: 15008522-15008522 on Build GRCh38
rs886177700 Chr.4: 15008553-15008553 on Build GRCh38
rs774470310 Chr.4: 15008344-15008344 on Build GRCh38
rs1047535614 Chr.4: 15008546-15008546 on Build GRCh38
rs762928600 Chr.4: 15008493-15008493 on Build GRCh38
rs558293988 Chr.4: 15008494-15008494 on Build GRCh38
rs376443106 Chr.4: 15008385-15008385 on Build GRCh38
rs892544070 Chr.4: 15008419-15008419 on Build GRCh38
rs536718134 Chr.4: 15008677-15008677 on Build GRCh38
rs547478894 Chr.4: 15008248-15008248 on Build GRCh38
rs756768908 Chr.4: 15008442-15008442 on Build GRCh38
rs1013143521 Chr.4: 15008406-15008406 on Build GRCh38
rs894770507 Chr.4: 15008636-15008636 on Build GRCh38
rs930355217 Chr.4: 15008543-15008543 on Build GRCh38
rs75021526 Chr.4: 15008489-15008489 on Build GRCh38
rs1011002516 Chr.4: 15008450-15008450 on Build GRCh38
rs1052570073 Chr.4: 15008672-15008672 on Build GRCh38
rs748231286 Chr.4: 15008357-15008357 on Build GRCh38
rs745466154 Chr.4: 15008272-15008272 on Build GRCh38
rs556836059 Chr.4: 15008448-15008448 on Build GRCh38
rs776924854 Chr.4: 15008388-15008388 on Build GRCh38
rs1031712574 Chr.4: 15008482-15008482 on Build GRCh38
rs1013592803 Chr.4: 15008645-15008645 on Build GRCh38
rs939188707 Chr.4: 15008269-15008269 on Build GRCh38
rs369684904 Chr.4: 15008461-15008461 on Build GRCh38
rs998333856 Chr.4: 15008395-15008395 on Build GRCh38
rs556605631 Chr.4: 15008619-15008619 on Build GRCh38
rs942922227 Chr.4: 15008313-15008313 on Build GRCh38
rs536403362 Chr.4: 15008446-15008446 on Build GRCh38
rs756354474 Chr.4: 15008530-15008531 on Build GRCh38
rs574687214 Chr.4: 15008678-15008678 on Build GRCh38
rs933067554 Chr.4: 15008263-15008263 on Build GRCh38
rs763694978 Chr.4: 15008411-15008411 on Build GRCh38
rs773760698 Chr.4: 15008369-15008369 on Build GRCh38
rs779176137 Chr.4: 15008470-15008470 on Build GRCh38
rs759918003 Chr.4: 15008405-15008405 on Build GRCh38
rs749096484 Chr.4: 15008288-15008288 on Build GRCh38
rs1017216098 Chr.4: 15008286-15008286 on Build GRCh38
rs372838374 Chr.4: 15008633-15008633 on Build GRCh38
rs1040464724 Chr.4: 15008631-15008631 on Build GRCh38
rs756045027 Chr.4: 15008455-15008455 on Build GRCh38
rs941762092 Chr.4: 15008564-15008564 on Build GRCh38
rs934263221 Chr.4: 15008665-15008665 on Build GRCh38
rs539300300 Chr.4: 15008492-15008492 on Build GRCh38
rs761153360 Chr.4: 15008380-15008380 on Build GRCh38
rs186083785 Chr.4: 15008588-15008588 on Build GRCh38
rs147887241 Chr.4: 15008277-15008277 on Build GRCh38

View Full Product Details