Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15469782-15470286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2A
Entrez Gene ID
57545
Gene Name
coiled-coil and C2 domain containing 2A
Gene Aliases
JBTS9, MKS6
UniGene
Hs.590928
Gene Chromosome Location
Chr.4: 15468660-15601971 on Build GRCh38
Transcripts
NM_001080522.2 NM_001164720.1 NM_020785.2 XM_005248177.1 XM_011513872.2 XM_011513874.2

SNP Detail

SNP ID Chromosome Location
rs920929600 Chr.4: 15469881-15469881 on Build GRCh38
rs534147397 Chr.4: 15470189-15470189 on Build GRCh38
rs145443888 Chr.4: 15469989-15469989 on Build GRCh38
rs6819598 Chr.4: 15470094-15470094 on Build GRCh38
rs764329163 Chr.4: 15469884-15469884 on Build GRCh38
rs879543998 Chr.4: 15469836-15469836 on Build GRCh38
rs781156446 Chr.4: 15470174-15470174 on Build GRCh38
rs989023559 Chr.4: 15470119-15470119 on Build GRCh38
rs746332512 Chr.4: 15470185-15470185 on Build GRCh38
rs554374624 Chr.4: 15470209-15470209 on Build GRCh38
rs1047684002 Chr.4: 15469965-15469965 on Build GRCh38
rs753631903 Chr.4: 15469891-15469891 on Build GRCh38
rs771006335 Chr.4: 15470110-15470110 on Build GRCh38
rs368395644 Chr.4: 15469920-15469920 on Build GRCh38
rs914717577 Chr.4: 15470156-15470156 on Build GRCh38
rs1051179602 Chr.4: 15470231-15470231 on Build GRCh38
rs911726071 Chr.4: 15469923-15469923 on Build GRCh38
rs970054011 Chr.4: 15470043-15470043 on Build GRCh38
rs1031335874 Chr.4: 15470146-15470146 on Build GRCh38
rs778112215 Chr.4: 15470029-15470029 on Build GRCh38
rs956301285 Chr.4: 15470112-15470112 on Build GRCh38
rs886450165 Chr.4: 15469987-15469987 on Build GRCh38
rs911673747 Chr.4: 15470224-15470224 on Build GRCh38
rs1022905055 Chr.4: 15470074-15470074 on Build GRCh38
rs1039351934 Chr.4: 15469944-15469944 on Build GRCh38
rs886059135 Chr.4: 15469898-15469898 on Build GRCh38
rs551669488 Chr.4: 15470087-15470087 on Build GRCh38
rs538142479 Chr.4: 15470040-15470040 on Build GRCh38
rs770432766 Chr.4: 15470212-15470212 on Build GRCh38
rs536387957 Chr.4: 15470226-15470226 on Build GRCh38
rs1043407714 Chr.4: 15469858-15469858 on Build GRCh38
rs934367736 Chr.4: 15469917-15469917 on Build GRCh38
rs572601157 Chr.4: 15470155-15470155 on Build GRCh38
rs775994292 Chr.4: 15470253-15470253 on Build GRCh38
rs890843272 Chr.4: 15469904-15469904 on Build GRCh38
rs1035104349 Chr.4: 15469994-15469994 on Build GRCh38
rs899051692 Chr.4: 15469964-15469964 on Build GRCh38
rs895298295 Chr.4: 15469998-15469998 on Build GRCh38
rs28463577 Chr.4: 15469808-15469808 on Build GRCh38
rs898447321 Chr.4: 15469930-15469930 on Build GRCh38
rs968982618 Chr.4: 15470167-15470167 on Build GRCh38
rs1050875313 Chr.4: 15469894-15469894 on Build GRCh38
rs1004071038 Chr.4: 15469906-15469906 on Build GRCh38
rs573640371 Chr.4: 15470211-15470211 on Build GRCh38
rs182259436 Chr.4: 15469907-15469907 on Build GRCh38

View Full Product Details