Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15478632-15479130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2A
Entrez Gene ID
57545
Gene Name
coiled-coil and C2 domain containing 2A
Gene Aliases
JBTS9, MKS6
UniGene
Hs.590928
Gene Chromosome Location
Chr.4: 15468660-15601971 on Build GRCh38
Transcripts
NM_001080522.2 NM_001164720.1 NM_020785.2 XM_005248177.1 XM_011513872.2 XM_011513874.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5818790 c.65C>A p.A22E Chr.4: 15478748-15478748 on Build GRCh38
5818789 c.65C>A p.A22E Chr.4: 15478748-15478748 on Build GRCh38
1594315 c.117G>T p.K39N Chr.4: 15478800-15478800 on Build GRCh38
1052279 c.100C>T p.R34* Chr.4: 15478783-15478783 on Build GRCh38
5528181 c.45C>T p.F15F Chr.4: 15478728-15478728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868073383 Chr.4: 15478864-15478864 on Build GRCh38
rs368612494 Chr.4: 15478838-15478838 on Build GRCh38
rs562505018 Chr.4: 15478969-15478969 on Build GRCh38
rs755345164 Chr.4: 15478738-15478738 on Build GRCh38
rs754222479 Chr.4: 15478698-15478698 on Build GRCh38
rs768395829 Chr.4: 15478822-15478822 on Build GRCh38
rs1004695681 Chr.4: 15479006-15479006 on Build GRCh38
rs886044284 Chr.4: 15478791-15478791 on Build GRCh38
rs758421113 Chr.4: 15478998-15478998 on Build GRCh38
rs80245100 Chr.4: 15479041-15479041 on Build GRCh38
rs530243571 Chr.4: 15478773-15478773 on Build GRCh38
rs1034116587 Chr.4: 15478829-15478829 on Build GRCh38
rs575647788 Chr.4: 15478811-15478811 on Build GRCh38
rs946155022 Chr.4: 15478894-15478894 on Build GRCh38
rs745409806 Chr.4: 15478780-15478780 on Build GRCh38
rs993841192 Chr.4: 15478787-15478787 on Build GRCh38
rs888123115 Chr.4: 15479029-15479029 on Build GRCh38
rs944854184 Chr.4: 15479009-15479009 on Build GRCh38
rs751235309 Chr.4: 15478765-15478765 on Build GRCh38
rs960925783 Chr.4: 15478826-15478827 on Build GRCh38
rs780506478 Chr.4: 15478774-15478774 on Build GRCh38
rs531616003 Chr.4: 15478977-15478977 on Build GRCh38
rs912791690 Chr.4: 15478934-15478934 on Build GRCh38
rs988729891 Chr.4: 15478933-15478933 on Build GRCh38
rs748252802 Chr.4: 15478834-15478834 on Build GRCh38
rs1032400794 Chr.4: 15479099-15479099 on Build GRCh38
rs565551298 Chr.4: 15479019-15479019 on Build GRCh38
rs767024593 Chr.4: 15478689-15478689 on Build GRCh38
rs1023647045 Chr.4: 15478840-15478842 on Build GRCh38
rs13353624 Chr.4: 15478858-15478858 on Build GRCh38
rs1020101511 Chr.4: 15479098-15479098 on Build GRCh38
rs772353449 Chr.4: 15478835-15478835 on Build GRCh38
rs1039983196 Chr.4: 15478948-15478948 on Build GRCh38
rs1048034941 Chr.4: 15479025-15479025 on Build GRCh38
rs16892028 Chr.4: 15479049-15479049 on Build GRCh38
rs778593083 Chr.4: 15478857-15478857 on Build GRCh38
rs756008877 Chr.4: 15478784-15478784 on Build GRCh38
rs956925810 Chr.4: 15478910-15478910 on Build GRCh38
rs146065951 Chr.4: 15478938-15478938 on Build GRCh38
rs980445355 Chr.4: 15478826-15478826 on Build GRCh38
rs780010340 Chr.4: 15478802-15478802 on Build GRCh38
rs979549049 Chr.4: 15478867-15478867 on Build GRCh38
rs749965862 Chr.4: 15478792-15478792 on Build GRCh38
rs932225717 Chr.4: 15478947-15478947 on Build GRCh38
rs183908060 Chr.4: 15478879-15478879 on Build GRCh38
rs529766543 Chr.4: 15479090-15479090 on Build GRCh38
rs542717294 Chr.4: 15478939-15478939 on Build GRCh38
rs887657124 Chr.4: 15478677-15478677 on Build GRCh38
rs959984118 Chr.4: 15479062-15479062 on Build GRCh38
rs545271288 Chr.4: 15478989-15478989 on Build GRCh38
rs763786765 Chr.4: 15478753-15478753 on Build GRCh38
rs547389759 Chr.4: 15478818-15478818 on Build GRCh38
rs978769259 Chr.4: 15478905-15478905 on Build GRCh38
rs931730321 Chr.4: 15478978-15478978 on Build GRCh38
rs896947430 Chr.4: 15478783-15478783 on Build GRCh38
rs753264367 Chr.4: 15478801-15478801 on Build GRCh38
rs1036568281 Chr.4: 15479011-15479011 on Build GRCh38
rs769889016 Chr.4: 15479108-15479108 on Build GRCh38
rs1048872800 Chr.4: 15478672-15478672 on Build GRCh38
rs139952452 Chr.4: 15479024-15479024 on Build GRCh38
rs751479717 Chr.4: 15479052-15479052 on Build GRCh38
rs142821756 Chr.4: 15478825-15478825 on Build GRCh38
rs546369496 Chr.4: 15478702-15478702 on Build GRCh38
rs754933522 Chr.4: 15479063-15479063 on Build GRCh38

View Full Product Details