Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17634752-17635251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184B
Entrez Gene ID
27146
Gene Name
family with sequence similarity 184 member B
Gene Aliases
-
UniGene
Hs.744951
Gene Chromosome Location
Chr.4: 17632073-17781512 on Build GRCh38
Transcripts
NM_015688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3337086 c.2886G>T p.M962I Chr.4: 17635012-17635012 on Build GRCh38
3337092 c.2822C>T p.A941V Chr.4: 17635076-17635076 on Build GRCh38
5975061 c.2865G>T p.P955P Chr.4: 17635033-17635033 on Build GRCh38
5400880 c.2785G>A p.E929K Chr.4: 17635113-17635113 on Build GRCh38
5627535 c.2847C>G p.F949L Chr.4: 17635051-17635051 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556556250 Chr.4: 17634920-17634920 on Build GRCh38
rs372390501 Chr.4: 17635120-17635120 on Build GRCh38
rs979493688 Chr.4: 17634979-17634979 on Build GRCh38
rs756465598 Chr.4: 17634983-17634983 on Build GRCh38
rs763203774 Chr.4: 17635080-17635080 on Build GRCh38
rs376247796 Chr.4: 17634894-17634895 on Build GRCh38
rs1021535255 Chr.4: 17635099-17635099 on Build GRCh38
rs143581443 Chr.4: 17635064-17635064 on Build GRCh38
rs921253019 Chr.4: 17634984-17634984 on Build GRCh38
rs569967878 Chr.4: 17634927-17634927 on Build GRCh38
rs775922853 Chr.4: 17635078-17635078 on Build GRCh38
rs1044091595 Chr.4: 17635069-17635069 on Build GRCh38
rs757968455 Chr.4: 17635033-17635033 on Build GRCh38
rs938448203 Chr.4: 17635091-17635091 on Build GRCh38
rs868086463 Chr.4: 17634932-17634932 on Build GRCh38
rs371383137 Chr.4: 17634959-17634959 on Build GRCh38
rs919888303 Chr.4: 17635063-17635063 on Build GRCh38
rs527882445 Chr.4: 17634787-17634787 on Build GRCh38
rs749576057 Chr.4: 17635102-17635104 on Build GRCh38
rs375853416 Chr.4: 17634893-17634894 on Build GRCh38
rs986883449 Chr.4: 17635164-17635164 on Build GRCh38
rs983087124 Chr.4: 17635202-17635202 on Build GRCh38
rs1011252914 Chr.4: 17634779-17634779 on Build GRCh38
rs565245907 Chr.4: 17635171-17635171 on Build GRCh38
rs753042649 Chr.4: 17635058-17635058 on Build GRCh38
rs1035169360 Chr.4: 17634916-17634916 on Build GRCh38
rs56911690 Chr.4: 17634900-17634903 on Build GRCh38
rs1001499153 Chr.4: 17635122-17635122 on Build GRCh38
rs781639950 Chr.4: 17634970-17634970 on Build GRCh38
rs921400495 Chr.4: 17634793-17634793 on Build GRCh38
rs76288358 Chr.4: 17634939-17634939 on Build GRCh38
rs567109446 Chr.4: 17634897-17634900 on Build GRCh38
rs757785061 Chr.4: 17634960-17634960 on Build GRCh38
rs369732253 Chr.4: 17634857-17634857 on Build GRCh38
rs986852222 Chr.4: 17635029-17635029 on Build GRCh38
rs61745212 Chr.4: 17635015-17635015 on Build GRCh38
rs961444950 Chr.4: 17635181-17635181 on Build GRCh38
rs140407107 Chr.4: 17634952-17634952 on Build GRCh38
rs778477202 Chr.4: 17635000-17635000 on Build GRCh38
rs755335607 Chr.4: 17635223-17635223 on Build GRCh38
rs890346507 Chr.4: 17635097-17635097 on Build GRCh38
rs954700882 Chr.4: 17634917-17634918 on Build GRCh38
rs762677740 Chr.4: 17635016-17635016 on Build GRCh38
rs894180726 Chr.4: 17634776-17634776 on Build GRCh38
rs912373588 Chr.4: 17634836-17634836 on Build GRCh38
rs780919796 Chr.4: 17635048-17635051 on Build GRCh38
rs186792009 Chr.4: 17634974-17634974 on Build GRCh38
rs16895365 Chr.4: 17635043-17635043 on Build GRCh38
rs190054948 Chr.4: 17635093-17635093 on Build GRCh38
rs200401027 Chr.4: 17634881-17634882 on Build GRCh38
rs3214318 Chr.4: 17634884-17634885 on Build GRCh38
rs376784221 Chr.4: 17634894-17634894 on Build GRCh38
rs541800438 Chr.4: 17634977-17634977 on Build GRCh38
rs376901720 Chr.4: 17635092-17635092 on Build GRCh38
rs527975115 Chr.4: 17635215-17635215 on Build GRCh38
rs905502745 Chr.4: 17635073-17635073 on Build GRCh38
rs182747127 Chr.4: 17635220-17635220 on Build GRCh38
rs763605381 Chr.4: 17635034-17635034 on Build GRCh38
rs995220722 Chr.4: 17634863-17634863 on Build GRCh38
rs1007810723 Chr.4: 17635105-17635106 on Build GRCh38
rs182144411 Chr.4: 17634858-17634858 on Build GRCh38
rs1017368294 Chr.4: 17635113-17635113 on Build GRCh38
rs972589858 Chr.4: 17635209-17635209 on Build GRCh38
rs954147919 Chr.4: 17635147-17635147 on Build GRCh38
rs77210007 Chr.4: 17634895-17634895 on Build GRCh38
rs372762782 Chr.4: 17635036-17635036 on Build GRCh38
rs55751748 Chr.4: 17634915-17634915 on Build GRCh38
rs964110106 Chr.4: 17635117-17635117 on Build GRCh38
rs567467695 Chr.4: 17634870-17634870 on Build GRCh38
rs750917547 Chr.4: 17635134-17635134 on Build GRCh38
rs905640911 Chr.4: 17634898-17634898 on Build GRCh38
rs796702267 Chr.4: 17634885-17634885 on Build GRCh38
rs932721818 Chr.4: 17634821-17634821 on Build GRCh38
rs111632471 Chr.4: 17635096-17635096 on Build GRCh38
rs1050937397 Chr.4: 17634830-17634830 on Build GRCh38
rs769619782 Chr.4: 17635074-17635074 on Build GRCh38
rs767946017 Chr.4: 17635129-17635129 on Build GRCh38
rs939930426 Chr.4: 17634842-17634842 on Build GRCh38

View Full Product Details