Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 17647655-17648185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM184B
Entrez Gene ID
27146
Gene Name
family with sequence similarity 184 member B
Gene Aliases
-
UniGene
Hs.744951
Gene Chromosome Location
Chr.4: 17632073-17781512 on Build GRCh38
Transcripts
NM_015688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6864948 c.2192-1G>C p.? Chr.4: 17647792-17647792 on Build GRCh38
4536640 c.2326G>A p.D776N Chr.4: 17647657-17647657 on Build GRCh38
6405350 c.2253G>T p.L751F Chr.4: 17647730-17647730 on Build GRCh38
6243296 c.2231G>A p.C744Y Chr.4: 17647752-17647752 on Build GRCh38
4796068 c.2229C>T p.A743A Chr.4: 17647754-17647754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055476227 Chr.4: 17648053-17648053 on Build GRCh38
rs772981465 Chr.4: 17647770-17647770 on Build GRCh38
rs910778432 Chr.4: 17647879-17647879 on Build GRCh38
rs770249260 Chr.4: 17647719-17647719 on Build GRCh38
rs977505364 Chr.4: 17648155-17648155 on Build GRCh38
rs761424699 Chr.4: 17647763-17647763 on Build GRCh38
rs909802313 Chr.4: 17648007-17648007 on Build GRCh38
rs775834250 Chr.4: 17647748-17647748 on Build GRCh38
rs1033038705 Chr.4: 17647801-17647801 on Build GRCh38
rs370717195 Chr.4: 17647783-17647783 on Build GRCh38
rs1053937303 Chr.4: 17648138-17648138 on Build GRCh38
rs1000117444 Chr.4: 17648096-17648096 on Build GRCh38
rs868773578 Chr.4: 17648019-17648019 on Build GRCh38
rs41268401 Chr.4: 17647906-17647906 on Build GRCh38
rs542919139 Chr.4: 17647738-17647738 on Build GRCh38
rs749715642 Chr.4: 17647754-17647754 on Build GRCh38
rs546637316 Chr.4: 17647872-17647872 on Build GRCh38
rs115241247 Chr.4: 17648060-17648060 on Build GRCh38
rs993254729 Chr.4: 17647928-17647928 on Build GRCh38
rs966910788 Chr.4: 17648148-17648148 on Build GRCh38
rs376068083 Chr.4: 17647714-17647714 on Build GRCh38
rs543998326 Chr.4: 17647854-17647854 on Build GRCh38
rs769455279 Chr.4: 17647802-17647802 on Build GRCh38
rs537976429 Chr.4: 17647954-17647954 on Build GRCh38
rs568762232 Chr.4: 17648064-17648064 on Build GRCh38
rs6813374 Chr.4: 17647871-17647871 on Build GRCh38
rs918675042 Chr.4: 17647933-17647933 on Build GRCh38
rs530656827 Chr.4: 17647769-17647769 on Build GRCh38
rs537688667 Chr.4: 17648068-17648068 on Build GRCh38
rs923273367 Chr.4: 17647874-17647874 on Build GRCh38
rs951312984 Chr.4: 17647964-17647964 on Build GRCh38
rs776230014 Chr.4: 17647836-17647836 on Build GRCh38
rs1033519348 Chr.4: 17647705-17647705 on Build GRCh38
rs942221741 Chr.4: 17647891-17647891 on Build GRCh38
rs769377412 Chr.4: 17647918-17647918 on Build GRCh38
rs563866496 Chr.4: 17647858-17647858 on Build GRCh38
rs1030396501 Chr.4: 17648160-17648160 on Build GRCh38
rs6813375 Chr.4: 17647876-17647876 on Build GRCh38
rs1046097882 Chr.4: 17648011-17648011 on Build GRCh38
rs761310589 Chr.4: 17648121-17648121 on Build GRCh38
rs983956925 Chr.4: 17647986-17647986 on Build GRCh38
rs906212484 Chr.4: 17648017-17648017 on Build GRCh38
rs747713164 Chr.4: 17647718-17647719 on Build GRCh38
rs894054433 Chr.4: 17648140-17648140 on Build GRCh38
rs533776342 Chr.4: 17647787-17647787 on Build GRCh38
rs937250836 Chr.4: 17648027-17648027 on Build GRCh38
rs1001803074 Chr.4: 17648024-17648024 on Build GRCh38
rs902721035 Chr.4: 17647772-17647772 on Build GRCh38
rs902921370 Chr.4: 17648089-17648089 on Build GRCh38
rs937031659 Chr.4: 17647716-17647716 on Build GRCh38
rs776945124 Chr.4: 17647802-17647803 on Build GRCh38
rs1055438812 Chr.4: 17647756-17647756 on Build GRCh38

View Full Product Details