Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25929458-25929956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM20
Entrez Gene ID
389203
Gene Name
small integral membrane protein 20
Gene Aliases
C4orf52
UniGene
Hs.479386
Gene Chromosome Location
Chr.4: 25914192-25929879 on Build GRCh38
Transcripts
NM_001145432.1

SNP Detail

SNP ID Chromosome Location
rs148941272 Chr.4: 25929737-25929737 on Build GRCh38
rs1046712427 Chr.4: 25929924-25929924 on Build GRCh38
rs191654144 Chr.4: 25929914-25929914 on Build GRCh38
rs760046813 Chr.4: 25929613-25929613 on Build GRCh38
rs554 Chr.4: 25929703-25929703 on Build GRCh38
rs541949609 Chr.4: 25929677-25929677 on Build GRCh38
rs566984489 Chr.4: 25929494-25929494 on Build GRCh38
rs1000977907 Chr.4: 25929918-25929918 on Build GRCh38
rs942559386 Chr.4: 25929891-25929891 on Build GRCh38
rs983246581 Chr.4: 25929867-25929867 on Build GRCh38
rs929121794 Chr.4: 25929556-25929556 on Build GRCh38
rs1006830065 Chr.4: 25929557-25929557 on Build GRCh38
rs931090050 Chr.4: 25929821-25929821 on Build GRCh38
rs887521367 Chr.4: 25929600-25929600 on Build GRCh38
rs972413485 Chr.4: 25929812-25929812 on Build GRCh38
rs556128418 Chr.4: 25929546-25929546 on Build GRCh38
rs941757170 Chr.4: 25929897-25929897 on Build GRCh38
rs975995125 Chr.4: 25929765-25929765 on Build GRCh38
rs895395203 Chr.4: 25929642-25929642 on Build GRCh38
rs923178702 Chr.4: 25929796-25929796 on Build GRCh38
rs533668535 Chr.4: 25929865-25929865 on Build GRCh38
rs11538095 Chr.4: 25929523-25929523 on Build GRCh38
rs1020981478 Chr.4: 25929911-25929911 on Build GRCh38
rs943264745 Chr.4: 25929497-25929497 on Build GRCh38
rs1048679694 Chr.4: 25929871-25929871 on Build GRCh38
rs143814445 Chr.4: 25929919-25929919 on Build GRCh38
rs535598157 Chr.4: 25929512-25929512 on Build GRCh38
rs999683214 Chr.4: 25929690-25929690 on Build GRCh38
rs145680612 Chr.4: 25929731-25929731 on Build GRCh38
rs564738023 Chr.4: 25929739-25929739 on Build GRCh38
rs773264844 Chr.4: 25929819-25929819 on Build GRCh38
rs762813320 Chr.4: 25929892-25929892 on Build GRCh38
rs576740812 Chr.4: 25929500-25929500 on Build GRCh38
rs997549730 Chr.4: 25929643-25929643 on Build GRCh38
rs901127219 Chr.4: 25929906-25929906 on Build GRCh38
rs1017577052 Chr.4: 25929579-25929579 on Build GRCh38
rs889671862 Chr.4: 25929536-25929536 on Build GRCh38
rs115907771 Chr.4: 25929872-25929872 on Build GRCh38
rs555863311 Chr.4: 25929587-25929587 on Build GRCh38
rs112349939 Chr.4: 25929730-25929730 on Build GRCh38
rs772559539 Chr.4: 25929632-25929632 on Build GRCh38
rs1048252067 Chr.4: 25929517-25929517 on Build GRCh38
rs910460541 Chr.4: 25929495-25929495 on Build GRCh38
rs1021453453 Chr.4: 25929653-25929653 on Build GRCh38
rs187542423 Chr.4: 25929676-25929676 on Build GRCh38
rs112149527 Chr.4: 25929787-25929787 on Build GRCh38
rs901850330 Chr.4: 25929633-25929633 on Build GRCh38
rs182862544 Chr.4: 25929515-25929515 on Build GRCh38
rs767656672 Chr.4: 25929907-25929907 on Build GRCh38
rs1032465720 Chr.4: 25929697-25929697 on Build GRCh38
rs941731901 Chr.4: 25929609-25929609 on Build GRCh38

View Full Product Details