Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 26490076-26490583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKAR
Entrez Gene ID
886
Gene Name
cholecystokinin A receptor
Gene Aliases
CCK-A, CCK1-R, CCK1R, CCKRA
UniGene
Hs.129
Gene Chromosome Location
Chr.4: 26481396-26490420 on Build GRCh38
Transcripts
NM_000730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829108 c.77C>T p.T26M Chr.4: 26490191-26490191 on Build GRCh38
1694227 c.46_47CC>TT p.P16F Chr.4: 26490221-26490222 on Build GRCh38
4913075 c.51C>T p.P17P Chr.4: 26490217-26490217 on Build GRCh38
6100133 c.60C>T p.L20L Chr.4: 26490208-26490208 on Build GRCh38
6482820 c.23T>C p.L8P Chr.4: 26490245-26490245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868034108 Chr.4: 26490201-26490201 on Build GRCh38
rs537322039 Chr.4: 26490267-26490267 on Build GRCh38
rs936912521 Chr.4: 26490273-26490273 on Build GRCh38
rs759109607 Chr.4: 26490242-26490242 on Build GRCh38
rs868528853 Chr.4: 26490519-26490519 on Build GRCh38
rs1016535607 Chr.4: 26490405-26490406 on Build GRCh38
rs200494718 Chr.4: 26490159-26490159 on Build GRCh38
rs747058691 Chr.4: 26490192-26490192 on Build GRCh38
rs115646101 Chr.4: 26490531-26490531 on Build GRCh38
rs781117941 Chr.4: 26490275-26490275 on Build GRCh38
rs201856546 Chr.4: 26490245-26490245 on Build GRCh38
rs6448456 Chr.4: 26490352-26490352 on Build GRCh38
rs942620036 Chr.4: 26490556-26490556 on Build GRCh38
rs368292493 Chr.4: 26490117-26490117 on Build GRCh38
rs1005510234 Chr.4: 26490399-26490399 on Build GRCh38
rs202031212 Chr.4: 26490196-26490196 on Build GRCh38
rs770111352 Chr.4: 26490200-26490200 on Build GRCh38
rs762243830 Chr.4: 26490137-26490139 on Build GRCh38
rs746846439 Chr.4: 26490375-26490375 on Build GRCh38
rs550581286 Chr.4: 26490147-26490147 on Build GRCh38
rs893590061 Chr.4: 26490260-26490260 on Build GRCh38
rs935017299 Chr.4: 26490502-26490502 on Build GRCh38
rs536379349 Chr.4: 26490168-26490168 on Build GRCh38
rs200917200 Chr.4: 26490232-26490232 on Build GRCh38
rs545284669 Chr.4: 26490354-26490354 on Build GRCh38
rs757826943 Chr.4: 26490270-26490270 on Build GRCh38
rs867332155 Chr.4: 26490417-26490417 on Build GRCh38
rs373086084 Chr.4: 26490474-26490474 on Build GRCh38
rs753405191 Chr.4: 26490166-26490166 on Build GRCh38
rs181156950 Chr.4: 26490396-26490396 on Build GRCh38
rs760707904 Chr.4: 26490228-26490228 on Build GRCh38
rs201311032 Chr.4: 26490408-26490408 on Build GRCh38
rs773378192 Chr.4: 26490153-26490154 on Build GRCh38
rs768485230 Chr.4: 26490465-26490465 on Build GRCh38
rs765785446 Chr.4: 26490229-26490229 on Build GRCh38
rs753450499 Chr.4: 26490239-26490239 on Build GRCh38
rs567297961 Chr.4: 26490149-26490149 on Build GRCh38
rs201796699 Chr.4: 26490291-26490291 on Build GRCh38
rs778435579 Chr.4: 26490182-26490182 on Build GRCh38
rs1799723 Chr.4: 26490553-26490553 on Build GRCh38
rs753055327 Chr.4: 26490252-26490252 on Build GRCh38
rs200177038 Chr.4: 26490368-26490368 on Build GRCh38
rs936692870 Chr.4: 26490516-26490516 on Build GRCh38
rs758853459 Chr.4: 26490255-26490255 on Build GRCh38
rs116476903 Chr.4: 26490209-26490209 on Build GRCh38
rs560986896 Chr.4: 26490419-26490419 on Build GRCh38
rs115609834 Chr.4: 26490208-26490208 on Build GRCh38
rs746055437 Chr.4: 26490198-26490198 on Build GRCh38
rs1046542169 Chr.4: 26490321-26490321 on Build GRCh38
rs906293897 Chr.4: 26490324-26490324 on Build GRCh38
rs201586028 Chr.4: 26490342-26490342 on Build GRCh38
rs751976222 Chr.4: 26490262-26490262 on Build GRCh38
rs199975181 Chr.4: 26490253-26490253 on Build GRCh38
rs778382078 Chr.4: 26490257-26490257 on Build GRCh38
rs747900043 Chr.4: 26490290-26490290 on Build GRCh38
rs1800856 Chr.4: 26490146-26490146 on Build GRCh38
rs104893833 Chr.4: 26490207-26490207 on Build GRCh38
rs186654279 Chr.4: 26490508-26490508 on Build GRCh38
rs746025462 Chr.4: 26490278-26490278 on Build GRCh38
rs774176022 Chr.4: 26490120-26490120 on Build GRCh38
rs376084549 Chr.4: 26490532-26490532 on Build GRCh38
rs774369395 Chr.4: 26490267-26490267 on Build GRCh38
rs761722603 Chr.4: 26490142-26490142 on Build GRCh38
rs963980643 Chr.4: 26490412-26490412 on Build GRCh38
rs948308484 Chr.4: 26490546-26490546 on Build GRCh38
rs200910008 Chr.4: 26490110-26490110 on Build GRCh38
rs989470666 Chr.4: 26490517-26490517 on Build GRCh38
rs954970398 Chr.4: 26490487-26490487 on Build GRCh38
rs377488910 Chr.4: 26490322-26490322 on Build GRCh38
rs906669428 Chr.4: 26490558-26490558 on Build GRCh38
rs61730704 Chr.4: 26490171-26490171 on Build GRCh38
rs200849488 Chr.4: 26490279-26490279 on Build GRCh38
rs199694452 Chr.4: 26490458-26490458 on Build GRCh38
rs36224751 Chr.4: 26490482-26490482 on Build GRCh38
rs201179899 Chr.4: 26490301-26490301 on Build GRCh38
rs756797239 Chr.4: 26490131-26490131 on Build GRCh38
rs776759777 Chr.4: 26490490-26490490 on Build GRCh38
rs1039106402 Chr.4: 26490313-26490313 on Build GRCh38
rs201415344 Chr.4: 26490119-26490119 on Build GRCh38
rs758910995 Chr.4: 26490173-26490173 on Build GRCh38
rs149993603 Chr.4: 26490195-26490195 on Build GRCh38
rs754709140 Chr.4: 26490167-26490167 on Build GRCh38
rs1025273389 Chr.4: 26490341-26490341 on Build GRCh38
rs780266352 Chr.4: 26490287-26490287 on Build GRCh38
rs202039613 Chr.4: 26490319-26490319 on Build GRCh38
rs200488470 Chr.4: 26490335-26490335 on Build GRCh38
rs190188968 Chr.4: 26490384-26490384 on Build GRCh38
rs773166071 Chr.4: 26490227-26490227 on Build GRCh38
rs200119191 Chr.4: 26490292-26490292 on Build GRCh38
rs41267459 Chr.4: 26490304-26490304 on Build GRCh38

View Full Product Details