Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37590787-37591297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf19
Entrez Gene ID
55286
Gene Name
chromosome 4 open reading frame 19
Gene Aliases
-
UniGene
Hs.107527
Gene Chromosome Location
Chr.4: 37453454-37596146 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1 NM_001085399.1

Gene Symbol
RELL1
Entrez Gene ID
768211
Gene Name
RELT like 1
Gene Aliases
-
UniGene
Hs.283378
Gene Chromosome Location
Chr.4: 37590800-37686377 on Build GRCh38
Transcripts
NM_001104629.1 NM_018302.2 XM_011513712.2 XM_011513713.2 XM_017008361.1 NM_001085399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6212084 c.867C>T p.D289D Chr.4: 37590922-37590922 on Build GRCh38
5975121 c.843_844insGATGGG p.G285_E286insDG Chr.4: 37590898-37590899 on Build GRCh38
4124490 c.737T>G p.V246G Chr.4: 37590792-37590792 on Build GRCh38
1738985 c.820G>A p.E274K Chr.4: 37590875-37590875 on Build GRCh38
7166767 c.847G>T p.G283W Chr.4: 37590902-37590902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757918394 Chr.4: 37591040-37591040 on Build GRCh38
rs576754765 Chr.4: 37591049-37591049 on Build GRCh38
rs888990468 Chr.4: 37590944-37590944 on Build GRCh38
rs565817662 Chr.4: 37591194-37591194 on Build GRCh38
rs764782712 Chr.4: 37591019-37591019 on Build GRCh38
rs917875165 Chr.4: 37590855-37590855 on Build GRCh38
rs754531707 Chr.4: 37590906-37590906 on Build GRCh38
rs765538593 Chr.4: 37590868-37590868 on Build GRCh38
rs777508193 Chr.4: 37591045-37591045 on Build GRCh38
rs1035876010 Chr.4: 37591245-37591245 on Build GRCh38
rs1018658826 Chr.4: 37590963-37590963 on Build GRCh38
rs139186850 Chr.4: 37590892-37590892 on Build GRCh38
rs754478384 Chr.4: 37591017-37591017 on Build GRCh38
rs188080064 Chr.4: 37590832-37590832 on Build GRCh38
rs146200909 Chr.4: 37590833-37590833 on Build GRCh38
rs770068930 Chr.4: 37590971-37590971 on Build GRCh38
rs771494998 Chr.4: 37590853-37590853 on Build GRCh38
rs551119350 Chr.4: 37590850-37590850 on Build GRCh38
rs763030177 Chr.4: 37590981-37590981 on Build GRCh38
rs777330778 Chr.4: 37590939-37590939 on Build GRCh38
rs918025065 Chr.4: 37591217-37591217 on Build GRCh38
rs372617352 Chr.4: 37590968-37590968 on Build GRCh38
rs943276610 Chr.4: 37590936-37590936 on Build GRCh38
rs778505316 Chr.4: 37590917-37590917 on Build GRCh38
rs775845020 Chr.4: 37590977-37590977 on Build GRCh38
rs3733500 Chr.4: 37590876-37590876 on Build GRCh38
rs959536058 Chr.4: 37591099-37591099 on Build GRCh38
rs745566095 Chr.4: 37590828-37590829 on Build GRCh38
rs747862182 Chr.4: 37590919-37590919 on Build GRCh38
rs757740018 Chr.4: 37591046-37591046 on Build GRCh38
rs772385827 Chr.4: 37590848-37590848 on Build GRCh38
rs367961468 Chr.4: 37590861-37590861 on Build GRCh38
rs764383387 Chr.4: 37590997-37590997 on Build GRCh38
rs368684445 Chr.4: 37590946-37590946 on Build GRCh38
rs752262990 Chr.4: 37591022-37591022 on Build GRCh38
rs778550533 Chr.4: 37590843-37590843 on Build GRCh38
rs759697697 Chr.4: 37590864-37590864 on Build GRCh38
rs937942 Chr.4: 37591057-37591057 on Build GRCh38
rs375131188 Chr.4: 37590933-37590933 on Build GRCh38
rs536702365 Chr.4: 37591038-37591038 on Build GRCh38
rs372748082 Chr.4: 37590922-37590922 on Build GRCh38
rs753032474 Chr.4: 37590871-37590871 on Build GRCh38
rs140013528 Chr.4: 37591047-37591047 on Build GRCh38
rs77303565 Chr.4: 37590940-37590940 on Build GRCh38
rs746438123 Chr.4: 37591048-37591048 on Build GRCh38
rs773740387 Chr.4: 37590849-37590849 on Build GRCh38
rs969774206 Chr.4: 37591073-37591073 on Build GRCh38
rs761635338 Chr.4: 37590873-37590873 on Build GRCh38
rs548436171 Chr.4: 37590825-37590825 on Build GRCh38
rs779998208 Chr.4: 37590894-37590894 on Build GRCh38
rs760541630 Chr.4: 37591006-37591006 on Build GRCh38
rs530995137 Chr.4: 37590846-37590846 on Build GRCh38
rs1043415626 Chr.4: 37590921-37590921 on Build GRCh38
rs539859782 Chr.4: 37590886-37590886 on Build GRCh38
rs766281698 Chr.4: 37591015-37591015 on Build GRCh38
rs143272655 Chr.4: 37590826-37590826 on Build GRCh38
rs1036663313 Chr.4: 37590878-37590878 on Build GRCh38
rs992204446 Chr.4: 37591188-37591188 on Build GRCh38
rs937941 Chr.4: 37591052-37591052 on Build GRCh38
rs755626408 Chr.4: 37590829-37590829 on Build GRCh38
rs768230851 Chr.4: 37590840-37590840 on Build GRCh38
rs535638130 Chr.4: 37590934-37590934 on Build GRCh38
rs372063210 Chr.4: 37590895-37590895 on Build GRCh38
rs988952739 Chr.4: 37590885-37590885 on Build GRCh38
rs772842830 Chr.4: 37591005-37591005 on Build GRCh38
rs573212043 Chr.4: 37591186-37591186 on Build GRCh38
rs13150138 Chr.4: 37591016-37591016 on Build GRCh38

View Full Product Details