Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38905431-38905946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A1
Entrez Gene ID
92689
Gene Name
family with sequence similarity 114 member A1
Gene Aliases
Noxp20
UniGene
Hs.476517
Gene Chromosome Location
Chr.4: 38867733-38945744 on Build GRCh38
Transcripts
NM_138389.2 NR_033290.1 XM_005262672.2 XM_006714033.2 XM_017008838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303900 c.450G>A p.T150T Chr.4: 38905535-38905535 on Build GRCh38
3917732 c.474C>T p.A158A Chr.4: 38905559-38905559 on Build GRCh38
1485943 c.547G>A p.E183K Chr.4: 38905632-38905632 on Build GRCh38
6100028 c.580G>T p.A194S Chr.4: 38905784-38905784 on Build GRCh38
6658108 c.617G>A p.R206Q Chr.4: 38905821-38905821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769096503 Chr.4: 38905657-38905657 on Build GRCh38
rs1013821417 Chr.4: 38905559-38905559 on Build GRCh38
rs372801338 Chr.4: 38905869-38905869 on Build GRCh38
rs1055208615 Chr.4: 38905736-38905736 on Build GRCh38
rs763959361 Chr.4: 38905678-38905678 on Build GRCh38
rs569608422 Chr.4: 38905622-38905622 on Build GRCh38
rs146872442 Chr.4: 38905615-38905615 on Build GRCh38
rs757409156 Chr.4: 38905478-38905478 on Build GRCh38
rs775617833 Chr.4: 38905870-38905870 on Build GRCh38
rs747803554 Chr.4: 38905816-38905816 on Build GRCh38
rs750460933 Chr.4: 38905583-38905583 on Build GRCh38
rs763196324 Chr.4: 38905877-38905877 on Build GRCh38
rs936815275 Chr.4: 38905505-38905505 on Build GRCh38
rs751009377 Chr.4: 38905894-38905894 on Build GRCh38
rs773250651 Chr.4: 38905710-38905710 on Build GRCh38
rs200436144 Chr.4: 38905476-38905476 on Build GRCh38
rs11940275 Chr.4: 38905457-38905457 on Build GRCh38
rs554734104 Chr.4: 38905735-38905735 on Build GRCh38
rs766584561 Chr.4: 38905739-38905739 on Build GRCh38
rs762187696 Chr.4: 38905576-38905576 on Build GRCh38
rs377135625 Chr.4: 38905608-38905608 on Build GRCh38
rs751249984 Chr.4: 38905679-38905679 on Build GRCh38
rs772457949 Chr.4: 38905720-38905720 on Build GRCh38
rs144537370 Chr.4: 38905778-38905778 on Build GRCh38
rs375301582 Chr.4: 38905561-38905561 on Build GRCh38
rs747527647 Chr.4: 38905824-38905824 on Build GRCh38
rs767974839 Chr.4: 38905660-38905660 on Build GRCh38
rs765676345 Chr.4: 38905713-38905713 on Build GRCh38
rs199533608 Chr.4: 38905670-38905670 on Build GRCh38
rs779553902 Chr.4: 38905785-38905785 on Build GRCh38
rs776742199 Chr.4: 38905909-38905909 on Build GRCh38
rs751998871 Chr.4: 38905477-38905477 on Build GRCh38
rs764345278 Chr.4: 38905567-38905567 on Build GRCh38
rs199977746 Chr.4: 38905868-38905868 on Build GRCh38
rs139440415 Chr.4: 38905496-38905496 on Build GRCh38
rs767324729 Chr.4: 38905879-38905879 on Build GRCh38
rs965243657 Chr.4: 38905835-38905835 on Build GRCh38
rs111831236 Chr.4: 38905721-38905721 on Build GRCh38
rs745452381 Chr.4: 38905645-38905645 on Build GRCh38
rs142423470 Chr.4: 38905786-38905786 on Build GRCh38
rs770501756 Chr.4: 38905545-38905545 on Build GRCh38
rs1018110269 Chr.4: 38905826-38905826 on Build GRCh38
rs774048175 Chr.4: 38905717-38905717 on Build GRCh38
rs775950939 Chr.4: 38905553-38905553 on Build GRCh38
rs745785618 Chr.4: 38905684-38905684 on Build GRCh38
rs761221211 Chr.4: 38905733-38905733 on Build GRCh38
rs565049279 Chr.4: 38905500-38905500 on Build GRCh38
rs760885494 Chr.4: 38905666-38905666 on Build GRCh38
rs906021746 Chr.4: 38905702-38905702 on Build GRCh38
rs777277396 Chr.4: 38905861-38905861 on Build GRCh38
rs779535699 Chr.4: 38905610-38905610 on Build GRCh38
rs1027418475 Chr.4: 38905815-38905815 on Build GRCh38
rs781101600 Chr.4: 38905490-38905490 on Build GRCh38
rs139238894 Chr.4: 38905784-38905784 on Build GRCh38
rs754142280 Chr.4: 38905675-38905675 on Build GRCh38
rs139789863 Chr.4: 38905820-38905820 on Build GRCh38
rs976632788 Chr.4: 38905631-38905631 on Build GRCh38
rs752953862 Chr.4: 38905765-38905765 on Build GRCh38
rs778231131 Chr.4: 38905821-38905821 on Build GRCh38
rs202220961 Chr.4: 38905534-38905534 on Build GRCh38
rs937615570 Chr.4: 38905705-38905705 on Build GRCh38
rs187220962 Chr.4: 38905663-38905663 on Build GRCh38
rs7698138 Chr.4: 38905535-38905535 on Build GRCh38
rs1018143211 Chr.4: 38905629-38905629 on Build GRCh38
rs752148855 Chr.4: 38905572-38905572 on Build GRCh38
rs149310340 Chr.4: 38905612-38905612 on Build GRCh38
rs758847341 Chr.4: 38905489-38905489 on Build GRCh38
rs771485860 Chr.4: 38905459-38905459 on Build GRCh38
rs767658785 Chr.4: 38905580-38905580 on Build GRCh38
rs748518530 Chr.4: 38905718-38905718 on Build GRCh38
rs748910174 Chr.4: 38905652-38905652 on Build GRCh38
rs769373805 Chr.4: 38905560-38905560 on Build GRCh38
rs755103652 Chr.4: 38905677-38905677 on Build GRCh38
rs372382288 Chr.4: 38905754-38905754 on Build GRCh38
rs895384993 Chr.4: 38905547-38905547 on Build GRCh38
rs750392881 Chr.4: 38905780-38905780 on Build GRCh38
rs117500892 Chr.4: 38905566-38905566 on Build GRCh38
rs976663662 Chr.4: 38905823-38905823 on Build GRCh38
rs978433767 Chr.4: 38905907-38905907 on Build GRCh38
rs199550702 Chr.4: 38905638-38905638 on Build GRCh38
rs200345622 Chr.4: 38905776-38905776 on Build GRCh38
rs17501308 Chr.4: 38905601-38905601 on Build GRCh38
rs760610875 Chr.4: 38905892-38905892 on Build GRCh38
rs430296 Chr.4: 38905806-38905806 on Build GRCh38
rs750161596 Chr.4: 38905889-38905889 on Build GRCh38
rs543114574 Chr.4: 38905768-38905768 on Build GRCh38
rs372440711 Chr.4: 38905602-38905602 on Build GRCh38
rs775174600 Chr.4: 38905651-38905651 on Build GRCh38
rs1029962015 Chr.4: 38905843-38905843 on Build GRCh38
rs776790371 Chr.4: 38905753-38905753 on Build GRCh38
rs759204380 Chr.4: 38905557-38905557 on Build GRCh38
rs532030041 Chr.4: 38905623-38905623 on Build GRCh38
rs762386796 Chr.4: 38905659-38905659 on Build GRCh38
rs781168047 Chr.4: 38905683-38905683 on Build GRCh38
rs376696883 Chr.4: 38905681-38905681 on Build GRCh38
rs765447912 Chr.4: 38905764-38905764 on Build GRCh38
rs1037975282 Chr.4: 38905481-38905481 on Build GRCh38
rs906072638 Chr.4: 38905492-38905492 on Build GRCh38
rs755662903 Chr.4: 38905685-38905685 on Build GRCh38
rs371122372 Chr.4: 38905894-38905894 on Build GRCh38
rs369704603 Chr.4: 38905647-38905647 on Build GRCh38
rs774530038 Chr.4: 38905658-38905658 on Build GRCh38
rs750706085 Chr.4: 38905493-38905493 on Build GRCh38
rs754497110 Chr.4: 38905790-38905790 on Build GRCh38
rs527437436 Chr.4: 38905522-38905522 on Build GRCh38
rs768136264 Chr.4: 38905716-38905716 on Build GRCh38

View Full Product Details