Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39551254-39551793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM14
Entrez Gene ID
201895
Gene Name
small integral membrane protein 14
Gene Aliases
C4orf34
UniGene
Hs.205952
Gene Chromosome Location
Chr.4: 39547297-39638998 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

Gene Symbol
UGDH-AS1
Entrez Gene ID
100885776
Gene Name
UGDH antisense RNA 1
Gene Aliases
-
UniGene
Hs.640769
Gene Chromosome Location
Chr.4: 39527839-39594707 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

SNP Detail

SNP ID Chromosome Location
rs151294923 Chr.4: 39551586-39551586 on Build GRCh38
rs981160850 Chr.4: 39551648-39551648 on Build GRCh38
rs540038576 Chr.4: 39551662-39551662 on Build GRCh38
rs915577924 Chr.4: 39551607-39551607 on Build GRCh38
rs1019103791 Chr.4: 39551543-39551543 on Build GRCh38
rs538897153 Chr.4: 39551685-39551685 on Build GRCh38
rs548295795 Chr.4: 39551555-39551555 on Build GRCh38
rs953099895 Chr.4: 39551529-39551529 on Build GRCh38
rs968463067 Chr.4: 39551285-39551285 on Build GRCh38
rs11558915 Chr.4: 39551731-39551731 on Build GRCh38
rs902015249 Chr.4: 39551554-39551554 on Build GRCh38
rs992022674 Chr.4: 39551384-39551384 on Build GRCh38
rs762909615 Chr.4: 39551724-39551725 on Build GRCh38
rs1008334683 Chr.4: 39551404-39551404 on Build GRCh38
rs937894941 Chr.4: 39551378-39551378 on Build GRCh38
rs1028768144 Chr.4: 39551309-39551309 on Build GRCh38
rs947302651 Chr.4: 39551719-39551719 on Build GRCh38
rs930986254 Chr.4: 39551475-39551475 on Build GRCh38
rs142268159 Chr.4: 39551725-39551725 on Build GRCh38
rs1049726064 Chr.4: 39551732-39551732 on Build GRCh38
rs747446118 Chr.4: 39551520-39551520 on Build GRCh38
rs917748795 Chr.4: 39551397-39551397 on Build GRCh38
rs1037192033 Chr.4: 39551409-39551409 on Build GRCh38
rs928439790 Chr.4: 39551659-39551659 on Build GRCh38
rs926553533 Chr.4: 39551362-39551362 on Build GRCh38
rs542534310 Chr.4: 39551723-39551723 on Build GRCh38
rs917801039 Chr.4: 39551750-39551750 on Build GRCh38
rs934490934 Chr.4: 39551663-39551663 on Build GRCh38
rs11558914 Chr.4: 39551636-39551636 on Build GRCh38
rs954081071 Chr.4: 39551354-39551354 on Build GRCh38
rs1027236043 Chr.4: 39551506-39551506 on Build GRCh38
rs893037899 Chr.4: 39551679-39551679 on Build GRCh38
rs979430021 Chr.4: 39551314-39551314 on Build GRCh38
rs1011557572 Chr.4: 39551301-39551301 on Build GRCh38
rs1021472973 Chr.4: 39551623-39551623 on Build GRCh38
rs959312217 Chr.4: 39551705-39551705 on Build GRCh38
rs868172787 Chr.4: 39551392-39551392 on Build GRCh38
rs772298297 Chr.4: 39551441-39551441 on Build GRCh38
rs192740842 Chr.4: 39551436-39551436 on Build GRCh38
rs182845092 Chr.4: 39551453-39551453 on Build GRCh38
rs889388576 Chr.4: 39551500-39551500 on Build GRCh38
rs1053384631 Chr.4: 39551674-39551674 on Build GRCh38
rs142415916 Chr.4: 39551493-39551493 on Build GRCh38

View Full Product Details