Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40335696-40336172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA9
Entrez Gene ID
55584
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
HSA243342, NACHRA9
UniGene
Hs.272278
Gene Chromosome Location
Chr.4: 40335329-40355217 on Build GRCh38
Transcripts
NM_017581.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672275 c.120A>C p.E40D Chr.4: 40335882-40335882 on Build GRCh38
1055315 c.74C>T p.T25M Chr.4: 40335836-40335836 on Build GRCh38
4672274 c.113delT p.F39fs*18 Chr.4: 40335875-40335875 on Build GRCh38
3661086 c.146T>C p.V49A Chr.4: 40335908-40335908 on Build GRCh38
4672276 c.185T>A p.I62N Chr.4: 40335947-40335947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55893318 Chr.4: 40336062-40336062 on Build GRCh38
rs370590960 Chr.4: 40335898-40335898 on Build GRCh38
rs142750616 Chr.4: 40335756-40335756 on Build GRCh38
rs200975157 Chr.4: 40336005-40336005 on Build GRCh38
rs747156948 Chr.4: 40335783-40335784 on Build GRCh38
rs754372957 Chr.4: 40335797-40335797 on Build GRCh38
rs772302634 Chr.4: 40335804-40335804 on Build GRCh38
rs561034737 Chr.4: 40335861-40335861 on Build GRCh38
rs1053482281 Chr.4: 40336079-40336079 on Build GRCh38
rs774116058 Chr.4: 40335850-40335850 on Build GRCh38
rs765799684 Chr.4: 40335895-40335895 on Build GRCh38
rs768574003 Chr.4: 40335922-40335922 on Build GRCh38
rs146393051 Chr.4: 40335845-40335845 on Build GRCh38
rs567536994 Chr.4: 40335810-40335810 on Build GRCh38
rs895092744 Chr.4: 40335730-40335730 on Build GRCh38
rs749139955 Chr.4: 40335952-40335952 on Build GRCh38
rs146064767 Chr.4: 40335921-40335921 on Build GRCh38
rs144769301 Chr.4: 40335929-40335929 on Build GRCh38
rs868057484 Chr.4: 40335812-40335812 on Build GRCh38
rs371099700 Chr.4: 40335984-40335984 on Build GRCh38
rs771777799 Chr.4: 40335973-40335973 on Build GRCh38
rs575755652 Chr.4: 40335755-40335755 on Build GRCh38
rs766840746 Chr.4: 40335796-40335796 on Build GRCh38
rs921250153 Chr.4: 40335772-40335772 on Build GRCh38
rs865904212 Chr.4: 40335974-40335974 on Build GRCh38
rs1046296937 Chr.4: 40336056-40336056 on Build GRCh38
rs541218458 Chr.4: 40335837-40335837 on Build GRCh38
rs56171884 Chr.4: 40335960-40335960 on Build GRCh38
rs375832887 Chr.4: 40335950-40335950 on Build GRCh38
rs916424476 Chr.4: 40336026-40336026 on Build GRCh38
rs984395938 Chr.4: 40336070-40336070 on Build GRCh38
rs754205007 Chr.4: 40335884-40335884 on Build GRCh38
rs747174659 Chr.4: 40335827-40335827 on Build GRCh38
rs758485973 Chr.4: 40335901-40335901 on Build GRCh38
rs755476861 Chr.4: 40335798-40335798 on Build GRCh38
rs148907815 Chr.4: 40335852-40335852 on Build GRCh38
rs111256153 Chr.4: 40335807-40335807 on Build GRCh38
rs1055443856 Chr.4: 40335729-40335729 on Build GRCh38
rs143699405 Chr.4: 40335975-40335975 on Build GRCh38
rs931199581 Chr.4: 40335723-40335723 on Build GRCh38
rs10022491 Chr.4: 40335891-40335891 on Build GRCh38
rs1024616069 Chr.4: 40335744-40335744 on Build GRCh38
rs887313309 Chr.4: 40335871-40335871 on Build GRCh38
rs55974552 Chr.4: 40335951-40335951 on Build GRCh38
rs944081564 Chr.4: 40335739-40335739 on Build GRCh38
rs535138649 Chr.4: 40336073-40336073 on Build GRCh38
rs765462024 Chr.4: 40336016-40336016 on Build GRCh38
rs777975444 Chr.4: 40335825-40335825 on Build GRCh38
rs56299220 Chr.4: 40335989-40335989 on Build GRCh38
rs748025938 Chr.4: 40335964-40335964 on Build GRCh38
rs999240207 Chr.4: 40335757-40335757 on Build GRCh38
rs748278341 Chr.4: 40335813-40335813 on Build GRCh38
rs760450437 Chr.4: 40335981-40335981 on Build GRCh38
rs758369911 Chr.4: 40335814-40335814 on Build GRCh38
rs768269193 Chr.4: 40335956-40335956 on Build GRCh38
rs56159866 Chr.4: 40335927-40335927 on Build GRCh38
rs766887870 Chr.4: 40335876-40335876 on Build GRCh38
rs996733030 Chr.4: 40335944-40335944 on Build GRCh38
rs760401427 Chr.4: 40335864-40335864 on Build GRCh38
rs369167285 Chr.4: 40335793-40335793 on Build GRCh38
rs761683376 Chr.4: 40335966-40335966 on Build GRCh38
rs373239007 Chr.4: 40335836-40335836 on Build GRCh38
rs370290456 Chr.4: 40335853-40335853 on Build GRCh38
rs902191804 Chr.4: 40335906-40335906 on Build GRCh38
rs1054990916 Chr.4: 40335873-40335873 on Build GRCh38
rs920723769 Chr.4: 40335856-40335856 on Build GRCh38
rs1013187374 Chr.4: 40335740-40335740 on Build GRCh38
rs921298784 Chr.4: 40336061-40336061 on Build GRCh38
rs776506529 Chr.4: 40335952-40335953 on Build GRCh38
rs376206135 Chr.4: 40335843-40335843 on Build GRCh38
rs879474228 Chr.4: 40336077-40336077 on Build GRCh38
rs188109471 Chr.4: 40335902-40335902 on Build GRCh38
rs763028831 Chr.4: 40335986-40335986 on Build GRCh38
rs753303534 Chr.4: 40336018-40336018 on Build GRCh38
rs949266274 Chr.4: 40336045-40336045 on Build GRCh38

View Full Product Details