Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40337130-40337604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA9
Entrez Gene ID
55584
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
HSA243342, NACHRA9
UniGene
Hs.272278
Gene Chromosome Location
Chr.4: 40335329-40355217 on Build GRCh38
Transcripts
NM_017581.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1055316 c.274C>A p.L92I Chr.4: 40337273-40337273 on Build GRCh38
5760108 c.264C>T p.H88H Chr.4: 40337263-40337263 on Build GRCh38
6628749 c.261G>A p.W87* Chr.4: 40337260-40337260 on Build GRCh38
7309070 c.211-5T>C p.? Chr.4: 40337205-40337205 on Build GRCh38
733487 c.251G>A p.R84H Chr.4: 40337250-40337250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999695097 Chr.4: 40337485-40337485 on Build GRCh38
rs114307446 Chr.4: 40337269-40337269 on Build GRCh38
rs763444225 Chr.4: 40337165-40337165 on Build GRCh38
rs754864594 Chr.4: 40337185-40337185 on Build GRCh38
rs754042546 Chr.4: 40337323-40337323 on Build GRCh38
rs10024518 Chr.4: 40337286-40337286 on Build GRCh38
rs573286592 Chr.4: 40337518-40337518 on Build GRCh38
rs137868682 Chr.4: 40337262-40337262 on Build GRCh38
rs554984075 Chr.4: 40337477-40337477 on Build GRCh38
rs138628295 Chr.4: 40337491-40337491 on Build GRCh38
rs750557311 Chr.4: 40337184-40337184 on Build GRCh38
rs148584579 Chr.4: 40337332-40337332 on Build GRCh38
rs144004481 Chr.4: 40337357-40337357 on Build GRCh38
rs780801569 Chr.4: 40337341-40337341 on Build GRCh38
rs777802615 Chr.4: 40337197-40337197 on Build GRCh38
rs773726053 Chr.4: 40337404-40337404 on Build GRCh38
rs139782381 Chr.4: 40337311-40337311 on Build GRCh38
rs1029805977 Chr.4: 40337522-40337522 on Build GRCh38
rs369825430 Chr.4: 40337379-40337379 on Build GRCh38
rs534300891 Chr.4: 40337249-40337249 on Build GRCh38
rs373470519 Chr.4: 40337325-40337325 on Build GRCh38
rs951787614 Chr.4: 40337349-40337349 on Build GRCh38
rs988789021 Chr.4: 40337546-40337546 on Build GRCh38
rs536543895 Chr.4: 40337358-40337358 on Build GRCh38
rs985396823 Chr.4: 40337264-40337264 on Build GRCh38
rs947443743 Chr.4: 40337568-40337568 on Build GRCh38
rs6819385 Chr.4: 40337557-40337557 on Build GRCh38
rs1043679212 Chr.4: 40337416-40337416 on Build GRCh38
rs746881748 Chr.4: 40337277-40337277 on Build GRCh38
rs762487131 Chr.4: 40337170-40337170 on Build GRCh38
rs750727697 Chr.4: 40337245-40337245 on Build GRCh38
rs376664779 Chr.4: 40337359-40337359 on Build GRCh38
rs773146638 Chr.4: 40337317-40337317 on Build GRCh38
rs546046694 Chr.4: 40337427-40337427 on Build GRCh38
rs756848796 Chr.4: 40337278-40337278 on Build GRCh38
rs774774039 Chr.4: 40337234-40337234 on Build GRCh38
rs909523521 Chr.4: 40337562-40337562 on Build GRCh38
rs570024431 Chr.4: 40337200-40337200 on Build GRCh38
rs878953978 Chr.4: 40337454-40337454 on Build GRCh38
rs749934894 Chr.4: 40337344-40337344 on Build GRCh38
rs775685200 Chr.4: 40337211-40337211 on Build GRCh38
rs530279257 Chr.4: 40337171-40337171 on Build GRCh38
rs374894532 Chr.4: 40337187-40337187 on Build GRCh38
rs755586295 Chr.4: 40337348-40337348 on Build GRCh38
rs767679206 Chr.4: 40337314-40337314 on Build GRCh38
rs767224480 Chr.4: 40337285-40337285 on Build GRCh38
rs1050268364 Chr.4: 40337578-40337578 on Build GRCh38
rs763255413 Chr.4: 40337225-40337225 on Build GRCh38
rs767485650 Chr.4: 40337240-40337240 on Build GRCh38
rs769354742 Chr.4: 40337296-40337296 on Build GRCh38
rs199910277 Chr.4: 40337210-40337210 on Build GRCh38
rs745493859 Chr.4: 40337209-40337209 on Build GRCh38
rs368167486 Chr.4: 40337207-40337207 on Build GRCh38
rs757109316 Chr.4: 40337333-40337333 on Build GRCh38
rs751096383 Chr.4: 40337458-40337458 on Build GRCh38
rs112093951 Chr.4: 40337162-40337162 on Build GRCh38
rs763967910 Chr.4: 40337329-40337329 on Build GRCh38
rs953719075 Chr.4: 40337547-40337547 on Build GRCh38
rs112843181 Chr.4: 40337180-40337180 on Build GRCh38
rs758384454 Chr.4: 40337196-40337196 on Build GRCh38
rs562476798 Chr.4: 40337350-40337350 on Build GRCh38
rs371625417 Chr.4: 40337239-40337239 on Build GRCh38
rs748740413 Chr.4: 40337372-40337372 on Build GRCh38
rs775085099 Chr.4: 40337297-40337297 on Build GRCh38
rs757287793 Chr.4: 40337173-40337173 on Build GRCh38
rs141874739 Chr.4: 40337507-40337507 on Build GRCh38
rs760757245 Chr.4: 40337319-40337319 on Build GRCh38
rs1007437061 Chr.4: 40337299-40337299 on Build GRCh38
rs755484152 Chr.4: 40337263-40337263 on Build GRCh38
rs557144551 Chr.4: 40337302-40337302 on Build GRCh38
rs779695577 Chr.4: 40337351-40337351 on Build GRCh38
rs772482609 Chr.4: 40337403-40337403 on Build GRCh38
rs779221626 Chr.4: 40337369-40337369 on Build GRCh38
rs762307789 Chr.4: 40337306-40337306 on Build GRCh38
rs191525343 Chr.4: 40337300-40337300 on Build GRCh38
rs538917094 Chr.4: 40337205-40337205 on Build GRCh38
rs55998310 Chr.4: 40337233-40337233 on Build GRCh38

View Full Product Details