Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40354764-40355269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA9
Entrez Gene ID
55584
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
HSA243342, NACHRA9
UniGene
Hs.272278
Gene Chromosome Location
Chr.4: 40335329-40355217 on Build GRCh38
Transcripts
NM_017581.3

SNP Detail

SNP ID Chromosome Location
rs1007849125 Chr.4: 40355104-40355104 on Build GRCh38
rs1024506072 Chr.4: 40355059-40355059 on Build GRCh38
rs749116770 Chr.4: 40355015-40355015 on Build GRCh38
rs969709592 Chr.4: 40355226-40355226 on Build GRCh38
rs947714505 Chr.4: 40354943-40354943 on Build GRCh38
rs983965595 Chr.4: 40355172-40355172 on Build GRCh38
rs1050871151 Chr.4: 40355110-40355110 on Build GRCh38
rs768735140 Chr.4: 40354977-40354977 on Build GRCh38
rs563992009 Chr.4: 40355090-40355090 on Build GRCh38
rs373155261 Chr.4: 40355167-40355167 on Build GRCh38
rs1025865626 Chr.4: 40354917-40354917 on Build GRCh38
rs181016648 Chr.4: 40355109-40355109 on Build GRCh38
rs188119190 Chr.4: 40354976-40354976 on Build GRCh38
rs901871904 Chr.4: 40355111-40355111 on Build GRCh38
rs767690903 Chr.4: 40354878-40354878 on Build GRCh38
rs889848486 Chr.4: 40354820-40354820 on Build GRCh38
rs545777062 Chr.4: 40354791-40354791 on Build GRCh38
rs760676947 Chr.4: 40355190-40355190 on Build GRCh38
rs771996210 Chr.4: 40355181-40355181 on Build GRCh38
rs566060985 Chr.4: 40355173-40355173 on Build GRCh38
rs532378551 Chr.4: 40355100-40355100 on Build GRCh38
rs559200096 Chr.4: 40354834-40354834 on Build GRCh38
rs993553551 Chr.4: 40355155-40355155 on Build GRCh38
rs946886349 Chr.4: 40355078-40355078 on Build GRCh38
rs557250378 Chr.4: 40355098-40355098 on Build GRCh38
rs1052147280 Chr.4: 40354978-40354978 on Build GRCh38
rs36082183 Chr.4: 40355098-40355099 on Build GRCh38
rs185689952 Chr.4: 40354881-40354881 on Build GRCh38
rs942797019 Chr.4: 40355129-40355129 on Build GRCh38
rs898502111 Chr.4: 40355194-40355194 on Build GRCh38
rs530609255 Chr.4: 40354888-40354888 on Build GRCh38
rs918411635 Chr.4: 40354829-40354829 on Build GRCh38
rs992302650 Chr.4: 40354953-40354953 on Build GRCh38
rs1047362244 Chr.4: 40355240-40355240 on Build GRCh38
rs528003546 Chr.4: 40354925-40354926 on Build GRCh38
rs866744729 Chr.4: 40355221-40355221 on Build GRCh38
rs56291234 Chr.4: 40354882-40354882 on Build GRCh38
rs773225639 Chr.4: 40355183-40355183 on Build GRCh38
rs1026420268 Chr.4: 40355161-40355161 on Build GRCh38
rs935636067 Chr.4: 40355093-40355093 on Build GRCh38
rs865863614 Chr.4: 40354839-40354839 on Build GRCh38
rs868864375 Chr.4: 40354802-40354802 on Build GRCh38
rs779214953 Chr.4: 40355003-40355003 on Build GRCh38
rs969816653 Chr.4: 40355086-40355086 on Build GRCh38
rs980839784 Chr.4: 40354968-40354968 on Build GRCh38
rs572867780 Chr.4: 40354854-40354854 on Build GRCh38
rs747855683 Chr.4: 40355089-40355089 on Build GRCh38
rs376470500 Chr.4: 40355232-40355232 on Build GRCh38
rs373342310 Chr.4: 40355033-40355033 on Build GRCh38
rs927692947 Chr.4: 40354973-40354973 on Build GRCh38

View Full Product Details