Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42116200-42116700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND4
Entrez Gene ID
389206
Gene Name
BEN domain containing 4
Gene Aliases
CCDC4
UniGene
Hs.120591
Gene Chromosome Location
Chr.4: 42110853-42152878 on Build GRCh38
Transcripts
NM_001159547.1 NM_207406.3 XM_017008185.1

SNP Detail

SNP ID Chromosome Location
rs534523207 Chr.4: 42116644-42116644 on Build GRCh38
rs905992518 Chr.4: 42116462-42116462 on Build GRCh38
rs867992352 Chr.4: 42116373-42116373 on Build GRCh38
rs559456393 Chr.4: 42116270-42116270 on Build GRCh38
rs1030864732 Chr.4: 42116660-42116660 on Build GRCh38
rs942029164 Chr.4: 42116665-42116665 on Build GRCh38
rs1044446643 Chr.4: 42116423-42116423 on Build GRCh38
rs904170592 Chr.4: 42116523-42116523 on Build GRCh38
rs1007814169 Chr.4: 42116672-42116672 on Build GRCh38
rs1032353976 Chr.4: 42116390-42116390 on Build GRCh38
rs1056930956 Chr.4: 42116496-42116496 on Build GRCh38
rs976687967 Chr.4: 42116381-42116381 on Build GRCh38
rs1010156771 Chr.4: 42116248-42116248 on Build GRCh38
rs144490183 Chr.4: 42116402-42116402 on Build GRCh38
rs1035604539 Chr.4: 42116228-42116228 on Build GRCh38
rs142336180 Chr.4: 42116596-42116596 on Build GRCh38
rs942061929 Chr.4: 42116419-42116419 on Build GRCh38
rs1000823737 Chr.4: 42116347-42116347 on Build GRCh38
rs1045280828 Chr.4: 42116562-42116562 on Build GRCh38
rs964860060 Chr.4: 42116344-42116344 on Build GRCh38
rs1021570252 Chr.4: 42116500-42116500 on Build GRCh38
rs1053732305 Chr.4: 42116612-42116612 on Build GRCh38
rs909498176 Chr.4: 42116652-42116652 on Build GRCh38
rs547921431 Chr.4: 42116408-42116408 on Build GRCh38
rs949624183 Chr.4: 42116552-42116552 on Build GRCh38
rs973080973 Chr.4: 42116439-42116439 on Build GRCh38
rs965160868 Chr.4: 42116616-42116616 on Build GRCh38
rs147858911 Chr.4: 42116409-42116409 on Build GRCh38
rs16854026 Chr.4: 42116645-42116645 on Build GRCh38
rs954564081 Chr.4: 42116667-42116667 on Build GRCh38
rs774749472 Chr.4: 42116469-42116469 on Build GRCh38
rs538622300 Chr.4: 42116497-42116497 on Build GRCh38
rs918988041 Chr.4: 42116455-42116455 on Build GRCh38
rs368118325 Chr.4: 42116420-42116420 on Build GRCh38
rs780562543 Chr.4: 42116324-42116324 on Build GRCh38
rs976529943 Chr.4: 42116620-42116620 on Build GRCh38
rs968385568 Chr.4: 42116306-42116306 on Build GRCh38
rs143214267 Chr.4: 42116631-42116631 on Build GRCh38
rs78970435 Chr.4: 42116463-42116463 on Build GRCh38
rs891746886 Chr.4: 42116237-42116237 on Build GRCh38
rs1024342504 Chr.4: 42116283-42116284 on Build GRCh38
rs1030889575 Chr.4: 42116633-42116633 on Build GRCh38
rs6842251 Chr.4: 42116410-42116410 on Build GRCh38
rs1018057998 Chr.4: 42116338-42116338 on Build GRCh38
rs878942460 Chr.4: 42116449-42116449 on Build GRCh38
rs569689534 Chr.4: 42116486-42116486 on Build GRCh38
rs891633932 Chr.4: 42116477-42116477 on Build GRCh38
rs1010540505 Chr.4: 42116478-42116478 on Build GRCh38
rs1006587685 Chr.4: 42116581-42116581 on Build GRCh38
rs35139058 Chr.4: 42116489-42116490 on Build GRCh38
rs1018026952 Chr.4: 42116586-42116586 on Build GRCh38

View Full Product Details