Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42408297-42408811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

SNP Detail

SNP ID Chromosome Location
rs998331243 Chr.4: 42408454-42408454 on Build GRCh38
rs543530590 Chr.4: 42408761-42408761 on Build GRCh38
rs757584519 Chr.4: 42408489-42408489 on Build GRCh38
rs1051192481 Chr.4: 42408466-42408466 on Build GRCh38
rs751763585 Chr.4: 42408458-42408458 on Build GRCh38
rs1017564096 Chr.4: 42408515-42408515 on Build GRCh38
rs563131271 Chr.4: 42408762-42408762 on Build GRCh38
rs953075651 Chr.4: 42408393-42408393 on Build GRCh38
rs924976653 Chr.4: 42408346-42408346 on Build GRCh38
rs966083059 Chr.4: 42408455-42408455 on Build GRCh38
rs901575165 Chr.4: 42408428-42408428 on Build GRCh38
rs1022141986 Chr.4: 42408386-42408386 on Build GRCh38
rs187621752 Chr.4: 42408571-42408571 on Build GRCh38
rs762143714 Chr.4: 42408518-42408519 on Build GRCh38
rs992029127 Chr.4: 42408599-42408599 on Build GRCh38
rs985892560 Chr.4: 42408406-42408406 on Build GRCh38
rs774368811 Chr.4: 42408550-42408550 on Build GRCh38
rs898510392 Chr.4: 42408539-42408539 on Build GRCh38
rs3200171 Chr.4: 42408591-42408591 on Build GRCh38
rs558437721 Chr.4: 42408421-42408421 on Build GRCh38
rs960768532 Chr.4: 42408589-42408589 on Build GRCh38
rs911540396 Chr.4: 42408427-42408427 on Build GRCh38
rs1025757746 Chr.4: 42408556-42408556 on Build GRCh38
rs538554446 Chr.4: 42408351-42408351 on Build GRCh38
rs540848366 Chr.4: 42408675-42408675 on Build GRCh38
rs867184275 Chr.4: 42408359-42408359 on Build GRCh38
rs892349403 Chr.4: 42408785-42408785 on Build GRCh38
rs935029050 Chr.4: 42408751-42408751 on Build GRCh38
rs1007138529 Chr.4: 42408491-42408491 on Build GRCh38
rs969590050 Chr.4: 42408661-42408661 on Build GRCh38
rs906641770 Chr.4: 42408683-42408683 on Build GRCh38
rs558293528 Chr.4: 42408334-42408334 on Build GRCh38
rs984213143 Chr.4: 42408561-42408561 on Build GRCh38
rs367696386 Chr.4: 42408602-42408602 on Build GRCh38
rs568769077 Chr.4: 42408411-42408414 on Build GRCh38
rs750514344 Chr.4: 42408611-42408611 on Build GRCh38
rs183911974 Chr.4: 42408439-42408439 on Build GRCh38
rs191420358 Chr.4: 42408677-42408677 on Build GRCh38
rs756043784 Chr.4: 42408757-42408757 on Build GRCh38
rs560613944 Chr.4: 42408676-42408676 on Build GRCh38

View Full Product Details