Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42410645-42411154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

SNP Detail

SNP ID Chromosome Location
rs1017676262 Chr.4: 42410751-42410751 on Build GRCh38
rs566356269 Chr.4: 42410900-42410900 on Build GRCh38
rs201192684 Chr.4: 42410789-42410789 on Build GRCh38
rs1004867193 Chr.4: 42411127-42411127 on Build GRCh38
rs980375059 Chr.4: 42410862-42410862 on Build GRCh38
rs941854112 Chr.4: 42410861-42410861 on Build GRCh38
rs749763795 Chr.4: 42411110-42411110 on Build GRCh38
rs375090958 Chr.4: 42411069-42411069 on Build GRCh38
rs1018229301 Chr.4: 42411091-42411091 on Build GRCh38
rs370710519 Chr.4: 42410824-42410824 on Build GRCh38
rs927058993 Chr.4: 42410863-42410863 on Build GRCh38
rs905593999 Chr.4: 42411004-42411004 on Build GRCh38
rs779796235 Chr.4: 42410816-42410816 on Build GRCh38
rs1002819759 Chr.4: 42411019-42411019 on Build GRCh38
rs1045525257 Chr.4: 42410912-42410912 on Build GRCh38
rs1001058555 Chr.4: 42410722-42410722 on Build GRCh38
rs147535199 Chr.4: 42410850-42410850 on Build GRCh38
rs983349457 Chr.4: 42410828-42410828 on Build GRCh38
rs891470830 Chr.4: 42410968-42410968 on Build GRCh38
rs1001027465 Chr.4: 42411061-42411061 on Build GRCh38
rs568367898 Chr.4: 42410823-42410823 on Build GRCh38
rs886113900 Chr.4: 42410908-42410908 on Build GRCh38
rs746516536 Chr.4: 42410955-42410955 on Build GRCh38
rs12639920 Chr.4: 42410670-42410670 on Build GRCh38
rs966649413 Chr.4: 42410747-42410747 on Build GRCh38
rs554505427 Chr.4: 42410706-42410706 on Build GRCh38
rs776234603 Chr.4: 42411058-42411058 on Build GRCh38
rs1042645076 Chr.4: 42411023-42411023 on Build GRCh38
rs377360279 Chr.4: 42410705-42410705 on Build GRCh38
rs868656484 Chr.4: 42410674-42410674 on Build GRCh38
rs546242498 Chr.4: 42411117-42411117 on Build GRCh38
rs533805447 Chr.4: 42411000-42411000 on Build GRCh38
rs777358559 Chr.4: 42410910-42410910 on Build GRCh38
rs1047638370 Chr.4: 42410885-42410885 on Build GRCh38
rs528473938 Chr.4: 42410750-42410750 on Build GRCh38
rs570584888 Chr.4: 42410848-42410848 on Build GRCh38
rs541868334 Chr.4: 42411103-42411103 on Build GRCh38
rs958409548 Chr.4: 42411049-42411049 on Build GRCh38
rs548134948 Chr.4: 42410770-42410770 on Build GRCh38
rs554930937 Chr.4: 42411120-42411120 on Build GRCh38
rs553146498 Chr.4: 42410916-42410916 on Build GRCh38
rs60851756 Chr.4: 42411057-42411057 on Build GRCh38
rs965191099 Chr.4: 42410755-42410755 on Build GRCh38
rs950700747 Chr.4: 42411123-42411123 on Build GRCh38
rs557104957 Chr.4: 42410826-42410826 on Build GRCh38

View Full Product Details