Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42423157-42423700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527164 c.3193C>T p.L1065F Chr.4: 42423636-42423636 on Build GRCh38
5527165 c.3148C>T p.L1050F Chr.4: 42423636-42423636 on Build GRCh38
5675356 c.3155T>C p.V1052A Chr.4: 42423629-42423629 on Build GRCh38
4542286 c.3111G>A p.W1037* Chr.4: 42423673-42423673 on Build GRCh38
5675355 c.3200T>C p.V1067A Chr.4: 42423629-42423629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565775374 Chr.4: 42423236-42423236 on Build GRCh38
rs1002583941 Chr.4: 42423311-42423311 on Build GRCh38
rs891327155 Chr.4: 42423430-42423430 on Build GRCh38
rs1020862221 Chr.4: 42423392-42423392 on Build GRCh38
rs766454856 Chr.4: 42423583-42423583 on Build GRCh38
rs200613613 Chr.4: 42423652-42423652 on Build GRCh38
rs767301299 Chr.4: 42423665-42423665 on Build GRCh38
rs757180900 Chr.4: 42423661-42423661 on Build GRCh38
rs374530990 Chr.4: 42423625-42423625 on Build GRCh38
rs57101361 Chr.4: 42423419-42423419 on Build GRCh38
rs974475896 Chr.4: 42423231-42423231 on Build GRCh38
rs576788347 Chr.4: 42423458-42423458 on Build GRCh38
rs188079945 Chr.4: 42423574-42423574 on Build GRCh38
rs896500736 Chr.4: 42423375-42423375 on Build GRCh38
rs1042282595 Chr.4: 42423655-42423655 on Build GRCh38
rs373255752 Chr.4: 42423408-42423408 on Build GRCh38
rs745500740 Chr.4: 42423604-42423604 on Build GRCh38
rs143412392 Chr.4: 42423545-42423545 on Build GRCh38
rs142532672 Chr.4: 42423478-42423478 on Build GRCh38
rs964108981 Chr.4: 42423222-42423222 on Build GRCh38
rs1018928169 Chr.4: 42423200-42423200 on Build GRCh38
rs976134569 Chr.4: 42423455-42423455 on Build GRCh38
rs1038421171 Chr.4: 42423264-42423264 on Build GRCh38
rs980742923 Chr.4: 42423365-42423365 on Build GRCh38
rs779661502 Chr.4: 42423616-42423616 on Build GRCh38
rs753954548 Chr.4: 42423589-42423589 on Build GRCh38
rs982996612 Chr.4: 42423485-42423485 on Build GRCh38
rs970568665 Chr.4: 42423561-42423561 on Build GRCh38
rs41265685 Chr.4: 42423242-42423242 on Build GRCh38
rs1004277718 Chr.4: 42423507-42423507 on Build GRCh38
rs769215914 Chr.4: 42423620-42423620 on Build GRCh38
rs1017763682 Chr.4: 42423438-42423438 on Build GRCh38
rs370413768 Chr.4: 42423595-42423595 on Build GRCh38
rs922937197 Chr.4: 42423477-42423477 on Build GRCh38
rs537059261 Chr.4: 42423404-42423404 on Build GRCh38
rs900084614 Chr.4: 42423476-42423476 on Build GRCh38
rs760858660 Chr.4: 42423580-42423580 on Build GRCh38
rs773492388 Chr.4: 42423627-42423627 on Build GRCh38
rs753857860 Chr.4: 42423647-42423647 on Build GRCh38
rs201225804 Chr.4: 42423598-42423598 on Build GRCh38
rs936389691 Chr.4: 42423377-42423377 on Build GRCh38
rs917767315 Chr.4: 42423276-42423276 on Build GRCh38
rs554371406 Chr.4: 42423251-42423251 on Build GRCh38
rs748819585 Chr.4: 42423618-42423618 on Build GRCh38
rs1009424824 Chr.4: 42423378-42423378 on Build GRCh38
rs774990376 Chr.4: 42423623-42423623 on Build GRCh38
rs908794119 Chr.4: 42423505-42423505 on Build GRCh38
rs913590426 Chr.4: 42423640-42423640 on Build GRCh38
rs775968018 Chr.4: 42423443-42423443 on Build GRCh38
rs191498738 Chr.4: 42423592-42423592 on Build GRCh38
rs766613625 Chr.4: 42423630-42423630 on Build GRCh38
rs979273655 Chr.4: 42423426-42423426 on Build GRCh38
rs572561396 Chr.4: 42423568-42423568 on Build GRCh38
rs377196430 Chr.4: 42423593-42423593 on Build GRCh38
rs765368933 Chr.4: 42423654-42423654 on Build GRCh38
rs1026947809 Chr.4: 42423232-42423236 on Build GRCh38
rs945122853 Chr.4: 42423425-42423425 on Build GRCh38
rs750206508 Chr.4: 42423676-42423676 on Build GRCh38
rs368422986 Chr.4: 42423601-42423601 on Build GRCh38
rs751502747 Chr.4: 42423660-42423660 on Build GRCh38
rs1053867127 Chr.4: 42423393-42423393 on Build GRCh38
rs769360705 Chr.4: 42423608-42423608 on Build GRCh38
rs951359150 Chr.4: 42423501-42423501 on Build GRCh38
rs762125576 Chr.4: 42423667-42423667 on Build GRCh38
rs905282564 Chr.4: 42423267-42423267 on Build GRCh38
rs200291526 Chr.4: 42423648-42423648 on Build GRCh38
rs760914967 Chr.4: 42423628-42423628 on Build GRCh38
rs543393144 Chr.4: 42423374-42423374 on Build GRCh38
rs949204719 Chr.4: 42423304-42423304 on Build GRCh38

View Full Product Details