Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42443412-42443933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2847782 c.2985G>A p.A995A Chr.4: 42443658-42443658 on Build GRCh38
159289 c.3029C>T p.A1010V Chr.4: 42443659-42443659 on Build GRCh38
2847781 c.3030G>A p.A1010A Chr.4: 42443658-42443658 on Build GRCh38
4809794 c.2984C>T p.A995V Chr.4: 42443659-42443659 on Build GRCh38
5449996 c.3054G>A p.P1018P Chr.4: 42443589-42443589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961588509 Chr.4: 42443866-42443866 on Build GRCh38
rs1028625896 Chr.4: 42443809-42443809 on Build GRCh38
rs912707082 Chr.4: 42443850-42443850 on Build GRCh38
rs140420171 Chr.4: 42443590-42443590 on Build GRCh38
rs1041223684 Chr.4: 42443759-42443759 on Build GRCh38
rs754038062 Chr.4: 42443488-42443488 on Build GRCh38
rs957804374 Chr.4: 42443775-42443775 on Build GRCh38
rs769852292 Chr.4: 42443754-42443756 on Build GRCh38
rs773007378 Chr.4: 42443667-42443667 on Build GRCh38
rs777031347 Chr.4: 42443608-42443608 on Build GRCh38
rs987103198 Chr.4: 42443829-42443829 on Build GRCh38
rs759181094 Chr.4: 42443556-42443556 on Build GRCh38
rs550026430 Chr.4: 42443887-42443887 on Build GRCh38
rs564983245 Chr.4: 42443583-42443583 on Build GRCh38
rs868693295 Chr.4: 42443591-42443591 on Build GRCh38
rs934966577 Chr.4: 42443755-42443755 on Build GRCh38
rs746048297 Chr.4: 42443612-42443612 on Build GRCh38
rs778148153 Chr.4: 42443589-42443589 on Build GRCh38
rs778269282 Chr.4: 42443502-42443502 on Build GRCh38
rs766331746 Chr.4: 42443565-42443565 on Build GRCh38
rs760740652 Chr.4: 42443673-42443673 on Build GRCh38
rs374633134 Chr.4: 42443575-42443575 on Build GRCh38
rs536321125 Chr.4: 42443485-42443485 on Build GRCh38
rs746243024 Chr.4: 42443720-42443720 on Build GRCh38
rs902379218 Chr.4: 42443739-42443739 on Build GRCh38
rs901369348 Chr.4: 42443791-42443791 on Build GRCh38
rs999669305 Chr.4: 42443751-42443751 on Build GRCh38
rs762946178 Chr.4: 42443643-42443643 on Build GRCh38
rs973329253 Chr.4: 42443874-42443874 on Build GRCh38
rs753719903 Chr.4: 42443679-42443679 on Build GRCh38
rs769935963 Chr.4: 42443723-42443723 on Build GRCh38
rs144348573 Chr.4: 42443691-42443691 on Build GRCh38
rs771273391 Chr.4: 42443596-42443596 on Build GRCh38
rs764297544 Chr.4: 42443652-42443652 on Build GRCh38
rs759813589 Chr.4: 42443522-42443522 on Build GRCh38
rs764931957 Chr.4: 42443712-42443712 on Build GRCh38
rs1055141971 Chr.4: 42443659-42443659 on Build GRCh38
rs556217815 Chr.4: 42443547-42443547 on Build GRCh38
rs766243870 Chr.4: 42443676-42443676 on Build GRCh38
rs995771544 Chr.4: 42443804-42443804 on Build GRCh38
rs748101930 Chr.4: 42443891-42443891 on Build GRCh38
rs757842591 Chr.4: 42443703-42443703 on Build GRCh38
rs750165044 Chr.4: 42443556-42443556 on Build GRCh38
rs940090860 Chr.4: 42443512-42443512 on Build GRCh38
rs1015941842 Chr.4: 42443481-42443481 on Build GRCh38
rs764814643 Chr.4: 42443699-42443699 on Build GRCh38
rs757802910 Chr.4: 42443707-42443707 on Build GRCh38
rs776830285 Chr.4: 42443517-42443517 on Build GRCh38
rs148611717 Chr.4: 42443441-42443441 on Build GRCh38
rs770024010 Chr.4: 42443618-42443618 on Build GRCh38
rs961204461 Chr.4: 42443492-42443492 on Build GRCh38
rs1005445904 Chr.4: 42443443-42443443 on Build GRCh38
rs767369013 Chr.4: 42443555-42443555 on Build GRCh38
rs927840685 Chr.4: 42443509-42443510 on Build GRCh38
rs954291343 Chr.4: 42443826-42443826 on Build GRCh38
rs752040758 Chr.4: 42443704-42443704 on Build GRCh38
rs758886673 Chr.4: 42443587-42443587 on Build GRCh38
rs924859230 Chr.4: 42443734-42443734 on Build GRCh38
rs754952867 Chr.4: 42443642-42443642 on Build GRCh38
rs199599944 Chr.4: 42443579-42443579 on Build GRCh38
rs765280728 Chr.4: 42443543-42443543 on Build GRCh38
rs919978183 Chr.4: 42443879-42443879 on Build GRCh38
rs371894314 Chr.4: 42443549-42443549 on Build GRCh38
rs1005976453 Chr.4: 42443898-42443898 on Build GRCh38
rs572199208 Chr.4: 42443586-42443586 on Build GRCh38
rs41265687 Chr.4: 42443717-42443717 on Build GRCh38
rs113742288 Chr.4: 42443909-42443909 on Build GRCh38
rs200909875 Chr.4: 42443557-42443557 on Build GRCh38
rs762385226 Chr.4: 42443859-42443859 on Build GRCh38
rs1032118712 Chr.4: 42443756-42443756 on Build GRCh38
rs990558567 Chr.4: 42443552-42443552 on Build GRCh38
rs931278936 Chr.4: 42443610-42443610 on Build GRCh38
rs752189374 Chr.4: 42443700-42443700 on Build GRCh38

View Full Product Details