Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42444436-42444956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8A1
Entrez Gene ID
10396
Gene Name
ATPase phospholipid transporting 8A1
Gene Aliases
ATPASEII, ATPIA, ATPP2
UniGene
Hs.435052
Gene Chromosome Location
Chr.4: 42408373-42657134 on Build GRCh38
Transcripts
NM_001105529.1 NM_006095.2 XM_005248043.2 XM_011513615.1 XM_011513616.2 XM_011513618.1 XM_017007645.1 XM_017007646.1 XM_017007647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3661108 c.2938A>G p.K980E Chr.4: 42444610-42444610 on Build GRCh38
3661110 c.2937G>T p.L979F Chr.4: 42444611-42444611 on Build GRCh38
4931163 c.2946A>G p.G982G Chr.4: 42444602-42444602 on Build GRCh38
4931162 c.2991A>G p.G997G Chr.4: 42444602-42444602 on Build GRCh38
733426 c.3009G>A p.W1003* Chr.4: 42444584-42444584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755055422 Chr.4: 42444590-42444590 on Build GRCh38
rs567940225 Chr.4: 42444695-42444695 on Build GRCh38
rs970799351 Chr.4: 42444732-42444732 on Build GRCh38
rs371792292 Chr.4: 42444643-42444643 on Build GRCh38
rs765146916 Chr.4: 42444562-42444562 on Build GRCh38
rs760646723 Chr.4: 42444541-42444541 on Build GRCh38
rs113731569 Chr.4: 42444807-42444807 on Build GRCh38
rs757702547 Chr.4: 42444593-42444593 on Build GRCh38
rs1050233284 Chr.4: 42444505-42444505 on Build GRCh38
rs770929409 Chr.4: 42444679-42444679 on Build GRCh38
rs915055340 Chr.4: 42444460-42444460 on Build GRCh38
rs200861548 Chr.4: 42444645-42444645 on Build GRCh38
rs768592229 Chr.4: 42444537-42444537 on Build GRCh38
rs372064110 Chr.4: 42444510-42444510 on Build GRCh38
rs888720121 Chr.4: 42444654-42444654 on Build GRCh38
rs910023846 Chr.4: 42444753-42444753 on Build GRCh38
rs999376663 Chr.4: 42444910-42444910 on Build GRCh38
rs749437099 Chr.4: 42444651-42444651 on Build GRCh38
rs901593393 Chr.4: 42444930-42444930 on Build GRCh38
rs759413545 Chr.4: 42444561-42444561 on Build GRCh38
rs558648950 Chr.4: 42444827-42444827 on Build GRCh38
rs748709079 Chr.4: 42444853-42444853 on Build GRCh38
rs780135360 Chr.4: 42444528-42444528 on Build GRCh38
rs985760288 Chr.4: 42444742-42444742 on Build GRCh38
rs774527266 Chr.4: 42444540-42444540 on Build GRCh38
rs867835352 Chr.4: 42444709-42444709 on Build GRCh38
rs144342471 Chr.4: 42444599-42444599 on Build GRCh38
rs903654749 Chr.4: 42444868-42444868 on Build GRCh38
rs150403408 Chr.4: 42444733-42444733 on Build GRCh38
rs554135249 Chr.4: 42444502-42444502 on Build GRCh38
rs71610018 Chr.4: 42444884-42444884 on Build GRCh38
rs113586205 Chr.4: 42444917-42444917 on Build GRCh38
rs186426291 Chr.4: 42444852-42444852 on Build GRCh38
rs140860952 Chr.4: 42444458-42444458 on Build GRCh38
rs768951280 Chr.4: 42444655-42444655 on Build GRCh38
rs374824679 Chr.4: 42444566-42444566 on Build GRCh38
rs751089444 Chr.4: 42444574-42444574 on Build GRCh38
rs749577888 Chr.4: 42444535-42444535 on Build GRCh38
rs953303846 Chr.4: 42444841-42444841 on Build GRCh38
rs758757211 Chr.4: 42444532-42444532 on Build GRCh38
rs199510254 Chr.4: 42444559-42444559 on Build GRCh38
rs779089543 Chr.4: 42444659-42444659 on Build GRCh38
rs147792614 Chr.4: 42444617-42444617 on Build GRCh38
rs371477752 Chr.4: 42444894-42444894 on Build GRCh38
rs1028705667 Chr.4: 42444722-42444722 on Build GRCh38
rs865784125 Chr.4: 42444591-42444591 on Build GRCh38
rs752279050 Chr.4: 42444564-42444564 on Build GRCh38
rs200581251 Chr.4: 42444611-42444611 on Build GRCh38
rs538712842 Chr.4: 42444818-42444818 on Build GRCh38
rs781666426 Chr.4: 42444608-42444608 on Build GRCh38
rs16854376 Chr.4: 42444515-42444515 on Build GRCh38
rs911752599 Chr.4: 42444865-42444865 on Build GRCh38
rs190002759 Chr.4: 42444861-42444861 on Build GRCh38
rs1024234454 Chr.4: 42444675-42444675 on Build GRCh38
rs746876156 Chr.4: 42444746-42444746 on Build GRCh38
rs931790729 Chr.4: 42444468-42444468 on Build GRCh38
rs944455721 Chr.4: 42444524-42444524 on Build GRCh38
rs563477863 Chr.4: 42444486-42444486 on Build GRCh38
rs547912421 Chr.4: 42444690-42444690 on Build GRCh38
rs748255237 Chr.4: 42444665-42444665 on Build GRCh38
rs780391575 Chr.4: 42444555-42444556 on Build GRCh38
rs369725675 Chr.4: 42444685-42444685 on Build GRCh38
rs745546365 Chr.4: 42444875-42444875 on Build GRCh38
rs554704314 Chr.4: 42444862-42444862 on Build GRCh38
rs10004341 Chr.4: 42444804-42444804 on Build GRCh38
rs752534643 Chr.4: 42444514-42444514 on Build GRCh38
rs769257365 Chr.4: 42444889-42444889 on Build GRCh38
rs543751053 Chr.4: 42444933-42444933 on Build GRCh38
rs369524554 Chr.4: 42444638-42444638 on Build GRCh38
rs956943557 Chr.4: 42444805-42444805 on Build GRCh38
rs201430358 Chr.4: 42444549-42444549 on Build GRCh38
rs776710617 Chr.4: 42444683-42444683 on Build GRCh38
rs918672636 Chr.4: 42444813-42444813 on Build GRCh38

View Full Product Details