Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46037874-46038421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG1
Entrez Gene ID
2565
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Gene Aliases
-
UniGene
Hs.375051
Gene Chromosome Location
Chr.4: 46035769-46124065 on Build GRCh38
Transcripts
NM_173536.3 XM_017007990.1

SNP Detail

SNP ID Chromosome Location
rs575773607 Chr.4: 46038287-46038287 on Build GRCh38
rs556386575 Chr.4: 46037948-46037948 on Build GRCh38
rs1020084651 Chr.4: 46038075-46038075 on Build GRCh38
rs939625745 Chr.4: 46038233-46038233 on Build GRCh38
rs1041071765 Chr.4: 46038076-46038076 on Build GRCh38
rs543796154 Chr.4: 46038222-46038222 on Build GRCh38
rs867588995 Chr.4: 46038266-46038266 on Build GRCh38
rs913007724 Chr.4: 46037944-46037944 on Build GRCh38
rs562347799 Chr.4: 46038229-46038229 on Build GRCh38
rs553497428 Chr.4: 46037997-46037997 on Build GRCh38
rs982714842 Chr.4: 46037933-46037933 on Build GRCh38
rs937257760 Chr.4: 46037969-46037969 on Build GRCh38
rs557733430 Chr.4: 46038001-46038001 on Build GRCh38
rs144527658 Chr.4: 46038193-46038193 on Build GRCh38
rs981034701 Chr.4: 46038205-46038205 on Build GRCh38
rs967108461 Chr.4: 46038195-46038195 on Build GRCh38
rs537771093 Chr.4: 46037934-46037934 on Build GRCh38
rs1032624982 Chr.4: 46038118-46038118 on Build GRCh38
rs982829075 Chr.4: 46038179-46038179 on Build GRCh38
rs1056994500 Chr.4: 46038249-46038249 on Build GRCh38
rs982622519 Chr.4: 46038292-46038292 on Build GRCh38
rs149325644 Chr.4: 46038105-46038105 on Build GRCh38
rs369185501 Chr.4: 46038028-46038028 on Build GRCh38
rs571798662 Chr.4: 46038054-46038054 on Build GRCh38
rs941346094 Chr.4: 46038206-46038206 on Build GRCh38
rs1055754058 Chr.4: 46037974-46037974 on Build GRCh38
rs964010319 Chr.4: 46038130-46038130 on Build GRCh38
rs1009709570 Chr.4: 46038176-46038176 on Build GRCh38
rs535247500 Chr.4: 46037968-46037968 on Build GRCh38
rs991800376 Chr.4: 46038275-46038275 on Build GRCh38
rs1029575885 Chr.4: 46038325-46038325 on Build GRCh38
rs980821174 Chr.4: 46038221-46038221 on Build GRCh38
rs951430830 Chr.4: 46038165-46038165 on Build GRCh38
rs962846623 Chr.4: 46038381-46038381 on Build GRCh38
rs775190427 Chr.4: 46038276-46038276 on Build GRCh38
rs1041186013 Chr.4: 46038371-46038371 on Build GRCh38
rs559948669 Chr.4: 46038386-46038386 on Build GRCh38
rs998117960 Chr.4: 46038313-46038313 on Build GRCh38
rs908591316 Chr.4: 46038329-46038329 on Build GRCh38
rs567945803 Chr.4: 46037963-46037963 on Build GRCh38
rs981085637 Chr.4: 46037954-46037954 on Build GRCh38
rs1497565 Chr.4: 46038350-46038350 on Build GRCh38
rs919745326 Chr.4: 46037989-46037989 on Build GRCh38
rs372851798 Chr.4: 46038038-46038042 on Build GRCh38
rs557470307 Chr.4: 46038134-46038134 on Build GRCh38
rs1008200634 Chr.4: 46038050-46038050 on Build GRCh38
rs899871992 Chr.4: 46038080-46038080 on Build GRCh38
rs533470700 Chr.4: 46038393-46038393 on Build GRCh38
rs932693552 Chr.4: 46038378-46038378 on Build GRCh38
rs931189440 Chr.4: 46037991-46037991 on Build GRCh38
rs148459731 Chr.4: 46038373-46038373 on Build GRCh38
rs757968043 Chr.4: 46038244-46038244 on Build GRCh38
rs1047158529 Chr.4: 46037992-46037992 on Build GRCh38
rs999891272 Chr.4: 46038083-46038083 on Build GRCh38
rs894071858 Chr.4: 46038398-46038398 on Build GRCh38
rs779463470 Chr.4: 46038259-46038259 on Build GRCh38
rs990293393 Chr.4: 46037927-46037927 on Build GRCh38

View Full Product Details