Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46040101-46040600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG1
Entrez Gene ID
2565
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Gene Aliases
-
UniGene
Hs.375051
Gene Chromosome Location
Chr.4: 46035769-46124065 on Build GRCh38
Transcripts
NM_173536.3 XM_017007990.1

SNP Detail

SNP ID Chromosome Location
rs962294763 Chr.4: 46040291-46040291 on Build GRCh38
rs962000924 Chr.4: 46040323-46040323 on Build GRCh38
rs953251849 Chr.4: 46040239-46040239 on Build GRCh38
rs916081369 Chr.4: 46040376-46040376 on Build GRCh38
rs555057416 Chr.4: 46040153-46040153 on Build GRCh38
rs935132096 Chr.4: 46040155-46040155 on Build GRCh38
rs912005292 Chr.4: 46040316-46040316 on Build GRCh38
rs1030912413 Chr.4: 46040319-46040319 on Build GRCh38
rs183017448 Chr.4: 46040501-46040501 on Build GRCh38
rs571110168 Chr.4: 46040413-46040413 on Build GRCh38
rs1056704262 Chr.4: 46040192-46040192 on Build GRCh38
rs530711204 Chr.4: 46040258-46040258 on Build GRCh38
rs569287318 Chr.4: 46040136-46040136 on Build GRCh38
rs999161182 Chr.4: 46040497-46040497 on Build GRCh38
rs997643087 Chr.4: 46040314-46040314 on Build GRCh38
rs567365091 Chr.4: 46040284-46040284 on Build GRCh38
rs957992408 Chr.4: 46040144-46040144 on Build GRCh38
rs1045531389 Chr.4: 46040476-46040476 on Build GRCh38
rs557335883 Chr.4: 46040557-46040557 on Build GRCh38
rs190528539 Chr.4: 46040183-46040183 on Build GRCh38
rs546582012 Chr.4: 46040412-46040412 on Build GRCh38
rs1008707127 Chr.4: 46040573-46040574 on Build GRCh38
rs897865238 Chr.4: 46040577-46040577 on Build GRCh38
rs1045180695 Chr.4: 46040252-46040252 on Build GRCh38
rs1028827876 Chr.4: 46040220-46040220 on Build GRCh38
rs950233375 Chr.4: 46040392-46040392 on Build GRCh38
rs549125548 Chr.4: 46040263-46040263 on Build GRCh38
rs935247519 Chr.4: 46040425-46040425 on Build GRCh38
rs565391512 Chr.4: 46040253-46040253 on Build GRCh38
rs923821642 Chr.4: 46040408-46040408 on Build GRCh38
rs536621966 Chr.4: 46040137-46040137 on Build GRCh38
rs947935980 Chr.4: 46040472-46040472 on Build GRCh38
rs896359976 Chr.4: 46040219-46040219 on Build GRCh38
rs917730944 Chr.4: 46040471-46040471 on Build GRCh38
rs979077287 Chr.4: 46040366-46040366 on Build GRCh38
rs985597296 Chr.4: 46040348-46040348 on Build GRCh38
rs566981800 Chr.4: 46040347-46040347 on Build GRCh38
rs1006244085 Chr.4: 46040275-46040275 on Build GRCh38
rs73813157 Chr.4: 46040361-46040361 on Build GRCh38
rs1029359219 Chr.4: 46040554-46040555 on Build GRCh38
rs910994637 Chr.4: 46040356-46040356 on Build GRCh38
rs1015155898 Chr.4: 46040277-46040277 on Build GRCh38
rs941976050 Chr.4: 46040508-46040508 on Build GRCh38
rs1056419356 Chr.4: 46040470-46040470 on Build GRCh38
rs1010822075 Chr.4: 46040514-46040514 on Build GRCh38
rs544950159 Chr.4: 46040177-46040177 on Build GRCh38
rs1012738805 Chr.4: 46040224-46040224 on Build GRCh38

View Full Product Details