Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46058081-46058577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRG1
Entrez Gene ID
2565
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Gene Aliases
-
UniGene
Hs.375051
Gene Chromosome Location
Chr.4: 46035769-46124065 on Build GRCh38
Transcripts
NM_173536.3 XM_017007990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188862 c.784A>T p.I262F Chr.4: 46058349-46058349 on Build GRCh38
1252998 c.827A>C p.Q276P Chr.4: 46058306-46058306 on Build GRCh38
5466163 c.809T>C p.M270T Chr.4: 46058324-46058324 on Build GRCh38
1694314 c.916G>A p.G306S Chr.4: 46058217-46058217 on Build GRCh38
123305 c.806G>A p.R269K Chr.4: 46058327-46058327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776002505 Chr.4: 46058450-46058450 on Build GRCh38
rs549592022 Chr.4: 46058220-46058220 on Build GRCh38
rs953963389 Chr.4: 46058442-46058442 on Build GRCh38
rs535099025 Chr.4: 46058300-46058300 on Build GRCh38
rs760149181 Chr.4: 46058448-46058448 on Build GRCh38
rs555570300 Chr.4: 46058438-46058438 on Build GRCh38
rs200409168 Chr.4: 46058461-46058461 on Build GRCh38
rs989928187 Chr.4: 46058108-46058108 on Build GRCh38
rs776476920 Chr.4: 46058377-46058377 on Build GRCh38
rs777219393 Chr.4: 46058421-46058421 on Build GRCh38
rs760463590 Chr.4: 46058354-46058366 on Build GRCh38
rs938521346 Chr.4: 46058435-46058435 on Build GRCh38
rs376780739 Chr.4: 46058475-46058475 on Build GRCh38
rs540114569 Chr.4: 46058441-46058441 on Build GRCh38
rs768281552 Chr.4: 46058376-46058377 on Build GRCh38
rs775153115 Chr.4: 46058184-46058184 on Build GRCh38
rs756652892 Chr.4: 46058298-46058298 on Build GRCh38
rs529329140 Chr.4: 46058423-46058423 on Build GRCh38
rs914212784 Chr.4: 46058120-46058120 on Build GRCh38
rs369176789 Chr.4: 46058383-46058383 on Build GRCh38
rs761725919 Chr.4: 46058459-46058459 on Build GRCh38
rs1000210963 Chr.4: 46058351-46058351 on Build GRCh38
rs41265717 Chr.4: 46058190-46058190 on Build GRCh38
rs112216756 Chr.4: 46058208-46058208 on Build GRCh38
rs778528542 Chr.4: 46058474-46058474 on Build GRCh38
rs369652618 Chr.4: 46058493-46058493 on Build GRCh38
rs777247413 Chr.4: 46058368-46058368 on Build GRCh38
rs375029255 Chr.4: 46058378-46058378 on Build GRCh38
rs568096519 Chr.4: 46058222-46058222 on Build GRCh38
rs769995991 Chr.4: 46058449-46058449 on Build GRCh38
rs545674513 Chr.4: 46058496-46058496 on Build GRCh38
rs372346061 Chr.4: 46058296-46058296 on Build GRCh38
rs769698999 Chr.4: 46058183-46058183 on Build GRCh38
rs888693202 Chr.4: 46058135-46058135 on Build GRCh38
rs754985301 Chr.4: 46058138-46058138 on Build GRCh38
rs1036846736 Chr.4: 46058165-46058165 on Build GRCh38
rs775645619 Chr.4: 46058340-46058341 on Build GRCh38
rs566379202 Chr.4: 46058377-46058377 on Build GRCh38
rs142799145 Chr.4: 46058518-46058518 on Build GRCh38
rs139933358 Chr.4: 46058492-46058492 on Build GRCh38
rs368244440 Chr.4: 46058293-46058293 on Build GRCh38
rs761135864 Chr.4: 46058348-46058348 on Build GRCh38
rs769209641 Chr.4: 46058517-46058517 on Build GRCh38
rs114674065 Chr.4: 46058159-46058159 on Build GRCh38
rs888456827 Chr.4: 46058549-46058549 on Build GRCh38
rs576537296 Chr.4: 46058414-46058414 on Build GRCh38
rs41310791 Chr.4: 46058498-46058498 on Build GRCh38
rs759791928 Chr.4: 46058372-46058372 on Build GRCh38
rs747853091 Chr.4: 46058337-46058337 on Build GRCh38
rs559291958 Chr.4: 46058479-46058479 on Build GRCh38
rs772301203 Chr.4: 46058341-46058341 on Build GRCh38
rs756392838 Chr.4: 46058500-46058500 on Build GRCh38
rs367962945 Chr.4: 46058211-46058211 on Build GRCh38
rs746294744 Chr.4: 46058503-46058503 on Build GRCh38
rs770517873 Chr.4: 46058439-46058439 on Build GRCh38
rs771367844 Chr.4: 46058362-46058362 on Build GRCh38
rs955757999 Chr.4: 46058107-46058107 on Build GRCh38
rs932324409 Chr.4: 46058123-46058123 on Build GRCh38
rs1004887803 Chr.4: 46058158-46058158 on Build GRCh38
rs749637208 Chr.4: 46058516-46058516 on Build GRCh38
rs541028130 Chr.4: 46058468-46058468 on Build GRCh38
rs557084864 Chr.4: 46058385-46058385 on Build GRCh38
rs764389097 Chr.4: 46058455-46058455 on Build GRCh38
rs758183396 Chr.4: 46058396-46058396 on Build GRCh38
rs1056935479 Chr.4: 46058458-46058458 on Build GRCh38
rs749013190 Chr.4: 46058302-46058302 on Build GRCh38
rs540110325 Chr.4: 46058384-46058384 on Build GRCh38
rs185565097 Chr.4: 46058338-46058338 on Build GRCh38
rs150607696 Chr.4: 46058554-46058554 on Build GRCh38
rs753658054 Chr.4: 46058466-46058466 on Build GRCh38
rs755182208 Chr.4: 46058244-46058244 on Build GRCh38
rs77939156 Chr.4: 46058395-46058395 on Build GRCh38
rs1048417257 Chr.4: 46058131-46058131 on Build GRCh38
rs41309282 Chr.4: 46058465-46058465 on Build GRCh38
rs758242317 Chr.4: 46058478-46058478 on Build GRCh38
rs773368605 Chr.4: 46058543-46058543 on Build GRCh38
rs747728361 Chr.4: 46058413-46058413 on Build GRCh38
rs1048478263 Chr.4: 46058281-46058281 on Build GRCh38
rs767426287 Chr.4: 46058525-46058525 on Build GRCh38
rs1030082627 Chr.4: 46058283-46058283 on Build GRCh38
rs773691921 Chr.4: 46058221-46058221 on Build GRCh38
rs373443309 Chr.4: 46058522-46058522 on Build GRCh38
rs147493703 Chr.4: 46058238-46058238 on Build GRCh38
rs142748494 Chr.4: 46058160-46058160 on Build GRCh38
rs778424074 Chr.4: 46058331-46058331 on Build GRCh38
rs753005289 Chr.4: 46058280-46058280 on Build GRCh38
rs564336520 Chr.4: 46058507-46058507 on Build GRCh38
rs372512898 Chr.4: 46058271-46058271 on Build GRCh38
rs745922740 Chr.4: 46058275-46058277 on Build GRCh38
rs776187307 Chr.4: 46058509-46058509 on Build GRCh38
rs768534395 Chr.4: 46058327-46058327 on Build GRCh38
rs373278401 Chr.4: 46058464-46058464 on Build GRCh38
rs778778454 Chr.4: 46058406-46058406 on Build GRCh38
rs1012870401 Chr.4: 46058106-46058106 on Build GRCh38
rs753877038 Chr.4: 46058400-46058400 on Build GRCh38
rs778898204 Chr.4: 46058201-46058207 on Build GRCh38
rs745868816 Chr.4: 46058180-46058180 on Build GRCh38
rs754712499 Chr.4: 46058404-46058404 on Build GRCh38

View Full Product Details