Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46923913-46924417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA4
Entrez Gene ID
2557
Gene Name
gamma-aminobutyric acid type A receptor alpha4 subunit
Gene Aliases
-
UniGene
Hs.248112
Gene Chromosome Location
Chr.4: 46918900-46994407 on Build GRCh38
Transcripts
NM_000809.3 NM_001204266.1 NM_001204267.1

SNP Detail

SNP ID Chromosome Location
rs942056723 Chr.4: 46924168-46924168 on Build GRCh38
rs1047808416 Chr.4: 46924143-46924143 on Build GRCh38
rs1052583916 Chr.4: 46924377-46924377 on Build GRCh38
rs773004155 Chr.4: 46923938-46923938 on Build GRCh38
rs892221084 Chr.4: 46924389-46924389 on Build GRCh38
rs1038670813 Chr.4: 46924193-46924193 on Build GRCh38
rs1039497062 Chr.4: 46924335-46924335 on Build GRCh38
rs903581762 Chr.4: 46924045-46924045 on Build GRCh38
rs767094484 Chr.4: 46924372-46924372 on Build GRCh38
rs1000559525 Chr.4: 46924313-46924313 on Build GRCh38
rs933283938 Chr.4: 46924310-46924310 on Build GRCh38
rs545804001 Chr.4: 46924246-46924246 on Build GRCh38
rs542407714 Chr.4: 46924212-46924212 on Build GRCh38
rs942631869 Chr.4: 46924330-46924330 on Build GRCh38
rs766254619 Chr.4: 46923982-46923982 on Build GRCh38
rs972597609 Chr.4: 46924239-46924239 on Build GRCh38
rs912444419 Chr.4: 46924320-46924320 on Build GRCh38
rs1020592917 Chr.4: 46924038-46924038 on Build GRCh38
rs896323093 Chr.4: 46924219-46924219 on Build GRCh38
rs146064217 Chr.4: 46924267-46924267 on Build GRCh38
rs759568699 Chr.4: 46924211-46924211 on Build GRCh38
rs975310922 Chr.4: 46924341-46924341 on Build GRCh38
rs919257725 Chr.4: 46924263-46924263 on Build GRCh38
rs891051668 Chr.4: 46923984-46923984 on Build GRCh38
rs933448610 Chr.4: 46924139-46924139 on Build GRCh38
rs1020728813 Chr.4: 46924253-46924253 on Build GRCh38
rs991816091 Chr.4: 46924035-46924035 on Build GRCh38
rs1004499708 Chr.4: 46923972-46923972 on Build GRCh38
rs965022496 Chr.4: 46924321-46924321 on Build GRCh38
rs960473311 Chr.4: 46924006-46924006 on Build GRCh38
rs753409501 Chr.4: 46924241-46924241 on Build GRCh38
rs188827709 Chr.4: 46924368-46924368 on Build GRCh38
rs77333767 Chr.4: 46924096-46924096 on Build GRCh38
rs774477281 Chr.4: 46924135-46924135 on Build GRCh38
rs762901377 Chr.4: 46923983-46923983 on Build GRCh38
rs963439929 Chr.4: 46924231-46924231 on Build GRCh38
rs183396596 Chr.4: 46924324-46924324 on Build GRCh38
rs974965655 Chr.4: 46924080-46924080 on Build GRCh38
rs987490627 Chr.4: 46924316-46924316 on Build GRCh38
rs575055301 Chr.4: 46924204-46924204 on Build GRCh38
rs892163216 Chr.4: 46924071-46924071 on Build GRCh38
rs770959948 Chr.4: 46923992-46923992 on Build GRCh38
rs1030646008 Chr.4: 46924067-46924067 on Build GRCh38
rs942497150 Chr.4: 46923989-46923989 on Build GRCh38
rs887026137 Chr.4: 46923971-46923971 on Build GRCh38
rs577162476 Chr.4: 46924008-46924008 on Build GRCh38
rs760381436 Chr.4: 46923968-46923968 on Build GRCh38
rs1000598126 Chr.4: 46924058-46924058 on Build GRCh38
rs768765786 Chr.4: 46923937-46923937 on Build GRCh38
rs966753861 Chr.4: 46924296-46924296 on Build GRCh38
rs191185367 Chr.4: 46924145-46924145 on Build GRCh38
rs142798105 Chr.4: 46924216-46924216 on Build GRCh38

View Full Product Details