Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47943158-47943699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
UniGene
Hs.1323
Gene Chromosome Location
Chr.4: 47935015-48016718 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Gene Symbol
LOC101927157
Entrez Gene ID
101927157
Gene Name
uncharacterized LOC101927157
Gene Aliases
-
UniGene
Hs.134190
Gene Chromosome Location
Chr.4: 47914227-47990737 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6231403 c.346T>A p.S116T Chr.4: 47943284-47943284 on Build GRCh38
3760650 c.352G>A p.D118N Chr.4: 47943278-47943278 on Build GRCh38
79345 c.347C>G p.S116* Chr.4: 47943283-47943283 on Build GRCh38
6331484 c.597G>A p.K199K Chr.4: 47943240-47943240 on Build GRCh38
4550755 c.301G>A p.E101K Chr.4: 47943411-47943411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867857102 Chr.4: 47943222-47943222 on Build GRCh38
rs769774868 Chr.4: 47943190-47943190 on Build GRCh38
rs76061451 Chr.4: 47943400-47943400 on Build GRCh38
rs1023306743 Chr.4: 47943407-47943407 on Build GRCh38
rs748314222 Chr.4: 47943276-47943276 on Build GRCh38
rs147591591 Chr.4: 47943258-47943258 on Build GRCh38
rs868859799 Chr.4: 47943623-47943623 on Build GRCh38
rs774237739 Chr.4: 47943326-47943326 on Build GRCh38
rs1043046270 Chr.4: 47943184-47943184 on Build GRCh38
rs771862788 Chr.4: 47943483-47943483 on Build GRCh38
rs573327416 Chr.4: 47943506-47943506 on Build GRCh38
rs755705235 Chr.4: 47943262-47943262 on Build GRCh38
rs28642966 Chr.4: 47943278-47943278 on Build GRCh38
rs761590177 Chr.4: 47943242-47943242 on Build GRCh38
rs767146100 Chr.4: 47943362-47943362 on Build GRCh38
rs867345447 Chr.4: 47943370-47943370 on Build GRCh38
rs969082016 Chr.4: 47943418-47943419 on Build GRCh38
rs773862362 Chr.4: 47943238-47943238 on Build GRCh38
rs1033952438 Chr.4: 47943661-47943661 on Build GRCh38
rs747049717 Chr.4: 47943282-47943282 on Build GRCh38
rs913453170 Chr.4: 47943523-47943523 on Build GRCh38
rs959230023 Chr.4: 47943673-47943673 on Build GRCh38
rs1057518486 Chr.4: 47943389-47943389 on Build GRCh38
rs769489990 Chr.4: 47943213-47943214 on Build GRCh38
rs541687198 Chr.4: 47943660-47943660 on Build GRCh38
rs771001514 Chr.4: 47943301-47943301 on Build GRCh38
rs202182638 Chr.4: 47943343-47943343 on Build GRCh38
rs1041639282 Chr.4: 47943518-47943518 on Build GRCh38
rs893525961 Chr.4: 47943571-47943571 on Build GRCh38
rs773033454 Chr.4: 47943377-47943377 on Build GRCh38
rs372983495 Chr.4: 47943618-47943618 on Build GRCh38
rs79647861 Chr.4: 47943408-47943408 on Build GRCh38
rs766955357 Chr.4: 47943264-47943264 on Build GRCh38
rs901278121 Chr.4: 47943662-47943662 on Build GRCh38
rs766903165 Chr.4: 47943244-47943244 on Build GRCh38
rs952618231 Chr.4: 47943372-47943372 on Build GRCh38
rs906453903 Chr.4: 47943652-47943652 on Build GRCh38
rs889829783 Chr.4: 47943245-47943245 on Build GRCh38
rs932427749 Chr.4: 47943563-47943563 on Build GRCh38
rs763963244 Chr.4: 47943232-47943232 on Build GRCh38
rs749281255 Chr.4: 47943309-47943309 on Build GRCh38
rs121909600 Chr.4: 47943203-47943203 on Build GRCh38
rs766004809 Chr.4: 47943399-47943399 on Build GRCh38
rs539600817 Chr.4: 47943269-47943269 on Build GRCh38
rs369466685 Chr.4: 47943585-47943585 on Build GRCh38
rs931322990 Chr.4: 47943479-47943479 on Build GRCh38
rs540653917 Chr.4: 47943578-47943578 on Build GRCh38
rs775436707 Chr.4: 47943218-47943218 on Build GRCh38
rs374429703 Chr.4: 47943579-47943579 on Build GRCh38
rs755244560 Chr.4: 47943270-47943270 on Build GRCh38
rs772595474 Chr.4: 47943241-47943241 on Build GRCh38
rs989008498 Chr.4: 47943519-47943519 on Build GRCh38
rs758688361 Chr.4: 47943277-47943277 on Build GRCh38
rs868453867 Chr.4: 47943254-47943254 on Build GRCh38
rs755575913 Chr.4: 47943412-47943412 on Build GRCh38
rs140354960 Chr.4: 47943311-47943311 on Build GRCh38
rs1053524663 Chr.4: 47943564-47943564 on Build GRCh38
rs944274645 Chr.4: 47943500-47943500 on Build GRCh38
rs6819506 Chr.4: 47943438-47943438 on Build GRCh38
rs775347192 Chr.4: 47943305-47943305 on Build GRCh38
rs760198986 Chr.4: 47943394-47943394 on Build GRCh38
rs576510245 Chr.4: 47943319-47943319 on Build GRCh38
rs11940319 Chr.4: 47943653-47943653 on Build GRCh38
rs938829476 Chr.4: 47943513-47943513 on Build GRCh38
rs899981489 Chr.4: 47943524-47943524 on Build GRCh38
rs908807841 Chr.4: 47943488-47943488 on Build GRCh38

View Full Product Details