Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53379101-53379642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125088 c.105G>A p.R35R Chr.4: 53379244-53379244 on Build GRCh38
6664177 c.131-5G>C p.? Chr.4: 53379220-53379220 on Build GRCh38
6041226 c.134_146del13 p.N46fs*29 Chr.4: 53379228-53379240 on Build GRCh38
6664178 c.86-5G>C p.? Chr.4: 53379220-53379220 on Build GRCh38
6031743 c.170+9A>T p.? Chr.4: 53379273-53379273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140615869 Chr.4: 53379294-53379294 on Build GRCh38
rs761724914 Chr.4: 53379299-53379299 on Build GRCh38
rs10011631 Chr.4: 53379332-53379332 on Build GRCh38
rs369434354 Chr.4: 53379291-53379291 on Build GRCh38
rs370747439 Chr.4: 53379171-53379171 on Build GRCh38
rs367710840 Chr.4: 53379237-53379237 on Build GRCh38
rs183792321 Chr.4: 53379392-53379392 on Build GRCh38
rs368745990 Chr.4: 53379216-53379216 on Build GRCh38
rs751392086 Chr.4: 53379191-53379191 on Build GRCh38
rs772403593 Chr.4: 53379191-53379191 on Build GRCh38
rs545307624 Chr.4: 53379614-53379614 on Build GRCh38
rs754483375 Chr.4: 53379313-53379313 on Build GRCh38
rs568098579 Chr.4: 53379437-53379437 on Build GRCh38
rs765896165 Chr.4: 53379310-53379310 on Build GRCh38
rs188184505 Chr.4: 53379516-53379516 on Build GRCh38
rs748416079 Chr.4: 53379159-53379159 on Build GRCh38
rs902885828 Chr.4: 53379346-53379346 on Build GRCh38
rs547204034 Chr.4: 53379320-53379320 on Build GRCh38
rs767365777 Chr.4: 53379300-53379300 on Build GRCh38
rs999849688 Chr.4: 53379378-53379378 on Build GRCh38
rs35209549 Chr.4: 53379175-53379175 on Build GRCh38
rs987553837 Chr.4: 53379528-53379528 on Build GRCh38
rs778273802 Chr.4: 53379283-53379283 on Build GRCh38
rs763878554 Chr.4: 53379182-53379182 on Build GRCh38
rs755528845 Chr.4: 53379221-53379221 on Build GRCh38
rs1471597 Chr.4: 53379582-53379582 on Build GRCh38
rs772748524 Chr.4: 53379366-53379366 on Build GRCh38
rs868523649 Chr.4: 53379187-53379187 on Build GRCh38
rs773777761 Chr.4: 53379204-53379207 on Build GRCh38
rs371676507 Chr.4: 53379260-53379260 on Build GRCh38
rs542366890 Chr.4: 53379506-53379506 on Build GRCh38
rs774195744 Chr.4: 53379298-53379298 on Build GRCh38
rs778459581 Chr.4: 53379173-53379173 on Build GRCh38
rs199730748 Chr.4: 53379232-53379232 on Build GRCh38
rs773612742 Chr.4: 53379170-53379170 on Build GRCh38
rs772459076 Chr.4: 53379161-53379161 on Build GRCh38
rs113914235 Chr.4: 53379286-53379286 on Build GRCh38
rs1471596 Chr.4: 53379309-53379309 on Build GRCh38
rs559312701 Chr.4: 53379389-53379389 on Build GRCh38
rs866427712 Chr.4: 53379175-53379178 on Build GRCh38
rs552704240 Chr.4: 53379588-53379588 on Build GRCh38
rs79100349 Chr.4: 53379379-53379379 on Build GRCh38
rs770116385 Chr.4: 53379193-53379196 on Build GRCh38
rs766187580 Chr.4: 53379408-53379408 on Build GRCh38
rs963940604 Chr.4: 53379167-53379168 on Build GRCh38
rs996704707 Chr.4: 53379440-53379440 on Build GRCh38
rs554995229 Chr.4: 53379489-53379489 on Build GRCh38
rs750135082 Chr.4: 53379211-53379211 on Build GRCh38
rs748599096 Chr.4: 53379129-53379131 on Build GRCh38
rs767269409 Chr.4: 53379207-53379207 on Build GRCh38
rs749779629 Chr.4: 53379225-53379230 on Build GRCh38
rs772922950 Chr.4: 53379308-53379308 on Build GRCh38
rs771009502 Chr.4: 53379247-53379249 on Build GRCh38
rs550587330 Chr.4: 53379282-53379282 on Build GRCh38
rs1014952023 Chr.4: 53379569-53379569 on Build GRCh38
rs761555170 Chr.4: 53379192-53379192 on Build GRCh38
rs375375636 Chr.4: 53379288-53379288 on Build GRCh38
rs1021099 Chr.4: 53379608-53379608 on Build GRCh38
rs776567006 Chr.4: 53379174-53379174 on Build GRCh38
rs954818637 Chr.4: 53379496-53379496 on Build GRCh38
rs949799670 Chr.4: 53379316-53379316 on Build GRCh38
rs747537858 Chr.4: 53379178-53379178 on Build GRCh38
rs1029874801 Chr.4: 53379447-53379447 on Build GRCh38
rs747056901 Chr.4: 53379601-53379602 on Build GRCh38
rs375164549 Chr.4: 53379125-53379125 on Build GRCh38
rs372183751 Chr.4: 53379220-53379220 on Build GRCh38
rs747304988 Chr.4: 53379285-53379285 on Build GRCh38
rs779351623 Chr.4: 53379155-53379155 on Build GRCh38

View Full Product Details