Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53383324-53383871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447049 c.290A>C p.H97P Chr.4: 53383834-53383834 on Build GRCh38
6909924 c.229-1G>T p.? Chr.4: 53383772-53383772 on Build GRCh38
4125090 c.268G>A p.D90N Chr.4: 53383812-53383812 on Build GRCh38
5664215 c.247G>T p.V83F Chr.4: 53383836-53383836 on Build GRCh38
4125092 c.314C>T p.T105M Chr.4: 53383858-53383858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781598231 Chr.4: 53383741-53383741 on Build GRCh38
rs946392888 Chr.4: 53383640-53383640 on Build GRCh38
rs749315906 Chr.4: 53383787-53383787 on Build GRCh38
rs535065508 Chr.4: 53383489-53383489 on Build GRCh38
rs768553857 Chr.4: 53383414-53383414 on Build GRCh38
rs913632431 Chr.4: 53383639-53383639 on Build GRCh38
rs533203943 Chr.4: 53383424-53383424 on Build GRCh38
rs746143689 Chr.4: 53383747-53383747 on Build GRCh38
rs775552354 Chr.4: 53383754-53383757 on Build GRCh38
rs181920644 Chr.4: 53383470-53383470 on Build GRCh38
rs759119551 Chr.4: 53383841-53383841 on Build GRCh38
rs767730400 Chr.4: 53383746-53383746 on Build GRCh38
rs537715289 Chr.4: 53383667-53383667 on Build GRCh38
rs774541640 Chr.4: 53383704-53383704 on Build GRCh38
rs756291969 Chr.4: 53383769-53383769 on Build GRCh38
rs557356309 Chr.4: 53383669-53383669 on Build GRCh38
rs776485515 Chr.4: 53383834-53383834 on Build GRCh38
rs998262695 Chr.4: 53383383-53383383 on Build GRCh38
rs879494652 Chr.4: 53383593-53383593 on Build GRCh38
rs578126526 Chr.4: 53383636-53383636 on Build GRCh38
rs565432427 Chr.4: 53383560-53383560 on Build GRCh38
rs184801542 Chr.4: 53383545-53383545 on Build GRCh38
rs189313408 Chr.4: 53383568-53383568 on Build GRCh38
rs34716081 Chr.4: 53383392-53383392 on Build GRCh38
rs574415896 Chr.4: 53383697-53383697 on Build GRCh38
rs763742713 Chr.4: 53383671-53383672 on Build GRCh38
rs558290441 Chr.4: 53383635-53383635 on Build GRCh38
rs549825943 Chr.4: 53383429-53383429 on Build GRCh38
rs1035227603 Chr.4: 53383553-53383554 on Build GRCh38
rs370672369 Chr.4: 53383792-53383792 on Build GRCh38
rs757603584 Chr.4: 53383730-53383730 on Build GRCh38
rs374375670 Chr.4: 53383819-53383819 on Build GRCh38
rs111840938 Chr.4: 53383473-53383473 on Build GRCh38
rs142716571 Chr.4: 53383421-53383421 on Build GRCh38
rs112601279 Chr.4: 53383471-53383471 on Build GRCh38
rs188826662 Chr.4: 53383398-53383398 on Build GRCh38
rs547408854 Chr.4: 53383699-53383699 on Build GRCh38
rs375673559 Chr.4: 53383706-53383706 on Build GRCh38
rs547606866 Chr.4: 53383402-53383402 on Build GRCh38
rs943248259 Chr.4: 53383676-53383676 on Build GRCh38
rs772025539 Chr.4: 53383788-53383788 on Build GRCh38
rs573336752 Chr.4: 53383727-53383727 on Build GRCh38
rs770685150 Chr.4: 53383829-53383829 on Build GRCh38

View Full Product Details