Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53389419-53389917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3604445 c.358C>T p.L120F Chr.4: 53389834-53389834 on Build GRCh38
1618904 c.338A>G p.Y113C Chr.4: 53389814-53389814 on Build GRCh38
3604446 c.313C>T p.L105F Chr.4: 53389834-53389834 on Build GRCh38
5577461 c.377G>A p.G126E Chr.4: 53389853-53389853 on Build GRCh38
5577462 c.332G>A p.G111E Chr.4: 53389853-53389853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765037352 Chr.4: 53389755-53389755 on Build GRCh38
rs75910510 Chr.4: 53389700-53389700 on Build GRCh38
rs763974409 Chr.4: 53389784-53389784 on Build GRCh38
rs755184908 Chr.4: 53389857-53389857 on Build GRCh38
rs773931448 Chr.4: 53389801-53389801 on Build GRCh38
rs149720675 Chr.4: 53389455-53389455 on Build GRCh38
rs774671103 Chr.4: 53389496-53389496 on Build GRCh38
rs3762878 Chr.4: 53389626-53389626 on Build GRCh38
rs998475747 Chr.4: 53389530-53389530 on Build GRCh38
rs375979439 Chr.4: 53389466-53389467 on Build GRCh38
rs398083072 Chr.4: 53389465-53389465 on Build GRCh38
rs539038956 Chr.4: 53389643-53389643 on Build GRCh38
rs772743149 Chr.4: 53389546-53389546 on Build GRCh38
rs980923132 Chr.4: 53389611-53389611 on Build GRCh38
rs3762877 Chr.4: 53389599-53389599 on Build GRCh38
rs766705266 Chr.4: 53389834-53389834 on Build GRCh38
rs753841262 Chr.4: 53389784-53389786 on Build GRCh38
rs202159892 Chr.4: 53389784-53389784 on Build GRCh38
rs189310322 Chr.4: 53389892-53389892 on Build GRCh38
rs764871693 Chr.4: 53389793-53389797 on Build GRCh38
rs762735943 Chr.4: 53389821-53389821 on Build GRCh38
rs113276070 Chr.4: 53389523-53389523 on Build GRCh38
rs1006846707 Chr.4: 53389465-53389465 on Build GRCh38
rs768339642 Chr.4: 53389797-53389797 on Build GRCh38
rs761484033 Chr.4: 53389732-53389732 on Build GRCh38
rs1016696267 Chr.4: 53389589-53389589 on Build GRCh38
rs781146855 Chr.4: 53389876-53389876 on Build GRCh38
rs1039744874 Chr.4: 53389470-53389470 on Build GRCh38
rs988095567 Chr.4: 53389634-53389634 on Build GRCh38
rs910381323 Chr.4: 53389697-53389697 on Build GRCh38
rs3762879 Chr.4: 53389720-53389720 on Build GRCh38
rs765309460 Chr.4: 53389859-53389859 on Build GRCh38
rs77992820 Chr.4: 53389789-53389789 on Build GRCh38
rs762673974 Chr.4: 53389793-53389793 on Build GRCh38
rs768154163 Chr.4: 53389679-53389679 on Build GRCh38
rs946558438 Chr.4: 53389651-53389651 on Build GRCh38
rs769944014 Chr.4: 53389667-53389670 on Build GRCh38
rs766198822 Chr.4: 53389848-53389848 on Build GRCh38
rs746215199 Chr.4: 53389527-53389527 on Build GRCh38
rs543041222 Chr.4: 53389650-53389650 on Build GRCh38
rs762411553 Chr.4: 53389587-53389587 on Build GRCh38
rs904580689 Chr.4: 53389461-53389461 on Build GRCh38
rs951544897 Chr.4: 53389548-53389548 on Build GRCh38
rs757166849 Chr.4: 53389870-53389870 on Build GRCh38
rs560146448 Chr.4: 53389456-53389457 on Build GRCh38
rs535187737 Chr.4: 53389728-53389728 on Build GRCh38
rs760585291 Chr.4: 53389783-53389784 on Build GRCh38
rs558153131 Chr.4: 53389815-53389815 on Build GRCh38
rs960635404 Chr.4: 53389633-53389633 on Build GRCh38
rs527553317 Chr.4: 53389577-53389577 on Build GRCh38
rs143506497 Chr.4: 53389824-53389824 on Build GRCh38
rs752768017 Chr.4: 53389864-53389864 on Build GRCh38
rs775323864 Chr.4: 53389790-53389790 on Build GRCh38
rs149622470 Chr.4: 53389747-53389747 on Build GRCh38
rs143587271 Chr.4: 53389453-53389453 on Build GRCh38
rs762238961 Chr.4: 53389807-53389807 on Build GRCh38
rs34929012 Chr.4: 53389456-53389456 on Build GRCh38
rs1005694298 Chr.4: 53389570-53389570 on Build GRCh38
rs757196045 Chr.4: 53389790-53389793 on Build GRCh38
rs750783002 Chr.4: 53389820-53389820 on Build GRCh38
rs397993363 Chr.4: 53389466-53389466 on Build GRCh38

View Full Product Details