Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 53399604-53400110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIP1L1
Entrez Gene ID
81608
Gene Name
factor interacting with PAPOLA and CPSF1
Gene Aliases
FIP1, Rhe, hFip1
UniGene
Hs.555109
Gene Chromosome Location
Chr.4: 53377572-53462611 on Build GRCh38
Transcripts
NM_001134937.1 NM_001134938.1 NM_030917.3 XM_005265769.4 XM_005265773.4 XM_005265774.4 XM_005265778.4 XM_005265779.4 XM_005265781.4 XM_005265782.4 XM_017008662.1 XM_017008663.1 XM_017008664.1 XM_017008665.1 XM_017008666.1 XM_017008667.1 XM_017008668.1 XM_017008669.1 XM_017008670.1 XM_017008671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3993715 c.770+1G>C p.? Chr.4: 53399840-53399840 on Build GRCh38
6664175 c.812G>T p.S271I Chr.4: 53399836-53399836 on Build GRCh38
7368312 c.714G>T p.Q238H Chr.4: 53399738-53399738 on Build GRCh38
5758307 c.724G>A p.G242R Chr.4: 53399748-53399748 on Build GRCh38
3301146 c.711G>A p.Q237Q Chr.4: 53399735-53399735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374275386 Chr.4: 53399688-53399688 on Build GRCh38
rs760370262 Chr.4: 53399871-53399871 on Build GRCh38
rs914534334 Chr.4: 53399894-53399894 on Build GRCh38
rs151105925 Chr.4: 53399777-53399777 on Build GRCh38
rs771016690 Chr.4: 53399679-53399679 on Build GRCh38
rs542567872 Chr.4: 53399873-53399873 on Build GRCh38
rs1018963115 Chr.4: 53399664-53399664 on Build GRCh38
rs149817693 Chr.4: 53399834-53399834 on Build GRCh38
rs868153388 Chr.4: 53399785-53399785 on Build GRCh38
rs587778363 Chr.4: 53399808-53399808 on Build GRCh38
rs1006411840 Chr.4: 53400067-53400067 on Build GRCh38
rs966409703 Chr.4: 53399678-53399678 on Build GRCh38
rs947280157 Chr.4: 53399904-53399904 on Build GRCh38
rs993944518 Chr.4: 53400018-53400018 on Build GRCh38
rs139886150 Chr.4: 53399833-53399833 on Build GRCh38
rs759117892 Chr.4: 53400045-53400045 on Build GRCh38
rs572704349 Chr.4: 53399822-53399822 on Build GRCh38
rs55709322 Chr.4: 53399691-53399691 on Build GRCh38
rs774155401 Chr.4: 53399850-53399850 on Build GRCh38
rs768525517 Chr.4: 53399849-53399849 on Build GRCh38
rs755442173 Chr.4: 53399766-53399766 on Build GRCh38
rs750059420 Chr.4: 53399715-53399715 on Build GRCh38
rs748472020 Chr.4: 53399803-53399803 on Build GRCh38
rs565006746 Chr.4: 53400059-53400059 on Build GRCh38
rs571126949 Chr.4: 53399650-53399650 on Build GRCh38
rs761468740 Chr.4: 53399699-53399699 on Build GRCh38
rs770135904 Chr.4: 53399809-53399809 on Build GRCh38
rs771831122 Chr.4: 53399864-53399864 on Build GRCh38
rs374455355 Chr.4: 53399872-53399872 on Build GRCh38
rs771383747 Chr.4: 53399717-53399718 on Build GRCh38
rs747234438 Chr.4: 53399816-53399816 on Build GRCh38
rs749187058 Chr.4: 53399842-53399842 on Build GRCh38
rs535057000 Chr.4: 53399994-53399994 on Build GRCh38
rs889598114 Chr.4: 53399979-53399979 on Build GRCh38
rs17082702 Chr.4: 53400012-53400012 on Build GRCh38
rs779469853 Chr.4: 53399800-53399802 on Build GRCh38
rs771542837 Chr.4: 53400008-53400008 on Build GRCh38
rs369079997 Chr.4: 53399716-53399717 on Build GRCh38
rs748518306 Chr.4: 53399668-53399668 on Build GRCh38
rs185025021 Chr.4: 53399865-53399865 on Build GRCh38
rs754368971 Chr.4: 53399756-53399756 on Build GRCh38
rs142996922 Chr.4: 53399713-53399715 on Build GRCh38
rs1049620127 Chr.4: 53399974-53399974 on Build GRCh38
rs1053047813 Chr.4: 53399877-53399877 on Build GRCh38
rs746008942 Chr.4: 53399806-53399808 on Build GRCh38
rs767108325 Chr.4: 53399701-53399701 on Build GRCh38
rs765899567 Chr.4: 53399735-53399735 on Build GRCh38
rs760139722 Chr.4: 53399878-53399878 on Build GRCh38
rs1026264053 Chr.4: 53400044-53400044 on Build GRCh38
rs545138623 Chr.4: 53400043-53400043 on Build GRCh38
rs778051742 Chr.4: 53399717-53399717 on Build GRCh38
rs1008036591 Chr.4: 53399657-53399657 on Build GRCh38
rs762937480 Chr.4: 53399685-53399685 on Build GRCh38
rs113967782 Chr.4: 53400072-53400072 on Build GRCh38
rs536883056 Chr.4: 53399863-53399863 on Build GRCh38
rs889240745 Chr.4: 53399629-53399629 on Build GRCh38
rs755626892 Chr.4: 53399732-53399732 on Build GRCh38
rs775341686 Chr.4: 53399682-53399682 on Build GRCh38

View Full Product Details