Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55959558-55960066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP135
Entrez Gene ID
9662
Gene Name
centrosomal protein 135
Gene Aliases
CEP4, KIAA0635, MCPH8
UniGene
Hs.518767
Gene Chromosome Location
Chr.4: 55948808-56033363 on Build GRCh38
Transcripts
NM_025009.4 XM_005265788.3 XM_006714055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528558 c.643C>T p.Q215* Chr.4: 55959710-55959710 on Build GRCh38
6991557 c.668T>C p.M223T Chr.4: 55959735-55959735 on Build GRCh38
4671585 c.687C>T p.D229D Chr.4: 55959754-55959754 on Build GRCh38
6991556 c.668T>C p.M223T Chr.4: 55959735-55959735 on Build GRCh38
1200770 c.679G>A p.V227M Chr.4: 55959746-55959746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749639731 Chr.4: 55959820-55959820 on Build GRCh38
rs766224211 Chr.4: 55959771-55959771 on Build GRCh38
rs928065367 Chr.4: 55959895-55959895 on Build GRCh38
rs568550315 Chr.4: 55959963-55959963 on Build GRCh38
rs144775748 Chr.4: 55959622-55959622 on Build GRCh38
rs1055740133 Chr.4: 55959912-55959912 on Build GRCh38
rs886470976 Chr.4: 55960025-55960025 on Build GRCh38
rs77701414 Chr.4: 55959849-55959849 on Build GRCh38
rs765158583 Chr.4: 55959794-55959794 on Build GRCh38
rs746305130 Chr.4: 55959636-55959636 on Build GRCh38
rs200179312 Chr.4: 55959650-55959650 on Build GRCh38
rs1046528093 Chr.4: 55959869-55959869 on Build GRCh38
rs535286763 Chr.4: 55959766-55959766 on Build GRCh38
rs749151464 Chr.4: 55959675-55959675 on Build GRCh38
rs929841020 Chr.4: 55959707-55959728 on Build GRCh38
rs1043227598 Chr.4: 55959900-55959900 on Build GRCh38
rs767993141 Chr.4: 55959734-55959734 on Build GRCh38
rs776379681 Chr.4: 55959772-55959772 on Build GRCh38
rs1047346388 Chr.4: 55959995-55959995 on Build GRCh38
rs150331261 Chr.4: 55959705-55959705 on Build GRCh38
rs761466910 Chr.4: 55959661-55959661 on Build GRCh38
rs1013769197 Chr.4: 55959994-55959994 on Build GRCh38
rs140961633 Chr.4: 55959605-55959605 on Build GRCh38
rs964588940 Chr.4: 55959647-55959647 on Build GRCh38
rs770414158 Chr.4: 55959640-55959640 on Build GRCh38
rs757551939 Chr.4: 55959699-55959699 on Build GRCh38
rs758365942 Chr.4: 55959691-55959691 on Build GRCh38
rs749824236 Chr.4: 55959723-55959723 on Build GRCh38
rs746576951 Chr.4: 55959667-55959667 on Build GRCh38
rs531802843 Chr.4: 55959600-55959600 on Build GRCh38
rs1016339697 Chr.4: 55959688-55959688 on Build GRCh38
rs905599870 Chr.4: 55959902-55959902 on Build GRCh38
rs759526475 Chr.4: 55959782-55959782 on Build GRCh38
rs758416884 Chr.4: 55959583-55959583 on Build GRCh38
rs751679914 Chr.4: 55959816-55959816 on Build GRCh38
rs781696190 Chr.4: 55959634-55959634 on Build GRCh38
rs762211566 Chr.4: 55959621-55959621 on Build GRCh38
rs947169659 Chr.4: 55959891-55959891 on Build GRCh38
rs142117607 Chr.4: 55959761-55959761 on Build GRCh38
rs776115990 Chr.4: 55959646-55959646 on Build GRCh38
rs565650078 Chr.4: 55959714-55959715 on Build GRCh38
rs768244275 Chr.4: 55959651-55959651 on Build GRCh38
rs938219084 Chr.4: 55959956-55959956 on Build GRCh38
rs750707873 Chr.4: 55959833-55959833 on Build GRCh38
rs76941356 Chr.4: 55959703-55959703 on Build GRCh38
rs1026328861 Chr.4: 55960002-55960002 on Build GRCh38
rs767819722 Chr.4: 55959831-55959831 on Build GRCh38
rs137876652 Chr.4: 55959753-55959753 on Build GRCh38
rs988496314 Chr.4: 55959796-55959796 on Build GRCh38
rs760518886 Chr.4: 55959680-55959680 on Build GRCh38
rs756386097 Chr.4: 55959839-55959839 on Build GRCh38
rs772985935 Chr.4: 55959677-55959677 on Build GRCh38
rs747722303 Chr.4: 55959746-55959746 on Build GRCh38
rs372748512 Chr.4: 55959694-55959694 on Build GRCh38
rs138715248 Chr.4: 55959687-55959687 on Build GRCh38
rs780747687 Chr.4: 55959715-55959715 on Build GRCh38
rs201936189 Chr.4: 55959741-55959741 on Build GRCh38
rs770788521 Chr.4: 55959720-55959721 on Build GRCh38
rs896749236 Chr.4: 55959968-55959968 on Build GRCh38
rs780386101 Chr.4: 55959718-55959718 on Build GRCh38
rs771698299 Chr.4: 55959747-55959747 on Build GRCh38
rs773825879 Chr.4: 55959652-55959652 on Build GRCh38
rs756587774 Chr.4: 55959713-55959713 on Build GRCh38

View Full Product Details