Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55971154-55971668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP135
Entrez Gene ID
9662
Gene Name
centrosomal protein 135
Gene Aliases
CEP4, KIAA0635, MCPH8
UniGene
Hs.518767
Gene Chromosome Location
Chr.4: 55948808-56033363 on Build GRCh38
Transcripts
NM_025009.4 XM_005265788.3 XM_006714055.2 XM_011534412.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7360843 c.1220G>A p.S407N Chr.4: 55971379-55971379 on Build GRCh38
6626817 c.1222delA p.V410fs*18 Chr.4: 55971381-55971381 on Build GRCh38
6100245 c.1132G>C p.E378Q Chr.4: 55971291-55971291 on Build GRCh38
353284 c.1118A>T p.N373I Chr.4: 55971277-55971277 on Build GRCh38
41149 c.1124G>A p.C375Y Chr.4: 55971283-55971283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372571768 Chr.4: 55971372-55971372 on Build GRCh38
rs747351997 Chr.4: 55971336-55971336 on Build GRCh38
rs377505434 Chr.4: 55971319-55971319 on Build GRCh38
rs774034582 Chr.4: 55971413-55971413 on Build GRCh38
rs755527911 Chr.4: 55971451-55971452 on Build GRCh38
rs757729898 Chr.4: 55971345-55971345 on Build GRCh38
rs1029703325 Chr.4: 55971622-55971622 on Build GRCh38
rs772764058 Chr.4: 55971540-55971540 on Build GRCh38
rs754720085 Chr.4: 55971235-55971235 on Build GRCh38
rs892527308 Chr.4: 55971629-55971629 on Build GRCh38
rs1045786626 Chr.4: 55971360-55971360 on Build GRCh38
rs1007930648 Chr.4: 55971481-55971484 on Build GRCh38
rs772964766 Chr.4: 55971421-55971421 on Build GRCh38
rs950086071 Chr.4: 55971342-55971342 on Build GRCh38
rs964001437 Chr.4: 55971214-55971214 on Build GRCh38
rs774951133 Chr.4: 55971246-55971246 on Build GRCh38
rs1039916033 Chr.4: 55971566-55971566 on Build GRCh38
rs890782163 Chr.4: 55971480-55971481 on Build GRCh38
rs752081636 Chr.4: 55971245-55971248 on Build GRCh38
rs769372576 Chr.4: 55971244-55971244 on Build GRCh38
rs573832516 Chr.4: 55971608-55971608 on Build GRCh38
rs149118125 Chr.4: 55971206-55971209 on Build GRCh38
rs766689006 Chr.4: 55971232-55971234 on Build GRCh38
rs566483169 Chr.4: 55971462-55971462 on Build GRCh38
rs1047206885 Chr.4: 55971531-55971531 on Build GRCh38
rs764374793 Chr.4: 55971459-55971459 on Build GRCh38
rs62641664 Chr.4: 55971379-55971379 on Build GRCh38
rs774120805 Chr.4: 55971282-55971282 on Build GRCh38
rs181459579 Chr.4: 55971618-55971618 on Build GRCh38
rs767320674 Chr.4: 55971416-55971416 on Build GRCh38
rs373032199 Chr.4: 55971312-55971312 on Build GRCh38
rs769631189 Chr.4: 55971500-55971500 on Build GRCh38
rs775062669 Chr.4: 55971368-55971368 on Build GRCh38
rs1025924507 Chr.4: 55971480-55971483 on Build GRCh38
rs755240152 Chr.4: 55971315-55971317 on Build GRCh38
rs745332925 Chr.4: 55971243-55971243 on Build GRCh38
rs754964499 Chr.4: 55971297-55971297 on Build GRCh38
rs373490268 Chr.4: 55971268-55971268 on Build GRCh38
rs748812802 Chr.4: 55971595-55971595 on Build GRCh38
rs747441980 Chr.4: 55971219-55971219 on Build GRCh38
rs189415989 Chr.4: 55971515-55971515 on Build GRCh38
rs748049381 Chr.4: 55971485-55971485 on Build GRCh38
rs544051392 Chr.4: 55971581-55971581 on Build GRCh38
rs752806624 Chr.4: 55971434-55971434 on Build GRCh38
rs550691490 Chr.4: 55971216-55971216 on Build GRCh38
rs557015750 Chr.4: 55971588-55971588 on Build GRCh38
rs760662400 Chr.4: 55971432-55971432 on Build GRCh38
rs763803448 Chr.4: 55971257-55971257 on Build GRCh38
rs964142374 Chr.4: 55971519-55971519 on Build GRCh38
rs763171083 Chr.4: 55971442-55971442 on Build GRCh38
rs1018201964 Chr.4: 55971183-55971183 on Build GRCh38
rs375661378 Chr.4: 55971415-55971415 on Build GRCh38
rs766290384 Chr.4: 55971433-55971433 on Build GRCh38
rs377265085 Chr.4: 55971593-55971593 on Build GRCh38
rs769123333 Chr.4: 55971363-55971363 on Build GRCh38
rs746403996 Chr.4: 55971349-55971349 on Build GRCh38
rs1005830405 Chr.4: 55971560-55971560 on Build GRCh38
rs781746934 Chr.4: 55971348-55971348 on Build GRCh38
rs200550529 Chr.4: 55971454-55971454 on Build GRCh38
rs141602415 Chr.4: 55971292-55971292 on Build GRCh38
rs1056261488 Chr.4: 55971477-55971477 on Build GRCh38
rs538509394 Chr.4: 55971463-55971463 on Build GRCh38
rs558820065 Chr.4: 55971501-55971501 on Build GRCh38
rs905979944 Chr.4: 55971366-55971366 on Build GRCh38
rs761647013 Chr.4: 55971287-55971287 on Build GRCh38
rs771548724 Chr.4: 55971224-55971224 on Build GRCh38
rs371016705 Chr.4: 55971321-55971321 on Build GRCh38
rs998249744 Chr.4: 55971217-55971217 on Build GRCh38
rs925191826 Chr.4: 55971198-55971198 on Build GRCh38
rs915161015 Chr.4: 55971402-55971402 on Build GRCh38
rs765371705 Chr.4: 55971301-55971301 on Build GRCh38
rs938994673 Chr.4: 55971474-55971474 on Build GRCh38
rs762672940 Chr.4: 55971253-55971253 on Build GRCh38
rs777320698 Chr.4: 55971229-55971229 on Build GRCh38

View Full Product Details