Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 65321004-65321523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA5
Entrez Gene ID
2044
Gene Name
EPH receptor A5
Gene Aliases
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4
UniGene
Hs.654492
Gene Chromosome Location
Chr.4: 65319563-65670838 on Build GRCh38
Transcripts
NM_001281765.2 NM_001281766.2 NM_001318761.1 NM_004439.7 NM_182472.4 XM_005265653.3 XM_011531735.2 XM_017007878.1 XM_017007880.1

SNP Detail

SNP ID Chromosome Location
rs73824281 Chr.4: 65321382-65321382 on Build GRCh38
rs386675381 Chr.4: 65321381-65321382 on Build GRCh38
rs985806665 Chr.4: 65321242-65321242 on Build GRCh38
rs868661601 Chr.4: 65321172-65321172 on Build GRCh38
rs998175682 Chr.4: 65321042-65321042 on Build GRCh38
rs140511519 Chr.4: 65321272-65321272 on Build GRCh38
rs1048184835 Chr.4: 65321251-65321251 on Build GRCh38
rs147062149 Chr.4: 65321400-65321400 on Build GRCh38
rs1022373179 Chr.4: 65321187-65321187 on Build GRCh38
rs910878633 Chr.4: 65321301-65321301 on Build GRCh38
rs779107943 Chr.4: 65321473-65321473 on Build GRCh38
rs1022301113 Chr.4: 65321499-65321499 on Build GRCh38
rs906717510 Chr.4: 65321417-65321417 on Build GRCh38
rs372946430 Chr.4: 65321178-65321178 on Build GRCh38
rs760512632 Chr.4: 65321137-65321137 on Build GRCh38
rs918342955 Chr.4: 65321391-65321391 on Build GRCh38
rs571019577 Chr.4: 65321140-65321140 on Build GRCh38
rs900830977 Chr.4: 65321323-65321323 on Build GRCh38
rs766150373 Chr.4: 65321165-65321165 on Build GRCh38
rs955337770 Chr.4: 65321216-65321216 on Build GRCh38
rs960091375 Chr.4: 65321179-65321179 on Build GRCh38
rs115670792 Chr.4: 65321487-65321487 on Build GRCh38
rs929427152 Chr.4: 65321398-65321398 on Build GRCh38
rs751613369 Chr.4: 65321449-65321449 on Build GRCh38
rs1047941676 Chr.4: 65321407-65321407 on Build GRCh38
rs1047971231 Chr.4: 65321110-65321110 on Build GRCh38
rs551374421 Chr.4: 65321171-65321171 on Build GRCh38
rs1038155220 Chr.4: 65321070-65321070 on Build GRCh38
rs906796574 Chr.4: 65321111-65321111 on Build GRCh38
rs929899066 Chr.4: 65321235-65321235 on Build GRCh38
rs777730459 Chr.4: 65321435-65321435 on Build GRCh38
rs1021820832 Chr.4: 65321141-65321141 on Build GRCh38
rs535496237 Chr.4: 65321205-65321205 on Build GRCh38
rs904730905 Chr.4: 65321459-65321459 on Build GRCh38
rs572885391 Chr.4: 65321199-65321199 on Build GRCh38
rs1010375561 Chr.4: 65321120-65321120 on Build GRCh38
rs148294196 Chr.4: 65321096-65321096 on Build GRCh38
rs968755760 Chr.4: 65321144-65321144 on Build GRCh38
rs777501814 Chr.4: 65321296-65321296 on Build GRCh38
rs1003929384 Chr.4: 65321133-65321133 on Build GRCh38
rs568210482 Chr.4: 65321038-65321038 on Build GRCh38
rs1029507187 Chr.4: 65321211-65321211 on Build GRCh38
rs546063430 Chr.4: 65321478-65321478 on Build GRCh38
rs895143485 Chr.4: 65321481-65321481 on Build GRCh38
rs193205873 Chr.4: 65321451-65321451 on Build GRCh38
rs370354587 Chr.4: 65321384-65321384 on Build GRCh38
rs561159150 Chr.4: 65321454-65321454 on Build GRCh38
rs73824279 Chr.4: 65321381-65321381 on Build GRCh38
rs73824278 Chr.4: 65321063-65321063 on Build GRCh38
rs145254769 Chr.4: 65321293-65321293 on Build GRCh38
rs956391497 Chr.4: 65321118-65321118 on Build GRCh38
rs183014781 Chr.4: 65321190-65321190 on Build GRCh38
rs929500863 Chr.4: 65321098-65321098 on Build GRCh38
rs892024133 Chr.4: 65321395-65321395 on Build GRCh38
rs375950346 Chr.4: 65321209-65321209 on Build GRCh38
rs76925001 Chr.4: 65321162-65321162 on Build GRCh38
rs1039469361 Chr.4: 65321321-65321321 on Build GRCh38
rs188758788 Chr.4: 65321207-65321207 on Build GRCh38

View Full Product Details