Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 65321752-65322297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA5
Entrez Gene ID
2044
Gene Name
EPH receptor A5
Gene Aliases
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4
UniGene
Hs.654492
Gene Chromosome Location
Chr.4: 65319563-65670838 on Build GRCh38
Transcripts
NM_001281765.2 NM_001281766.2 NM_001318761.1 NM_004439.7 NM_182472.4 XM_005265653.3 XM_011531735.2 XM_017007878.1 XM_017007880.1

SNP Detail

SNP ID Chromosome Location
rs1016909828 Chr.4: 65322089-65322089 on Build GRCh38
rs996727327 Chr.4: 65322121-65322122 on Build GRCh38
rs538315474 Chr.4: 65322038-65322038 on Build GRCh38
rs890958862 Chr.4: 65321887-65321887 on Build GRCh38
rs962634152 Chr.4: 65321944-65321944 on Build GRCh38
rs979051648 Chr.4: 65322221-65322221 on Build GRCh38
rs28580139 Chr.4: 65322106-65322106 on Build GRCh38
rs967807284 Chr.4: 65322208-65322208 on Build GRCh38
rs898360575 Chr.4: 65322191-65322191 on Build GRCh38
rs192237920 Chr.4: 65322091-65322091 on Build GRCh38
rs1051653203 Chr.4: 65321880-65321880 on Build GRCh38
rs575299242 Chr.4: 65322014-65322014 on Build GRCh38
rs535410557 Chr.4: 65321985-65321985 on Build GRCh38
rs879048232 Chr.4: 65322215-65322215 on Build GRCh38
rs928374638 Chr.4: 65322035-65322035 on Build GRCh38
rs1043727769 Chr.4: 65321786-65321786 on Build GRCh38
rs926167600 Chr.4: 65321903-65321903 on Build GRCh38
rs1007138880 Chr.4: 65321901-65321901 on Build GRCh38
rs960869782 Chr.4: 65322040-65322040 on Build GRCh38
rs999554423 Chr.4: 65321869-65321869 on Build GRCh38
rs1047615783 Chr.4: 65322054-65322054 on Build GRCh38
rs1029494725 Chr.4: 65322160-65322160 on Build GRCh38
rs922430873 Chr.4: 65321782-65321782 on Build GRCh38
rs547261906 Chr.4: 65321875-65321875 on Build GRCh38
rs573187444 Chr.4: 65322249-65322249 on Build GRCh38
rs923937362 Chr.4: 65322132-65322132 on Build GRCh38
rs935376123 Chr.4: 65322138-65322138 on Build GRCh38
rs902548484 Chr.4: 65321816-65321816 on Build GRCh38
rs937566406 Chr.4: 65322272-65322272 on Build GRCh38
rs891214143 Chr.4: 65322172-65322172 on Build GRCh38
rs559642016 Chr.4: 65322239-65322239 on Build GRCh38
rs555350207 Chr.4: 65321986-65321986 on Build GRCh38
rs1025300844 Chr.4: 65321973-65321973 on Build GRCh38
rs995445751 Chr.4: 65321946-65321946 on Build GRCh38
rs545728859 Chr.4: 65322125-65322125 on Build GRCh38
rs1055936906 Chr.4: 65321928-65321928 on Build GRCh38
rs371395978 Chr.4: 65322274-65322274 on Build GRCh38
rs916814000 Chr.4: 65322083-65322083 on Build GRCh38
rs913612324 Chr.4: 65322183-65322184 on Build GRCh38
rs955407990 Chr.4: 65322165-65322165 on Build GRCh38
rs530661549 Chr.4: 65322011-65322011 on Build GRCh38
rs539677637 Chr.4: 65321848-65321848 on Build GRCh38
rs1051147788 Chr.4: 65322158-65322158 on Build GRCh38
rs1006696821 Chr.4: 65322176-65322176 on Build GRCh38
rs4860650 Chr.4: 65321909-65321909 on Build GRCh38
rs1039923414 Chr.4: 65322178-65322178 on Build GRCh38
rs991407127 Chr.4: 65322079-65322079 on Build GRCh38
rs896008327 Chr.4: 65321939-65321939 on Build GRCh38
rs755558631 Chr.4: 65321860-65321860 on Build GRCh38
rs933722732 Chr.4: 65321827-65321827 on Build GRCh38

View Full Product Details