Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70336027-70336539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABS1
Entrez Gene ID
85438
Gene Name
calcium binding protein, spermatid associated 1
Gene Aliases
C4orf35, CLPH, NYD-SP26
UniGene
Hs.120316
Gene Chromosome Location
Chr.4: 70334954-70337116 on Build GRCh38
Transcripts
NM_033122.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249804 c.1124C>A p.T375N Chr.4: 70336163-70336163 on Build GRCh38
5934860 c.1169C>T p.P390L Chr.4: 70336208-70336208 on Build GRCh38
3605427 c.1116G>A p.K372K Chr.4: 70336155-70336155 on Build GRCh38
3605426 c.1010C>T p.S337L Chr.4: 70336049-70336049 on Build GRCh38
7169430 c.1031C>T p.S344F Chr.4: 70336070-70336070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563383918 Chr.4: 70336503-70336503 on Build GRCh38
rs752623263 Chr.4: 70336197-70336197 on Build GRCh38
rs959300672 Chr.4: 70336392-70336392 on Build GRCh38
rs925260963 Chr.4: 70336210-70336210 on Build GRCh38
rs1024785774 Chr.4: 70336426-70336426 on Build GRCh38
rs559093578 Chr.4: 70336407-70336407 on Build GRCh38
rs1036788601 Chr.4: 70336311-70336311 on Build GRCh38
rs945420066 Chr.4: 70336167-70336167 on Build GRCh38
rs765906296 Chr.4: 70336185-70336185 on Build GRCh38
rs774967853 Chr.4: 70336151-70336151 on Build GRCh38
rs772877782 Chr.4: 70336263-70336263 on Build GRCh38
rs773936121 Chr.4: 70336145-70336146 on Build GRCh38
rs757383116 Chr.4: 70336201-70336201 on Build GRCh38
rs191296434 Chr.4: 70336161-70336161 on Build GRCh38
rs959503798 Chr.4: 70336106-70336106 on Build GRCh38
rs753687991 Chr.4: 70336187-70336187 on Build GRCh38
rs13039 Chr.4: 70336226-70336226 on Build GRCh38
rs149882290 Chr.4: 70336092-70336092 on Build GRCh38
rs1014133307 Chr.4: 70336359-70336359 on Build GRCh38
rs759224161 Chr.4: 70336199-70336201 on Build GRCh38
rs201451430 Chr.4: 70336181-70336181 on Build GRCh38
rs890031318 Chr.4: 70336218-70336218 on Build GRCh38
rs567242455 Chr.4: 70336090-70336090 on Build GRCh38
rs926559363 Chr.4: 70336094-70336094 on Build GRCh38
rs779915158 Chr.4: 70336052-70336052 on Build GRCh38
rs754706316 Chr.4: 70336190-70336190 on Build GRCh38
rs1003859529 Chr.4: 70336286-70336286 on Build GRCh38
rs925350578 Chr.4: 70336445-70336445 on Build GRCh38
rs376560270 Chr.4: 70336245-70336245 on Build GRCh38
rs771867150 Chr.4: 70336124-70336124 on Build GRCh38
rs760378146 Chr.4: 70336184-70336184 on Build GRCh38
rs368438643 Chr.4: 70336112-70336112 on Build GRCh38
rs895559363 Chr.4: 70336330-70336330 on Build GRCh38
rs932809537 Chr.4: 70336452-70336452 on Build GRCh38
rs769848096 Chr.4: 70336113-70336113 on Build GRCh38
rs773456105 Chr.4: 70336479-70336479 on Build GRCh38
rs936684744 Chr.4: 70336449-70336449 on Build GRCh38
rs1032903926 Chr.4: 70336425-70336425 on Build GRCh38
rs770080535 Chr.4: 70336209-70336209 on Build GRCh38
rs776902349 Chr.4: 70336101-70336101 on Build GRCh38
rs200977898 Chr.4: 70336255-70336255 on Build GRCh38
rs866735630 Chr.4: 70336304-70336304 on Build GRCh38
rs914022713 Chr.4: 70336432-70336432 on Build GRCh38
rs544820706 Chr.4: 70336438-70336438 on Build GRCh38
rs887485213 Chr.4: 70336285-70336285 on Build GRCh38
rs992625940 Chr.4: 70336143-70336143 on Build GRCh38
rs1041616166 Chr.4: 70336505-70336505 on Build GRCh38
rs115006059 Chr.4: 70336433-70336433 on Build GRCh38
rs1000533156 Chr.4: 70336416-70336417 on Build GRCh38
rs1048088734 Chr.4: 70336280-70336280 on Build GRCh38
rs770962721 Chr.4: 70336097-70336097 on Build GRCh38
rs765285487 Chr.4: 70336339-70336339 on Build GRCh38
rs764377711 Chr.4: 70336201-70336220 on Build GRCh38
rs182928869 Chr.4: 70336338-70336338 on Build GRCh38
rs573600956 Chr.4: 70336279-70336279 on Build GRCh38
rs988068921 Chr.4: 70336428-70336428 on Build GRCh38
rs770456606 Chr.4: 70336056-70336058 on Build GRCh38
rs1023199411 Chr.4: 70336364-70336364 on Build GRCh38
rs745910463 Chr.4: 70336207-70336207 on Build GRCh38
rs369019291 Chr.4: 70336497-70336497 on Build GRCh38
rs780121111 Chr.4: 70336211-70336211 on Build GRCh38
rs1005879607 Chr.4: 70336334-70336334 on Build GRCh38
rs138774071 Chr.4: 70336244-70336244 on Build GRCh38
rs748967987 Chr.4: 70336060-70336060 on Build GRCh38
rs186468035 Chr.4: 70336132-70336132 on Build GRCh38
rs746891726 Chr.4: 70336183-70336183 on Build GRCh38
rs781260280 Chr.4: 70336203-70336203 on Build GRCh38
rs372435553 Chr.4: 70336082-70336082 on Build GRCh38
rs1008833357 Chr.4: 70336224-70336224 on Build GRCh38
rs754178541 Chr.4: 70336235-70336236 on Build GRCh38
rs1042426905 Chr.4: 70336178-70336178 on Build GRCh38
rs140154037 Chr.4: 70336068-70336068 on Build GRCh38
rs958033545 Chr.4: 70336214-70336214 on Build GRCh38
rs868566594 Chr.4: 70336509-70336509 on Build GRCh38
rs369578007 Chr.4: 70336248-70336248 on Build GRCh38
rs764715464 Chr.4: 70336142-70336142 on Build GRCh38
rs372887159 Chr.4: 70336179-70336179 on Build GRCh38
rs911479456 Chr.4: 70336496-70336496 on Build GRCh38

View Full Product Details