Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70599524-70599997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMBN
Entrez Gene ID
258
Gene Name
ameloblastin
Gene Aliases
AI1F
UniGene
Hs.272396
Gene Chromosome Location
Chr.4: 70592258-70607288 on Build GRCh38
Transcripts
NM_016519.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598989 c.232G>A p.G78S Chr.4: 70599584-70599584 on Build GRCh38
5401476 c.251C>T p.S84F Chr.4: 70599603-70599603 on Build GRCh38
7242547 c.235C>G p.L79V Chr.4: 70599587-70599587 on Build GRCh38
6598992 c.195C>T p.Y65Y Chr.4: 70599547-70599547 on Build GRCh38
3918087 c.240C>T p.L80L Chr.4: 70599592-70599592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143941749 Chr.4: 70599894-70599894 on Build GRCh38
rs759822612 Chr.4: 70599681-70599681 on Build GRCh38
rs146148316 Chr.4: 70599561-70599561 on Build GRCh38
rs766493438 Chr.4: 70599668-70599668 on Build GRCh38
rs773146871 Chr.4: 70599650-70599650 on Build GRCh38
rs554370118 Chr.4: 70599876-70599876 on Build GRCh38
rs191765265 Chr.4: 70599835-70599835 on Build GRCh38
rs770460128 Chr.4: 70599625-70599625 on Build GRCh38
rs775414238 Chr.4: 70599639-70599639 on Build GRCh38
rs143795139 Chr.4: 70599584-70599584 on Build GRCh38
rs929808247 Chr.4: 70599913-70599913 on Build GRCh38
rs776557419 Chr.4: 70599627-70599627 on Build GRCh38
rs911911357 Chr.4: 70599860-70599860 on Build GRCh38
rs541599647 Chr.4: 70599870-70599870 on Build GRCh38
rs751027728 Chr.4: 70599692-70599692 on Build GRCh38
rs200620822 Chr.4: 70599578-70599578 on Build GRCh38
rs944765973 Chr.4: 70599890-70599890 on Build GRCh38
rs745638949 Chr.4: 70599632-70599632 on Build GRCh38
rs1015717932 Chr.4: 70599695-70599695 on Build GRCh38
rs989384883 Chr.4: 70599882-70599882 on Build GRCh38
rs1014293600 Chr.4: 70599731-70599731 on Build GRCh38
rs149467865 Chr.4: 70599768-70599768 on Build GRCh38
rs1020409188 Chr.4: 70599766-70599766 on Build GRCh38
rs774473809 Chr.4: 70599555-70599555 on Build GRCh38
rs888080677 Chr.4: 70599667-70599667 on Build GRCh38
rs1034451563 Chr.4: 70599672-70599672 on Build GRCh38
rs777465699 Chr.4: 70599691-70599691 on Build GRCh38
rs756993915 Chr.4: 70599696-70599696 on Build GRCh38
rs1040887902 Chr.4: 70599583-70599583 on Build GRCh38
rs137900627 Chr.4: 70599649-70599649 on Build GRCh38
rs761020179 Chr.4: 70599651-70599651 on Build GRCh38
rs777368202 Chr.4: 70599608-70599608 on Build GRCh38
rs1048764590 Chr.4: 70599940-70599940 on Build GRCh38
rs1013991336 Chr.4: 70599732-70599732 on Build GRCh38
rs184092253 Chr.4: 70599912-70599912 on Build GRCh38
rs986131002 Chr.4: 70599825-70599825 on Build GRCh38
rs776875648 Chr.4: 70599804-70599804 on Build GRCh38
rs765655841 Chr.4: 70599683-70599683 on Build GRCh38
rs1048892888 Chr.4: 70599602-70599602 on Build GRCh38
rs145763811 Chr.4: 70599585-70599585 on Build GRCh38
rs779268740 Chr.4: 70599598-70599598 on Build GRCh38
rs921570386 Chr.4: 70599934-70599934 on Build GRCh38
rs201505714 Chr.4: 70599554-70599554 on Build GRCh38
rs1033604142 Chr.4: 70599781-70599781 on Build GRCh38
rs767521239 Chr.4: 70599562-70599562 on Build GRCh38
rs1033281764 Chr.4: 70599783-70599783 on Build GRCh38
rs866630412 Chr.4: 70599633-70599633 on Build GRCh38
rs758950730 Chr.4: 70599687-70599687 on Build GRCh38
rs538877012 Chr.4: 70599679-70599679 on Build GRCh38
rs895554955 Chr.4: 70599721-70599721 on Build GRCh38
rs758011947 Chr.4: 70599599-70599599 on Build GRCh38
rs746724705 Chr.4: 70599619-70599619 on Build GRCh38
rs754031212 Chr.4: 70599676-70599676 on Build GRCh38
rs769523083 Chr.4: 70599634-70599634 on Build GRCh38
rs953479833 Chr.4: 70599803-70599803 on Build GRCh38
rs186527935 Chr.4: 70599767-70599767 on Build GRCh38
rs762840330 Chr.4: 70599643-70599643 on Build GRCh38
rs779573135 Chr.4: 70599596-70599596 on Build GRCh38
rs748344001 Chr.4: 70599817-70599817 on Build GRCh38
rs536021304 Chr.4: 70599789-70599789 on Build GRCh38
rs1048314022 Chr.4: 70599641-70599641 on Build GRCh38
rs145190129 Chr.4: 70599580-70599580 on Build GRCh38

View Full Product Details