Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70993727-70994212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7410852 c.91+2T>G p.? Chr.4: 70993928-70993928 on Build GRCh38
5978863 c.2T>A p.M1K Chr.4: 70993837-70993837 on Build GRCh38
5021447 c.15C>T p.P5P Chr.4: 70993850-70993850 on Build GRCh38
7204441 c.24C>A p.S8R Chr.4: 70993859-70993859 on Build GRCh38
3826155 c.51G>T p.E17D Chr.4: 70993886-70993886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772840327 Chr.4: 70993993-70993993 on Build GRCh38
rs368749344 Chr.4: 70993893-70993893 on Build GRCh38
rs762118458 Chr.4: 70993863-70993863 on Build GRCh38
rs907602129 Chr.4: 70994076-70994076 on Build GRCh38
rs776078836 Chr.4: 70994004-70994004 on Build GRCh38
rs368983488 Chr.4: 70993833-70993833 on Build GRCh38
rs933116528 Chr.4: 70994061-70994061 on Build GRCh38
rs1045316520 Chr.4: 70994185-70994185 on Build GRCh38
rs757622404 Chr.4: 70993793-70993793 on Build GRCh38
rs560647240 Chr.4: 70993758-70993758 on Build GRCh38
rs72552090 Chr.4: 70993995-70993995 on Build GRCh38
rs969014267 Chr.4: 70994148-70994148 on Build GRCh38
rs72552089 Chr.4: 70993784-70993784 on Build GRCh38
rs200834004 Chr.4: 70993807-70993807 on Build GRCh38
rs763783997 Chr.4: 70994018-70994018 on Build GRCh38
rs887633342 Chr.4: 70994033-70994033 on Build GRCh38
rs377182313 Chr.4: 70993907-70993907 on Build GRCh38
rs536825195 Chr.4: 70994065-70994065 on Build GRCh38
rs1025096771 Chr.4: 70994147-70994147 on Build GRCh38
rs767118073 Chr.4: 70993801-70993801 on Build GRCh38
rs759067543 Chr.4: 70994007-70994007 on Build GRCh38
rs72552093 Chr.4: 70994160-70994160 on Build GRCh38
rs769974178 Chr.4: 70994062-70994062 on Build GRCh38
rs765100675 Chr.4: 70994012-70994012 on Build GRCh38
rs772742361 Chr.4: 70993865-70993865 on Build GRCh38
rs974674289 Chr.4: 70994030-70994030 on Build GRCh38
rs1013501960 Chr.4: 70994139-70994139 on Build GRCh38
rs759403464 Chr.4: 70994000-70994001 on Build GRCh38
rs989856357 Chr.4: 70993999-70993999 on Build GRCh38
rs1037881777 Chr.4: 70994161-70994161 on Build GRCh38
rs201444059 Chr.4: 70993828-70993828 on Build GRCh38
rs964414556 Chr.4: 70994014-70994014 on Build GRCh38
rs761508555 Chr.4: 70993796-70993796 on Build GRCh38
rs749800459 Chr.4: 70993850-70993850 on Build GRCh38
rs754962228 Chr.4: 70993805-70993805 on Build GRCh38
rs989483827 Chr.4: 70993820-70993820 on Build GRCh38
rs568859559 Chr.4: 70993834-70993834 on Build GRCh38
rs200970510 Chr.4: 70993964-70993964 on Build GRCh38
rs572108629 Chr.4: 70993997-70993997 on Build GRCh38
rs142214180 Chr.4: 70993896-70993896 on Build GRCh38
rs963672787 Chr.4: 70993749-70993749 on Build GRCh38
rs540570195 Chr.4: 70994013-70994013 on Build GRCh38
rs760439706 Chr.4: 70993881-70993881 on Build GRCh38
rs1023824634 Chr.4: 70993771-70993771 on Build GRCh38
rs145221238 Chr.4: 70994176-70994176 on Build GRCh38
rs756463162 Chr.4: 70993943-70993943 on Build GRCh38
rs769248746 Chr.4: 70993958-70993958 on Build GRCh38
rs897697701 Chr.4: 70994115-70994115 on Build GRCh38
rs1013636917 Chr.4: 70993819-70993819 on Build GRCh38
rs1012483698 Chr.4: 70993929-70993929 on Build GRCh38
rs564125219 Chr.4: 70994017-70994017 on Build GRCh38
rs531912291 Chr.4: 70994075-70994075 on Build GRCh38
rs9997790 Chr.4: 70993963-70993963 on Build GRCh38
rs765857928 Chr.4: 70993889-70993889 on Build GRCh38
rs769340545 Chr.4: 70993853-70993853 on Build GRCh38
rs921884529 Chr.4: 70994059-70994059 on Build GRCh38
rs369545832 Chr.4: 70993977-70993977 on Build GRCh38
rs906785290 Chr.4: 70994189-70994189 on Build GRCh38
rs913448647 Chr.4: 70993955-70993955 on Build GRCh38
rs72553922 Chr.4: 70993774-70993774 on Build GRCh38
rs375049604 Chr.4: 70994098-70994098 on Build GRCh38
rs940465625 Chr.4: 70994089-70994089 on Build GRCh38
rs749423580 Chr.4: 70993954-70993954 on Build GRCh38
rs886191977 Chr.4: 70993876-70993876 on Build GRCh38
rs376018179 Chr.4: 70993823-70993823 on Build GRCh38
rs781411822 Chr.4: 70993922-70993922 on Build GRCh38
rs371808298 Chr.4: 70993794-70993794 on Build GRCh38
rs906693284 Chr.4: 70993972-70993972 on Build GRCh38
rs1012800657 Chr.4: 70993752-70993752 on Build GRCh38
rs72552087 Chr.4: 70993803-70993803 on Build GRCh38
rs538122704 Chr.4: 70993926-70993926 on Build GRCh38
rs1022313236 Chr.4: 70994006-70994006 on Build GRCh38
rs368316219 Chr.4: 70993788-70993788 on Build GRCh38
rs773680922 Chr.4: 70993978-70993978 on Build GRCh38
rs138159945 Chr.4: 70993859-70993859 on Build GRCh38
rs1006034549 Chr.4: 70994066-70994066 on Build GRCh38
rs72552098 Chr.4: 70993956-70993956 on Build GRCh38
rs368554642 Chr.4: 70993804-70993804 on Build GRCh38
rs66878317 Chr.4: 70993905-70993905 on Build GRCh38
rs200193775 Chr.4: 70993824-70993824 on Build GRCh38
rs545760162 Chr.4: 70994010-70994010 on Build GRCh38
rs372945476 Chr.4: 70993825-70993825 on Build GRCh38
rs779457928 Chr.4: 70993839-70993846 on Build GRCh38
rs868553565 Chr.4: 70993925-70993925 on Build GRCh38
rs111402794 Chr.4: 70994180-70994180 on Build GRCh38
rs780262850 Chr.4: 70993947-70993947 on Build GRCh38
rs72553921 Chr.4: 70993757-70993757 on Build GRCh38
rs199540553 Chr.4: 70993848-70993848 on Build GRCh38
rs767152030 Chr.4: 70993864-70993864 on Build GRCh38

View Full Product Details