Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 71023472-71023985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3737532 c.489T>A p.N163K Chr.4: 71023646-71023646 on Build GRCh38
4934971 c.504A>G p.Q168Q Chr.4: 71023661-71023661 on Build GRCh38
1284426 c.535C>A p.Q179K Chr.4: 71023692-71023692 on Build GRCh38
6573977 c.549+2T>G p.? Chr.4: 71023708-71023708 on Build GRCh38
6293691 c.530A>T p.Y177F Chr.4: 71023687-71023687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576990251 Chr.4: 71023652-71023652 on Build GRCh38
rs777492188 Chr.4: 71023617-71023617 on Build GRCh38
rs941186434 Chr.4: 71023867-71023868 on Build GRCh38
rs779186479 Chr.4: 71023732-71023732 on Build GRCh38
rs759057659 Chr.4: 71023608-71023628 on Build GRCh38
rs761816762 Chr.4: 71023678-71023678 on Build GRCh38
rs889655629 Chr.4: 71023861-71023861 on Build GRCh38
rs1049826986 Chr.4: 71023850-71023850 on Build GRCh38
rs761906822 Chr.4: 71023535-71023535 on Build GRCh38
rs780397016 Chr.4: 71023832-71023832 on Build GRCh38
rs137995308 Chr.4: 71023953-71023953 on Build GRCh38
rs767126142 Chr.4: 71023677-71023678 on Build GRCh38
rs761210008 Chr.4: 71023687-71023687 on Build GRCh38
rs139754308 Chr.4: 71023570-71023570 on Build GRCh38
rs201460017 Chr.4: 71023633-71023633 on Build GRCh38
rs777002353 Chr.4: 71023691-71023691 on Build GRCh38
rs528401408 Chr.4: 71023857-71023857 on Build GRCh38
rs758181636 Chr.4: 71023699-71023699 on Build GRCh38
rs767234705 Chr.4: 71023586-71023586 on Build GRCh38
rs970493696 Chr.4: 71023771-71023771 on Build GRCh38
rs781015507 Chr.4: 71023732-71023732 on Build GRCh38
rs12503595 Chr.4: 71023829-71023829 on Build GRCh38
rs185092624 Chr.4: 71023718-71023718 on Build GRCh38
rs942994565 Chr.4: 71023922-71023922 on Build GRCh38
rs146937779 Chr.4: 71023935-71023935 on Build GRCh38
rs751071896 Chr.4: 71023723-71023723 on Build GRCh38
rs772138151 Chr.4: 71023683-71023683 on Build GRCh38
rs375222793 Chr.4: 71023592-71023592 on Build GRCh38
rs1030647628 Chr.4: 71023805-71023805 on Build GRCh38
rs372497422 Chr.4: 71023580-71023580 on Build GRCh38
rs72553931 Chr.4: 71023670-71023670 on Build GRCh38
rs745621246 Chr.4: 71023743-71023743 on Build GRCh38
rs1016162315 Chr.4: 71023717-71023717 on Build GRCh38
rs752391013 Chr.4: 71023693-71023693 on Build GRCh38
rs745683761 Chr.4: 71023657-71023657 on Build GRCh38
rs199911146 Chr.4: 71023692-71023692 on Build GRCh38
rs956548837 Chr.4: 71023809-71023809 on Build GRCh38
rs982296813 Chr.4: 71023785-71023785 on Build GRCh38
rs760481266 Chr.4: 71023833-71023833 on Build GRCh38
rs762926099 Chr.4: 71023513-71023513 on Build GRCh38
rs750945857 Chr.4: 71023747-71023747 on Build GRCh38
rs575244378 Chr.4: 71023746-71023746 on Build GRCh38
rs1047568520 Chr.4: 71023955-71023955 on Build GRCh38
rs766821671 Chr.4: 71023689-71023689 on Build GRCh38
rs141119958 Chr.4: 71023913-71023913 on Build GRCh38
rs1038280950 Chr.4: 71023924-71023924 on Build GRCh38
rs766010862 Chr.4: 71023513-71023514 on Build GRCh38
rs752549246 Chr.4: 71023758-71023758 on Build GRCh38
rs776346903 Chr.4: 71023509-71023509 on Build GRCh38
rs768818120 Chr.4: 71023668-71023668 on Build GRCh38
rs774376139 Chr.4: 71023531-71023531 on Build GRCh38
rs3833976 Chr.4: 71023514-71023514 on Build GRCh38
rs975544844 Chr.4: 71023931-71023931 on Build GRCh38
rs750020287 Chr.4: 71023526-71023526 on Build GRCh38
rs757178515 Chr.4: 71023725-71023725 on Build GRCh38
rs757985137 Chr.4: 71023616-71023616 on Build GRCh38
rs774572004 Chr.4: 71023669-71023669 on Build GRCh38
rs190573557 Chr.4: 71023848-71023848 on Build GRCh38
rs763212879 Chr.4: 71023516-71023516 on Build GRCh38
rs752194823 Chr.4: 71023594-71023594 on Build GRCh38
rs915459386 Chr.4: 71023903-71023903 on Build GRCh38
rs757688428 Chr.4: 71023713-71023713 on Build GRCh38
rs750948462 Chr.4: 71023550-71023550 on Build GRCh38
rs1047045779 Chr.4: 71023663-71023663 on Build GRCh38
rs181745362 Chr.4: 71023707-71023707 on Build GRCh38
rs766741021 Chr.4: 71023565-71023565 on Build GRCh38
rs752383778 Chr.4: 71023692-71023693 on Build GRCh38
rs1023509647 Chr.4: 71023739-71023739 on Build GRCh38
rs978063021 Chr.4: 71023783-71023783 on Build GRCh38
rs558748148 Chr.4: 71023637-71023637 on Build GRCh38
rs767957001 Chr.4: 71023548-71023548 on Build GRCh38
rs546891172 Chr.4: 71023905-71023905 on Build GRCh38
rs542646281 Chr.4: 71023800-71023800 on Build GRCh38

View Full Product Details