Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87650177-87650677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
UniGene
Hs.652366
Gene Chromosome Location
Chr.4: 87650302-87664361 on Build GRCh38
Transcripts
NM_001079911.2 NM_004407.3

SNP Detail

SNP ID Chromosome Location
rs886450022 Chr.4: 87650276-87650276 on Build GRCh38
rs1004738729 Chr.4: 87650293-87650293 on Build GRCh38
rs985812828 Chr.4: 87650455-87650455 on Build GRCh38
rs913669349 Chr.4: 87650479-87650479 on Build GRCh38
rs1047697901 Chr.4: 87650216-87650216 on Build GRCh38
rs866307066 Chr.4: 87650207-87650207 on Build GRCh38
rs973869291 Chr.4: 87650635-87650635 on Build GRCh38
rs1002195277 Chr.4: 87650277-87650277 on Build GRCh38
rs955245145 Chr.4: 87650539-87650539 on Build GRCh38
rs929878919 Chr.4: 87650210-87650210 on Build GRCh38
rs1007962082 Chr.4: 87650574-87650574 on Build GRCh38
rs531125325 Chr.4: 87650304-87650304 on Build GRCh38
rs535082691 Chr.4: 87650592-87650592 on Build GRCh38
rs552537339 Chr.4: 87650306-87650306 on Build GRCh38
rs906028032 Chr.4: 87650392-87650392 on Build GRCh38
rs1019921107 Chr.4: 87650501-87650501 on Build GRCh38
rs546700305 Chr.4: 87650483-87650483 on Build GRCh38
rs1033147174 Chr.4: 87650482-87650482 on Build GRCh38
rs570513659 Chr.4: 87650402-87650402 on Build GRCh38
rs371000476 Chr.4: 87650244-87650244 on Build GRCh38
rs1028566860 Chr.4: 87650419-87650419 on Build GRCh38
rs138366492 Chr.4: 87650496-87650496 on Build GRCh38
rs965751104 Chr.4: 87650460-87650460 on Build GRCh38
rs36089093 Chr.4: 87650293-87650293 on Build GRCh38
rs950148567 Chr.4: 87650285-87650285 on Build GRCh38
rs1015686758 Chr.4: 87650321-87650321 on Build GRCh38
rs951247141 Chr.4: 87650652-87650652 on Build GRCh38
rs926521420 Chr.4: 87650629-87650629 on Build GRCh38
rs919774419 Chr.4: 87650650-87650650 on Build GRCh38
rs954302450 Chr.4: 87650444-87650444 on Build GRCh38
rs754509857 Chr.4: 87650473-87650473 on Build GRCh38
rs1046258277 Chr.4: 87650305-87650305 on Build GRCh38
rs1018482703 Chr.4: 87650576-87650576 on Build GRCh38
rs911584026 Chr.4: 87650459-87650459 on Build GRCh38
rs753416234 Chr.4: 87650310-87650310 on Build GRCh38
rs534448935 Chr.4: 87650418-87650418 on Build GRCh38

View Full Product Details