Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87656438-87656972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
UniGene
Hs.652366
Gene Chromosome Location
Chr.4: 87650302-87664361 on Build GRCh38
Transcripts
NM_001079911.2 NM_004407.3 XM_011531705.2 XM_011531706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619200 c.20T>A p.L7H Chr.4: 87656512-87656512 on Build GRCh38
4412302 c.54+4A>C p.? Chr.4: 87656550-87656550 on Build GRCh38
5605677 c.21C>T p.L7L Chr.4: 87656513-87656513 on Build GRCh38
348065 c.15C>A p.I5I Chr.4: 87656507-87656507 on Build GRCh38
3606853 c.34G>A p.G12R Chr.4: 87656526-87656526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925803091 Chr.4: 87656655-87656655 on Build GRCh38
rs920371451 Chr.4: 87656675-87656675 on Build GRCh38
rs868149050 Chr.4: 87656530-87656530 on Build GRCh38
rs879096748 Chr.4: 87656706-87656706 on Build GRCh38
rs765601823 Chr.4: 87656487-87656487 on Build GRCh38
rs374190026 Chr.4: 87656609-87656609 on Build GRCh38
rs373552880 Chr.4: 87656584-87656584 on Build GRCh38
rs745396074 Chr.4: 87656532-87656532 on Build GRCh38
rs773787734 Chr.4: 87656791-87656791 on Build GRCh38
rs376396990 Chr.4: 87656569-87656569 on Build GRCh38
rs765915591 Chr.4: 87656818-87656818 on Build GRCh38
rs566234658 Chr.4: 87656670-87656670 on Build GRCh38
rs950968947 Chr.4: 87656498-87656498 on Build GRCh38
rs1035222851 Chr.4: 87656893-87656893 on Build GRCh38
rs755630320 Chr.4: 87656537-87656537 on Build GRCh38
rs779724160 Chr.4: 87656543-87656543 on Build GRCh38
rs766307823 Chr.4: 87656571-87656572 on Build GRCh38
rs558296261 Chr.4: 87656943-87656943 on Build GRCh38
rs777171983 Chr.4: 87656586-87656586 on Build GRCh38
rs933909381 Chr.4: 87656732-87656732 on Build GRCh38
rs369626347 Chr.4: 87656564-87656564 on Build GRCh38
rs905298880 Chr.4: 87656849-87656849 on Build GRCh38
rs768312745 Chr.4: 87656661-87656661 on Build GRCh38
rs961740147 Chr.4: 87656637-87656637 on Build GRCh38
rs140768637 Chr.4: 87656504-87656504 on Build GRCh38
rs104893834 Chr.4: 87656493-87656493 on Build GRCh38
rs748892144 Chr.4: 87656657-87656657 on Build GRCh38
rs1051742044 Chr.4: 87656733-87656733 on Build GRCh38
rs559222197 Chr.4: 87656712-87656712 on Build GRCh38
rs79180984 Chr.4: 87656570-87656570 on Build GRCh38
rs751314577 Chr.4: 87656514-87656514 on Build GRCh38
rs372799088 Chr.4: 87656566-87656566 on Build GRCh38
rs4693851 Chr.4: 87656602-87656602 on Build GRCh38
rs1015872608 Chr.4: 87656940-87656940 on Build GRCh38
rs946749230 Chr.4: 87656810-87656810 on Build GRCh38
rs376221415 Chr.4: 87656519-87656519 on Build GRCh38
rs373562639 Chr.4: 87656503-87656503 on Build GRCh38
rs554700691 Chr.4: 87656846-87656846 on Build GRCh38
rs1027020354 Chr.4: 87656496-87656496 on Build GRCh38
rs186164610 Chr.4: 87656605-87656605 on Build GRCh38
rs931795480 Chr.4: 87656691-87656691 on Build GRCh38
rs188758040 Chr.4: 87656911-87656911 on Build GRCh38
rs537199422 Chr.4: 87656933-87656933 on Build GRCh38
rs531731094 Chr.4: 87656578-87656578 on Build GRCh38
rs748935080 Chr.4: 87656565-87656565 on Build GRCh38
rs957472881 Chr.4: 87656690-87656690 on Build GRCh38
rs943420977 Chr.4: 87656778-87656778 on Build GRCh38
rs1002371584 Chr.4: 87656871-87656871 on Build GRCh38
rs969930433 Chr.4: 87656572-87656572 on Build GRCh38
rs575811118 Chr.4: 87656468-87656468 on Build GRCh38
rs892435284 Chr.4: 87656789-87656789 on Build GRCh38
rs896666558 Chr.4: 87656898-87656899 on Build GRCh38
rs528157814 Chr.4: 87656836-87656836 on Build GRCh38
rs879272596 Chr.4: 87656708-87656708 on Build GRCh38
rs747655605 Chr.4: 87656796-87656796 on Build GRCh38
rs773010324 Chr.4: 87656464-87656465 on Build GRCh38
rs763964054 Chr.4: 87656512-87656512 on Build GRCh38
rs899327011 Chr.4: 87656817-87656817 on Build GRCh38
rs747358988 Chr.4: 87656579-87656579 on Build GRCh38
rs762846022 Chr.4: 87656475-87656476 on Build GRCh38
rs907506694 Chr.4: 87656624-87656624 on Build GRCh38
rs536840167 Chr.4: 87656769-87656769 on Build GRCh38

View Full Product Details