Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87656947-87657453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
UniGene
Hs.652366
Gene Chromosome Location
Chr.4: 87650302-87664361 on Build GRCh38
Transcripts
NM_001079911.2 NM_004407.3 XM_011531705.2 XM_011531706.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887904 c.81T>G p.S27S Chr.4: 87657058-87657058 on Build GRCh38
2957176 c.91G>A p.E31K Chr.4: 87657068-87657068 on Build GRCh38
328149 c.55-3T>G p.? Chr.4: 87657029-87657029 on Build GRCh38
6645638 c.102G>A p.K34K Chr.4: 87657079-87657079 on Build GRCh38
735392 c.85G>T p.D29Y Chr.4: 87657062-87657062 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926608612 Chr.4: 87657417-87657417 on Build GRCh38
rs769026068 Chr.4: 87657115-87657115 on Build GRCh38
rs866248509 Chr.4: 87657346-87657346 on Build GRCh38
rs985820121 Chr.4: 87657324-87657324 on Build GRCh38
rs758529495 Chr.4: 87657419-87657419 on Build GRCh38
rs765550566 Chr.4: 87657295-87657295 on Build GRCh38
rs754656539 Chr.4: 87657060-87657060 on Build GRCh38
rs770363297 Chr.4: 87657112-87657112 on Build GRCh38
rs775720445 Chr.4: 87657113-87657113 on Build GRCh38
rs376796407 Chr.4: 87657009-87657009 on Build GRCh38
rs755687466 Chr.4: 87657050-87657050 on Build GRCh38
rs771292808 Chr.4: 87657082-87657082 on Build GRCh38
rs150105108 Chr.4: 87657056-87657056 on Build GRCh38
rs965836247 Chr.4: 87657349-87657349 on Build GRCh38
rs2615479 Chr.4: 87657158-87657158 on Build GRCh38
rs898773411 Chr.4: 87657226-87657226 on Build GRCh38
rs774613442 Chr.4: 87657116-87657116 on Build GRCh38
rs1056566231 Chr.4: 87657148-87657148 on Build GRCh38
rs751312821 Chr.4: 87657423-87657426 on Build GRCh38
rs116792016 Chr.4: 87657292-87657292 on Build GRCh38
rs769988025 Chr.4: 87656991-87656991 on Build GRCh38
rs750344899 Chr.4: 87657048-87657048 on Build GRCh38
rs938045650 Chr.4: 87657147-87657147 on Build GRCh38
rs930755192 Chr.4: 87657221-87657221 on Build GRCh38
rs202210004 Chr.4: 87657068-87657068 on Build GRCh38
rs766601503 Chr.4: 87657075-87657075 on Build GRCh38
rs946691714 Chr.4: 87657400-87657400 on Build GRCh38
rs200752295 Chr.4: 87657007-87657007 on Build GRCh38
rs552859119 Chr.4: 87657039-87657039 on Build GRCh38
rs374841779 Chr.4: 87657379-87657379 on Build GRCh38
rs1001574565 Chr.4: 87657404-87657404 on Build GRCh38
rs773797362 Chr.4: 87656984-87656984 on Build GRCh38
rs368215646 Chr.4: 87657407-87657407 on Build GRCh38
rs550214616 Chr.4: 87657400-87657405 on Build GRCh38
rs920640210 Chr.4: 87657189-87657189 on Build GRCh38
rs367681762 Chr.4: 87657092-87657092 on Build GRCh38
rs367899184 Chr.4: 87657281-87657281 on Build GRCh38
rs181490843 Chr.4: 87657029-87657029 on Build GRCh38
rs11935907 Chr.4: 87657291-87657291 on Build GRCh38
rs141269375 Chr.4: 87657185-87657185 on Build GRCh38
rs746369070 Chr.4: 87657096-87657096 on Build GRCh38
rs763278171 Chr.4: 87657017-87657017 on Build GRCh38
rs979767466 Chr.4: 87657414-87657414 on Build GRCh38
rs1004397795 Chr.4: 87657171-87657171 on Build GRCh38
rs138537025 Chr.4: 87657057-87657057 on Build GRCh38
rs761623032 Chr.4: 87657036-87657036 on Build GRCh38
rs1545735 Chr.4: 87657335-87657335 on Build GRCh38
rs562686862 Chr.4: 87657019-87657019 on Build GRCh38
rs1047859899 Chr.4: 87657228-87657228 on Build GRCh38
rs767405803 Chr.4: 87657047-87657047 on Build GRCh38
rs376999442 Chr.4: 87656981-87656981 on Build GRCh38
rs953164688 Chr.4: 87657322-87657322 on Build GRCh38
rs761554083 Chr.4: 87657121-87657121 on Build GRCh38
rs950297209 Chr.4: 87657306-87657306 on Build GRCh38
rs943402772 Chr.4: 87657159-87657159 on Build GRCh38
rs186024777 Chr.4: 87657114-87657114 on Build GRCh38
rs369119886 Chr.4: 87656994-87656994 on Build GRCh38
rs587776697 Chr.4: 87657031-87657031 on Build GRCh38

View Full Product Details