Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88738950-88739453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6453096 c.2510G>A p.R837K Chr.4: 88739082-88739082 on Build GRCh38
304727 c.2522T>G p.V841G Chr.4: 88739070-88739070 on Build GRCh38
1661745 c.1496A>G p.K499R Chr.4: 88739118-88739118 on Build GRCh38
6101179 c.2517G>A p.R839R Chr.4: 88739075-88739075 on Build GRCh38
6658574 c.1504C>T p.R502W Chr.4: 88739110-88739110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942745659 Chr.4: 88739268-88739268 on Build GRCh38
rs759024023 Chr.4: 88739031-88739031 on Build GRCh38
rs775435079 Chr.4: 88738998-88738998 on Build GRCh38
rs200147664 Chr.4: 88739162-88739162 on Build GRCh38
rs200812331 Chr.4: 88739070-88739070 on Build GRCh38
rs2290356 Chr.4: 88739011-88739011 on Build GRCh38
rs770042385 Chr.4: 88739109-88739109 on Build GRCh38
rs760372862 Chr.4: 88739144-88739144 on Build GRCh38
rs201522307 Chr.4: 88739103-88739103 on Build GRCh38
rs189951620 Chr.4: 88739193-88739193 on Build GRCh38
rs957774979 Chr.4: 88739186-88739186 on Build GRCh38
rs778203509 Chr.4: 88739093-88739093 on Build GRCh38
rs918551257 Chr.4: 88739251-88739251 on Build GRCh38
rs756878381 Chr.4: 88739160-88739160 on Build GRCh38
rs963088606 Chr.4: 88739249-88739249 on Build GRCh38
rs761523453 Chr.4: 88739065-88739065 on Build GRCh38
rs1056398178 Chr.4: 88739078-88739078 on Build GRCh38
rs895101380 Chr.4: 88739086-88739086 on Build GRCh38
rs747342480 Chr.4: 88739099-88739099 on Build GRCh38
rs753387663 Chr.4: 88739146-88739146 on Build GRCh38
rs1052135216 Chr.4: 88739415-88739415 on Build GRCh38
rs948019395 Chr.4: 88739317-88739317 on Build GRCh38
rs766201180 Chr.4: 88739145-88739145 on Build GRCh38
rs151124976 Chr.4: 88739357-88739357 on Build GRCh38
rs773112301 Chr.4: 88739143-88739143 on Build GRCh38
rs771172333 Chr.4: 88739052-88739052 on Build GRCh38
rs778251680 Chr.4: 88739032-88739032 on Build GRCh38
rs181335730 Chr.4: 88739329-88739329 on Build GRCh38
rs759594197 Chr.4: 88738989-88738989 on Build GRCh38
rs902425053 Chr.4: 88739407-88739409 on Build GRCh38
rs369806368 Chr.4: 88739167-88739167 on Build GRCh38
rs939330011 Chr.4: 88739259-88739259 on Build GRCh38
rs757831044 Chr.4: 88739165-88739165 on Build GRCh38
rs901522628 Chr.4: 88739155-88739155 on Build GRCh38
rs138276268 Chr.4: 88739054-88739054 on Build GRCh38
rs376269141 Chr.4: 88739114-88739114 on Build GRCh38
rs750077363 Chr.4: 88739069-88739069 on Build GRCh38
rs755586625 Chr.4: 88739018-88739018 on Build GRCh38
rs935038919 Chr.4: 88739391-88739391 on Build GRCh38
rs775539140 Chr.4: 88739113-88739113 on Build GRCh38
rs376246251 Chr.4: 88738986-88738986 on Build GRCh38
rs775594545 Chr.4: 88739060-88739060 on Build GRCh38
rs949097601 Chr.4: 88739246-88739246 on Build GRCh38
rs767133757 Chr.4: 88739068-88739068 on Build GRCh38
rs573963851 Chr.4: 88739179-88739179 on Build GRCh38
rs115629193 Chr.4: 88739087-88739087 on Build GRCh38
rs762686972 Chr.4: 88739002-88739002 on Build GRCh38
rs200222917 Chr.4: 88739110-88739110 on Build GRCh38
rs373060384 Chr.4: 88739049-88739049 on Build GRCh38
rs779529385 Chr.4: 88739368-88739368 on Build GRCh38
rs954348459 Chr.4: 88739216-88739216 on Build GRCh38
rs771789121 Chr.4: 88739141-88739141 on Build GRCh38
rs757593108 Chr.4: 88739101-88739101 on Build GRCh38
rs755673064 Chr.4: 88739273-88739273 on Build GRCh38
rs753439131 Chr.4: 88739077-88739077 on Build GRCh38
rs916485896 Chr.4: 88739315-88739315 on Build GRCh38
rs767120551 Chr.4: 88739234-88739234 on Build GRCh38
rs771098241 Chr.4: 88738981-88738981 on Build GRCh38
rs762842310 Chr.4: 88739061-88739061 on Build GRCh38
rs1007015089 Chr.4: 88739100-88739100 on Build GRCh38
rs371929675 Chr.4: 88738988-88738988 on Build GRCh38
rs765090739 Chr.4: 88738995-88738995 on Build GRCh38
rs779526161 Chr.4: 88739022-88739022 on Build GRCh38
rs763850726 Chr.4: 88739003-88739003 on Build GRCh38
rs370289351 Chr.4: 88739006-88739006 on Build GRCh38
rs776720799 Chr.4: 88739053-88739053 on Build GRCh38
rs34309843 Chr.4: 88739116-88739117 on Build GRCh38
rs748690836 Chr.4: 88739023-88739023 on Build GRCh38
rs916228320 Chr.4: 88739226-88739226 on Build GRCh38
rs764620574 Chr.4: 88739154-88739154 on Build GRCh38
rs747402226 Chr.4: 88739047-88739047 on Build GRCh38
rs774163076 Chr.4: 88739133-88739133 on Build GRCh38
rs144309025 Chr.4: 88739121-88739121 on Build GRCh38
rs939216863 Chr.4: 88739059-88739059 on Build GRCh38
rs116320485 Chr.4: 88739010-88739010 on Build GRCh38
rs759132305 Chr.4: 88739152-88739152 on Build GRCh38
rs114111202 Chr.4: 88739248-88739248 on Build GRCh38
rs760301767 Chr.4: 88739140-88739141 on Build GRCh38
rs781378526 Chr.4: 88739171-88739171 on Build GRCh38
rs149846247 Chr.4: 88739263-88739264 on Build GRCh38
rs569685131 Chr.4: 88739191-88739191 on Build GRCh38
rs1030624259 Chr.4: 88739386-88739387 on Build GRCh38
rs972848725 Chr.4: 88739250-88739250 on Build GRCh38
rs774180726 Chr.4: 88739063-88739063 on Build GRCh38
rs992191662 Chr.4: 88739275-88739275 on Build GRCh38
rs184743658 Chr.4: 88739017-88739017 on Build GRCh38

View Full Product Details