Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 93829170-93829670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH1
Entrez Gene ID
474
Gene Name
atonal bHLH transcription factor 1
Gene Aliases
ATH1, HATH1, MATH-1, bHLHa14
UniGene
Hs.532680
Gene Chromosome Location
Chr.4: 93828927-93829991 on Build GRCh38
Transcripts
NM_005172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137409 c.500G>A p.R167K Chr.4: 93829426-93829426 on Build GRCh38
7356877 c.321G>C p.R107S Chr.4: 93829247-93829247 on Build GRCh38
1059020 c.599C>T p.T200I Chr.4: 93829525-93829525 on Build GRCh38
5401838 c.339C>T p.A113A Chr.4: 93829265-93829265 on Build GRCh38
7267738 c.652C>G p.P218A Chr.4: 93829578-93829578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774159886 Chr.4: 93829203-93829203 on Build GRCh38
rs942784851 Chr.4: 93829457-93829457 on Build GRCh38
rs369636991 Chr.4: 93829334-93829334 on Build GRCh38
rs769357921 Chr.4: 93829341-93829341 on Build GRCh38
rs760041029 Chr.4: 93829211-93829211 on Build GRCh38
rs200577041 Chr.4: 93829209-93829209 on Build GRCh38
rs780656881 Chr.4: 93829265-93829267 on Build GRCh38
rs757248475 Chr.4: 93829228-93829228 on Build GRCh38
rs753993575 Chr.4: 93829392-93829392 on Build GRCh38
rs942857356 Chr.4: 93829217-93829217 on Build GRCh38
rs533752955 Chr.4: 93829604-93829604 on Build GRCh38
rs762223368 Chr.4: 93829459-93829459 on Build GRCh38
rs771487306 Chr.4: 93829652-93829652 on Build GRCh38
rs772754916 Chr.4: 93829346-93829346 on Build GRCh38
rs930220938 Chr.4: 93829527-93829527 on Build GRCh38
rs796357340 Chr.4: 93829366-93829366 on Build GRCh38
rs773671771 Chr.4: 93829589-93829589 on Build GRCh38
rs969557427 Chr.4: 93829357-93829357 on Build GRCh38
rs751381751 Chr.4: 93829553-93829553 on Build GRCh38
rs781377479 Chr.4: 93829229-93829229 on Build GRCh38
rs761125136 Chr.4: 93829598-93829598 on Build GRCh38
rs758188041 Chr.4: 93829432-93829432 on Build GRCh38
rs764478508 Chr.4: 93829297-93829297 on Build GRCh38
rs757448327 Chr.4: 93829299-93829299 on Build GRCh38
rs773870667 Chr.4: 93829262-93829262 on Build GRCh38
rs756274872 Chr.4: 93829331-93829331 on Build GRCh38
rs369046364 Chr.4: 93829388-93829388 on Build GRCh38
rs537422021 Chr.4: 93829368-93829368 on Build GRCh38
rs373671734 Chr.4: 93829635-93829635 on Build GRCh38
rs766737827 Chr.4: 93829485-93829485 on Build GRCh38
rs745424790 Chr.4: 93829218-93829218 on Build GRCh38
rs765269254 Chr.4: 93829601-93829601 on Build GRCh38
rs763307747 Chr.4: 93829296-93829296 on Build GRCh38
rs780811958 Chr.4: 93829627-93829627 on Build GRCh38
rs746512569 Chr.4: 93829354-93829354 on Build GRCh38
rs747363887 Chr.4: 93829649-93829649 on Build GRCh38
rs770498732 Chr.4: 93829361-93829361 on Build GRCh38
rs780142317 Chr.4: 93829232-93829232 on Build GRCh38
rs756638857 Chr.4: 93829450-93829450 on Build GRCh38
rs746708814 Chr.4: 93829441-93829441 on Build GRCh38
rs767718503 Chr.4: 93829389-93829389 on Build GRCh38
rs750143677 Chr.4: 93829631-93829631 on Build GRCh38
rs748860929 Chr.4: 93829636-93829636 on Build GRCh38
rs373848308 Chr.4: 93829568-93829568 on Build GRCh38
rs745410157 Chr.4: 93829445-93829445 on Build GRCh38
rs369884611 Chr.4: 93829561-93829561 on Build GRCh38
rs35182771 Chr.4: 93829637-93829637 on Build GRCh38
rs771217805 Chr.4: 93829599-93829599 on Build GRCh38
rs780830478 Chr.4: 93829555-93829555 on Build GRCh38
rs769137416 Chr.4: 93829447-93829447 on Build GRCh38
rs867898378 Chr.4: 93829380-93829380 on Build GRCh38
rs769812244 Chr.4: 93829231-93829231 on Build GRCh38
rs767094967 Chr.4: 93829208-93829208 on Build GRCh38
rs778722533 Chr.4: 93829411-93829411 on Build GRCh38
rs772384185 Chr.4: 93829467-93829467 on Build GRCh38
rs139664764 Chr.4: 93829585-93829585 on Build GRCh38
rs376812790 Chr.4: 93829364-93829364 on Build GRCh38
rs771645916 Chr.4: 93829276-93829276 on Build GRCh38
rs997548725 Chr.4: 93829233-93829233 on Build GRCh38
rs760109556 Chr.4: 93829281-93829281 on Build GRCh38
rs1052248211 Chr.4: 93829471-93829471 on Build GRCh38
rs371205873 Chr.4: 93829201-93829201 on Build GRCh38
rs754926613 Chr.4: 93829404-93829404 on Build GRCh38
rs748643346 Chr.4: 93829583-93829583 on Build GRCh38
rs143419348 Chr.4: 93829298-93829298 on Build GRCh38
rs774791660 Chr.4: 93829348-93829348 on Build GRCh38
rs142870406 Chr.4: 93829600-93829600 on Build GRCh38
rs763789767 Chr.4: 93829390-93829390 on Build GRCh38
rs149240097 Chr.4: 93829273-93829273 on Build GRCh38
rs370710738 Chr.4: 93829612-93829612 on Build GRCh38
rs777481278 Chr.4: 93829439-93829439 on Build GRCh38
rs768629371 Chr.4: 93829236-93829236 on Build GRCh38
rs531803049 Chr.4: 93829626-93829626 on Build GRCh38
rs761088395 Chr.4: 93829624-93829624 on Build GRCh38
rs373777032 Chr.4: 93829444-93829444 on Build GRCh38
rs774732105 Chr.4: 93829458-93829458 on Build GRCh38
rs115524348 Chr.4: 93829340-93829340 on Build GRCh38
rs764053065 Chr.4: 93829618-93829618 on Build GRCh38
rs202025292 Chr.4: 93829344-93829344 on Build GRCh38
rs750448934 Chr.4: 93829305-93829305 on Build GRCh38
rs368475681 Chr.4: 93829213-93829213 on Build GRCh38
rs888717321 Chr.4: 93829592-93829592 on Build GRCh38
rs547129931 Chr.4: 93829235-93829235 on Build GRCh38
rs377300000 Chr.4: 93829593-93829593 on Build GRCh38
rs752995405 Chr.4: 93829214-93829214 on Build GRCh38
rs751879356 Chr.4: 93829224-93829224 on Build GRCh38
rs1051774230 Chr.4: 93829596-93829596 on Build GRCh38
rs201509026 Chr.4: 93829220-93829220 on Build GRCh38
rs773761668 Chr.4: 93829517-93829517 on Build GRCh38
rs776038071 Chr.4: 93829289-93829289 on Build GRCh38
rs759699829 Chr.4: 93829500-93829500 on Build GRCh38
rs34543561 Chr.4: 93829499-93829499 on Build GRCh38
rs1045137930 Chr.4: 93829605-93829605 on Build GRCh38
rs749304452 Chr.4: 93829336-93829336 on Build GRCh38
rs947764486 Chr.4: 93829190-93829190 on Build GRCh38
rs142207012 Chr.4: 93829391-93829391 on Build GRCh38
rs201671383 Chr.4: 93829573-93829573 on Build GRCh38
rs760930701 Chr.4: 93829484-93829484 on Build GRCh38
rs765205948 Chr.4: 93829508-93829508 on Build GRCh38
rs767661587 Chr.4: 93829300-93829300 on Build GRCh38
rs773536945 Chr.4: 93829469-93829469 on Build GRCh38
rs779471383 Chr.4: 93829582-93829582 on Build GRCh38
rs1030719508 Chr.4: 93829269-93829269 on Build GRCh38
rs34477680 Chr.4: 93829373-93829374 on Build GRCh38
rs756866699 Chr.4: 93829554-93829554 on Build GRCh38
rs751291709 Chr.4: 93829623-93829623 on Build GRCh38
rs777474577 Chr.4: 93829539-93829539 on Build GRCh38
rs761216588 Chr.4: 93829207-93829207 on Build GRCh38
rs772739464 Chr.4: 93829277-93829277 on Build GRCh38
rs376429461 Chr.4: 93829230-93829230 on Build GRCh38
rs758178289 Chr.4: 93829538-93829538 on Build GRCh38
rs200824879 Chr.4: 93829430-93829430 on Build GRCh38
rs548738966 Chr.4: 93829362-93829362 on Build GRCh38
rs755620274 Chr.4: 93829632-93829632 on Build GRCh38

View Full Product Details