Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 93829568-93830055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATOH1
Entrez Gene ID
474
Gene Name
atonal bHLH transcription factor 1
Gene Aliases
ATH1, HATH1, MATH-1, bHLHa14
UniGene
Hs.532680
Gene Chromosome Location
Chr.4: 93828927-93829991 on Build GRCh38
Transcripts
NM_005172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5737033 c.784G>T p.G262W Chr.4: 93829710-93829710 on Build GRCh38
3768058 c.973G>T p.E325* Chr.4: 93829899-93829899 on Build GRCh38
5491825 c.1013A>G p.D338G Chr.4: 93829939-93829939 on Build GRCh38
1059022 c.1018G>A p.E340K Chr.4: 93829944-93829944 on Build GRCh38
7324407 c.900C>T p.S300S Chr.4: 93829826-93829826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770072676 Chr.4: 93829665-93829665 on Build GRCh38
rs533752955 Chr.4: 93829604-93829604 on Build GRCh38
rs771487306 Chr.4: 93829652-93829652 on Build GRCh38
rs770793566 Chr.4: 93829948-93829948 on Build GRCh38
rs543328241 Chr.4: 93829903-93829903 on Build GRCh38
rs892241315 Chr.4: 93829923-93829923 on Build GRCh38
rs773671771 Chr.4: 93829589-93829589 on Build GRCh38
rs776122789 Chr.4: 93829850-93829850 on Build GRCh38
rs756563436 Chr.4: 93829810-93829810 on Build GRCh38
rs753590807 Chr.4: 93829777-93829777 on Build GRCh38
rs761125136 Chr.4: 93829598-93829598 on Build GRCh38
rs200054812 Chr.4: 93829891-93829891 on Build GRCh38
rs960325653 Chr.4: 93829676-93829676 on Build GRCh38
rs374364595 Chr.4: 93829864-93829866 on Build GRCh38
rs751008859 Chr.4: 93829889-93829889 on Build GRCh38
rs35227734 Chr.4: 93829734-93829734 on Build GRCh38
rs373671734 Chr.4: 93829635-93829635 on Build GRCh38
rs754504277 Chr.4: 93829792-93829792 on Build GRCh38
rs145270864 Chr.4: 93829743-93829743 on Build GRCh38
rs767465667 Chr.4: 93829764-93829764 on Build GRCh38
rs201443762 Chr.4: 93829843-93829843 on Build GRCh38
rs765269254 Chr.4: 93829601-93829601 on Build GRCh38
rs780811958 Chr.4: 93829627-93829627 on Build GRCh38
rs943398640 Chr.4: 93829842-93829842 on Build GRCh38
rs778916265 Chr.4: 93829829-93829829 on Build GRCh38
rs747363887 Chr.4: 93829649-93829649 on Build GRCh38
rs760381446 Chr.4: 93829840-93829840 on Build GRCh38
rs759405519 Chr.4: 93829965-93829965 on Build GRCh38
rs982925751 Chr.4: 93829813-93829813 on Build GRCh38
rs761788317 Chr.4: 93829758-93829758 on Build GRCh38
rs939103703 Chr.4: 93829892-93829892 on Build GRCh38
rs750143677 Chr.4: 93829631-93829631 on Build GRCh38
rs748860929 Chr.4: 93829636-93829636 on Build GRCh38
rs776442303 Chr.4: 93829952-93829952 on Build GRCh38
rs762550204 Chr.4: 93829995-93829995 on Build GRCh38
rs753900320 Chr.4: 93829723-93829723 on Build GRCh38
rs35182771 Chr.4: 93829637-93829637 on Build GRCh38
rs771217805 Chr.4: 93829599-93829599 on Build GRCh38
rs555824120 Chr.4: 93829717-93829717 on Build GRCh38
rs142131282 Chr.4: 93829825-93829825 on Build GRCh38
rs766059965 Chr.4: 93829719-93829719 on Build GRCh38
rs748213328 Chr.4: 93829912-93829912 on Build GRCh38
rs774333038 Chr.4: 93829683-93829683 on Build GRCh38
rs775797343 Chr.4: 93829736-93829736 on Build GRCh38
rs768701507 Chr.4: 93829827-93829827 on Build GRCh38
rs373713422 Chr.4: 93829735-93829735 on Build GRCh38
rs763702912 Chr.4: 93829996-93829996 on Build GRCh38
rs140137565 Chr.4: 93829932-93829932 on Build GRCh38
rs755154222 Chr.4: 93829898-93829898 on Build GRCh38
rs993647078 Chr.4: 93829657-93829657 on Build GRCh38
rs142870406 Chr.4: 93829600-93829600 on Build GRCh38
rs529589968 Chr.4: 93829900-93829900 on Build GRCh38
rs370710738 Chr.4: 93829612-93829612 on Build GRCh38
rs769516625 Chr.4: 93829967-93829967 on Build GRCh38
rs761593903 Chr.4: 93829696-93829696 on Build GRCh38
rs531803049 Chr.4: 93829626-93829626 on Build GRCh38
rs777540023 Chr.4: 93829924-93829924 on Build GRCh38
rs752679675 Chr.4: 93829772-93829772 on Build GRCh38
rs761088395 Chr.4: 93829624-93829624 on Build GRCh38
rs750068829 Chr.4: 93829715-93829715 on Build GRCh38
rs910104204 Chr.4: 93829821-93829821 on Build GRCh38
rs764053065 Chr.4: 93829618-93829618 on Build GRCh38
rs199789935 Chr.4: 93829749-93829749 on Build GRCh38
rs753229261 Chr.4: 93829720-93829720 on Build GRCh38
rs781726770 Chr.4: 93829798-93829798 on Build GRCh38
rs375846661 Chr.4: 93829661-93829661 on Build GRCh38
rs764885802 Chr.4: 93829794-93829794 on Build GRCh38
rs888717321 Chr.4: 93829592-93829592 on Build GRCh38
rs151169302 Chr.4: 93829919-93829919 on Build GRCh38
rs780137669 Chr.4: 93829817-93829817 on Build GRCh38
rs149770942 Chr.4: 93829682-93829682 on Build GRCh38
rs759213179 Chr.4: 93829856-93829856 on Build GRCh38
rs377300000 Chr.4: 93829593-93829593 on Build GRCh38
rs573894468 Chr.4: 93829664-93829664 on Build GRCh38
rs750699452 Chr.4: 93829804-93829804 on Build GRCh38
rs1037644837 Chr.4: 93829886-93829886 on Build GRCh38
rs1051774230 Chr.4: 93829596-93829596 on Build GRCh38
rs371160490 Chr.4: 93829895-93829895 on Build GRCh38
rs763102271 Chr.4: 93829681-93829681 on Build GRCh38
rs1045137930 Chr.4: 93829605-93829605 on Build GRCh38
rs754224658 Chr.4: 93829897-93829897 on Build GRCh38
rs558292896 Chr.4: 93829835-93829835 on Build GRCh38
rs1056642816 Chr.4: 93829894-93829894 on Build GRCh38
rs536528718 Chr.4: 93829670-93829670 on Build GRCh38
rs757670696 Chr.4: 93829731-93829731 on Build GRCh38
rs144338372 Chr.4: 93829901-93829901 on Build GRCh38
rs562808631 Chr.4: 93829982-93829982 on Build GRCh38
rs1026411537 Chr.4: 93829669-93829669 on Build GRCh38
rs370454058 Chr.4: 93829724-93829724 on Build GRCh38
rs747853211 Chr.4: 93829963-93829965 on Build GRCh38
rs773259389 Chr.4: 93829837-93829837 on Build GRCh38
rs200758862 Chr.4: 93829883-93829883 on Build GRCh38
rs145666261 Chr.4: 93829728-93829728 on Build GRCh38
rs765932446 Chr.4: 93829774-93829774 on Build GRCh38
rs772815944 Chr.4: 93829770-93829770 on Build GRCh38
rs371446004 Chr.4: 93829756-93829756 on Build GRCh38
rs781645737 Chr.4: 93829732-93829732 on Build GRCh38
rs767899405 Chr.4: 93829884-93829884 on Build GRCh38
rs368034043 Chr.4: 93829785-93829785 on Build GRCh38
rs913687437 Chr.4: 93829767-93829767 on Build GRCh38
rs767341255 Chr.4: 93829706-93829706 on Build GRCh38
rs749567383 Chr.4: 93829826-93829826 on Build GRCh38
rs540814845 Chr.4: 93829874-93829874 on Build GRCh38
rs147620158 Chr.4: 93829797-93829797 on Build GRCh38
rs751291709 Chr.4: 93829623-93829623 on Build GRCh38
rs748258469 Chr.4: 93829830-93829830 on Build GRCh38
rs1009371963 Chr.4: 93829930-93829930 on Build GRCh38
rs755620274 Chr.4: 93829632-93829632 on Build GRCh38

View Full Product Details