Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 97943764-97944264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STPG2
Entrez Gene ID
285555
Gene Name
sperm tail PG-rich repeat containing 2
Gene Aliases
C4orf37
UniGene
Hs.680226
Gene Chromosome Location
Chr.4: 97441450-98143473 on Build GRCh38
Transcripts
NM_174952.2 XM_011531886.2 XM_017008049.1 XM_017008050.1 XM_017008051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6484121 c.934-2A>T p.? Chr.4: 97944009-97944009 on Build GRCh38
2989319 c.974T>C p.L325S Chr.4: 97943967-97943967 on Build GRCh38
3357831 c.967G>A p.D323N Chr.4: 97943974-97943974 on Build GRCh38
6101106 c.982T>C p.L328L Chr.4: 97943959-97943959 on Build GRCh38
3946695 c.1012A>G p.R338G Chr.4: 97943929-97943929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781115465 Chr.4: 97943952-97943952 on Build GRCh38
rs982207790 Chr.4: 97943903-97943903 on Build GRCh38
rs150178155 Chr.4: 97943895-97943895 on Build GRCh38
rs759519883 Chr.4: 97943891-97943891 on Build GRCh38
rs969011318 Chr.4: 97943791-97943791 on Build GRCh38
rs753158689 Chr.4: 97944029-97944029 on Build GRCh38
rs890106123 Chr.4: 97943981-97943981 on Build GRCh38
rs534447160 Chr.4: 97943794-97943794 on Build GRCh38
rs761557604 Chr.4: 97943908-97943908 on Build GRCh38
rs775508159 Chr.4: 97943972-97943972 on Build GRCh38
rs542468900 Chr.4: 97944164-97944164 on Build GRCh38
rs1019788419 Chr.4: 97944177-97944177 on Build GRCh38
rs548790851 Chr.4: 97943793-97943793 on Build GRCh38
rs952981794 Chr.4: 97944162-97944162 on Build GRCh38
rs186384214 Chr.4: 97944099-97944099 on Build GRCh38
rs377127074 Chr.4: 97943819-97943819 on Build GRCh38
rs554323137 Chr.4: 97943982-97943982 on Build GRCh38
rs534707615 Chr.4: 97944217-97944217 on Build GRCh38
rs1009086887 Chr.4: 97943999-97943999 on Build GRCh38
rs762768245 Chr.4: 97943898-97943898 on Build GRCh38
rs915159730 Chr.4: 97943938-97943938 on Build GRCh38
rs781405934 Chr.4: 97944059-97944059 on Build GRCh38
rs769908300 Chr.4: 97943892-97943892 on Build GRCh38
rs953923925 Chr.4: 97944158-97944158 on Build GRCh38
rs780776653 Chr.4: 97943863-97943863 on Build GRCh38
rs770871065 Chr.4: 97943887-97943887 on Build GRCh38
rs754260547 Chr.4: 97943874-97943874 on Build GRCh38
rs972386583 Chr.4: 97944204-97944204 on Build GRCh38
rs764807931 Chr.4: 97944036-97944038 on Build GRCh38
rs560754943 Chr.4: 97944206-97944206 on Build GRCh38
rs755519686 Chr.4: 97943877-97943877 on Build GRCh38
rs780077483 Chr.4: 97943941-97943941 on Build GRCh38
rs534314293 Chr.4: 97944031-97944031 on Build GRCh38
rs572921036 Chr.4: 97944022-97944022 on Build GRCh38
rs964993226 Chr.4: 97944195-97944195 on Build GRCh38
rs923995640 Chr.4: 97943905-97943905 on Build GRCh38
rs749999268 Chr.4: 97943916-97943916 on Build GRCh38
rs199699367 Chr.4: 97943906-97943906 on Build GRCh38
rs558739501 Chr.4: 97944034-97944034 on Build GRCh38
rs931895637 Chr.4: 97943944-97943944 on Build GRCh38
rs765802299 Chr.4: 97943936-97943936 on Build GRCh38
rs139836757 Chr.4: 97943833-97943833 on Build GRCh38
rs768295550 Chr.4: 97943986-97943986 on Build GRCh38
rs999530815 Chr.4: 97944062-97944062 on Build GRCh38
rs577168975 Chr.4: 97944091-97944091 on Build GRCh38
rs1021482052 Chr.4: 97944138-97944141 on Build GRCh38
rs372621822 Chr.4: 97943862-97943862 on Build GRCh38
rs201025912 Chr.4: 97943861-97943861 on Build GRCh38
rs772280874 Chr.4: 97943881-97943881 on Build GRCh38
rs1031602165 Chr.4: 97943818-97943818 on Build GRCh38
rs767260391 Chr.4: 97943992-97943992 on Build GRCh38
rs546346125 Chr.4: 97943822-97943822 on Build GRCh38
rs138653131 Chr.4: 97943896-97943896 on Build GRCh38
rs746009793 Chr.4: 97943960-97943960 on Build GRCh38
rs774243599 Chr.4: 97943989-97943989 on Build GRCh38
rs538211011 Chr.4: 97943828-97943828 on Build GRCh38
rs757544801 Chr.4: 97943947-97943947 on Build GRCh38
rs769996165 Chr.4: 97943964-97943964 on Build GRCh38
rs575170150 Chr.4: 97944156-97944156 on Build GRCh38
rs755466908 Chr.4: 97943926-97943926 on Build GRCh38
rs761442566 Chr.4: 97943942-97943942 on Build GRCh38
rs181835924 Chr.4: 97943806-97943806 on Build GRCh38
rs563215402 Chr.4: 97944129-97944129 on Build GRCh38
rs376961745 Chr.4: 97943880-97943880 on Build GRCh38
rs752015599 Chr.4: 97944055-97944055 on Build GRCh38
rs763961371 Chr.4: 97943900-97943900 on Build GRCh38
rs546678202 Chr.4: 97944240-97944240 on Build GRCh38
rs970701828 Chr.4: 97944225-97944225 on Build GRCh38
rs772965298 Chr.4: 97944047-97944048 on Build GRCh38
rs952907198 Chr.4: 97943866-97943866 on Build GRCh38
rs368781633 Chr.4: 97943946-97943946 on Build GRCh38
rs374105338 Chr.4: 97943984-97943984 on Build GRCh38
rs373049652 Chr.4: 97944023-97944023 on Build GRCh38
rs369863947 Chr.4: 97944098-97944098 on Build GRCh38
rs146023525 Chr.4: 97943827-97943827 on Build GRCh38
rs747273101 Chr.4: 97943885-97943885 on Build GRCh38
rs764340861 Chr.4: 97944053-97944053 on Build GRCh38
rs1006672291 Chr.4: 97944171-97944171 on Build GRCh38
rs751226924 Chr.4: 97943851-97943851 on Build GRCh38
rs757414789 Chr.4: 97944057-97944057 on Build GRCh38
rs779229450 Chr.4: 97943879-97943879 on Build GRCh38
rs766907171 Chr.4: 97943914-97943914 on Build GRCh38
rs763904806 Chr.4: 97943847-97943847 on Build GRCh38
rs761349814 Chr.4: 97943991-97943991 on Build GRCh38
rs776809314 Chr.4: 97943888-97943888 on Build GRCh38
rs953848803 Chr.4: 97943872-97943872 on Build GRCh38
rs778226443 Chr.4: 97943942-97943942 on Build GRCh38
rs527842862 Chr.4: 97944230-97944230 on Build GRCh38
rs961549590 Chr.4: 97943805-97943805 on Build GRCh38

View Full Product Details