Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 98884814-98885346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E
Entrez Gene ID
1977
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
AUTS19, CBP, EIF4E1, EIF4EL1, EIF4F, eIF-4E
UniGene
Hs.249718
Gene Chromosome Location
Chr.4: 98878456-98930635 on Build GRCh38
Transcripts
NM_001130678.1 NM_001130679.1 NM_001968.3 XM_006714126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680641 c.531A>G p.T177T Chr.4: 98884930-98884930 on Build GRCh38
6535393 c.418G>C p.E140Q Chr.4: 98885043-98885043 on Build GRCh38
6545634 c.481G>A p.D161N Chr.4: 98884980-98884980 on Build GRCh38
35428 c.453C>T p.G151G Chr.4: 98885008-98885008 on Build GRCh38
6652925 c.402T>C p.L134L Chr.4: 98885059-98885059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953313839 Chr.4: 98885146-98885146 on Build GRCh38
rs977551705 Chr.4: 98885203-98885203 on Build GRCh38
rs190358403 Chr.4: 98885266-98885266 on Build GRCh38
rs753775224 Chr.4: 98885093-98885093 on Build GRCh38
rs767620602 Chr.4: 98885065-98885065 on Build GRCh38
rs538640561 Chr.4: 98884896-98884896 on Build GRCh38
rs573869577 Chr.4: 98885205-98885205 on Build GRCh38
rs910675000 Chr.4: 98885109-98885109 on Build GRCh38
rs746510895 Chr.4: 98884915-98884915 on Build GRCh38
rs1048533592 Chr.4: 98885233-98885233 on Build GRCh38
rs1002688766 Chr.4: 98885208-98885208 on Build GRCh38
rs928598070 Chr.4: 98885210-98885210 on Build GRCh38
rs189400211 Chr.4: 98884858-98884858 on Build GRCh38
rs369619373 Chr.4: 98885151-98885151 on Build GRCh38
rs773473491 Chr.4: 98885067-98885067 on Build GRCh38
rs3833620 Chr.4: 98884875-98884876 on Build GRCh38
rs1009609045 Chr.4: 98885284-98885284 on Build GRCh38
rs374882767 Chr.4: 98884897-98884897 on Build GRCh38
rs1049204084 Chr.4: 98885170-98885170 on Build GRCh38
rs758129582 Chr.4: 98885111-98885111 on Build GRCh38
rs777386723 Chr.4: 98885112-98885112 on Build GRCh38
rs752434060 Chr.4: 98885105-98885105 on Build GRCh38
rs760614564 Chr.4: 98885064-98885065 on Build GRCh38
rs369930220 Chr.4: 98884903-98884903 on Build GRCh38
rs914436736 Chr.4: 98885161-98885161 on Build GRCh38
rs997807306 Chr.4: 98885127-98885127 on Build GRCh38
rs373421201 Chr.4: 98884901-98884901 on Build GRCh38
rs773512920 Chr.4: 98884937-98884937 on Build GRCh38
rs918996795 Chr.4: 98885128-98885128 on Build GRCh38
rs780115717 Chr.4: 98884891-98884891 on Build GRCh38
rs1028914797 Chr.4: 98885130-98885130 on Build GRCh38
rs185066219 Chr.4: 98885008-98885008 on Build GRCh38
rs537630770 Chr.4: 98885260-98885260 on Build GRCh38
rs766446576 Chr.4: 98885088-98885088 on Build GRCh38
rs192939764 Chr.4: 98884880-98884880 on Build GRCh38
rs773199991 Chr.4: 98885185-98885185 on Build GRCh38
rs774451037 Chr.4: 98885020-98885020 on Build GRCh38
rs553289886 Chr.4: 98885291-98885291 on Build GRCh38
rs777335682 Chr.4: 98884912-98884912 on Build GRCh38
rs34861020 Chr.4: 98884938-98884939 on Build GRCh38
rs775645971 Chr.4: 98885065-98885065 on Build GRCh38
rs929010012 Chr.4: 98885167-98885167 on Build GRCh38
rs916162795 Chr.4: 98885221-98885221 on Build GRCh38
rs1036128326 Chr.4: 98885267-98885267 on Build GRCh38
rs746565912 Chr.4: 98885113-98885113 on Build GRCh38
rs1004666356 Chr.4: 98885091-98885091 on Build GRCh38
rs765013808 Chr.4: 98885099-98885099 on Build GRCh38
rs372999306 Chr.4: 98884966-98884966 on Build GRCh38
rs760805893 Chr.4: 98885070-98885070 on Build GRCh38
rs763245360 Chr.4: 98885237-98885237 on Build GRCh38
rs968700652 Chr.4: 98885016-98885016 on Build GRCh38
rs770337295 Chr.4: 98884932-98884932 on Build GRCh38
rs762161932 Chr.4: 98885050-98885050 on Build GRCh38
rs555608209 Chr.4: 98885150-98885150 on Build GRCh38
rs1055933078 Chr.4: 98885219-98885219 on Build GRCh38
rs114497468 Chr.4: 98885095-98885095 on Build GRCh38
rs749648246 Chr.4: 98884989-98884989 on Build GRCh38
rs887704603 Chr.4: 98885181-98885181 on Build GRCh38
rs572809877 Chr.4: 98885149-98885149 on Build GRCh38
rs931600944 Chr.4: 98885232-98885232 on Build GRCh38
rs938704555 Chr.4: 98885217-98885217 on Build GRCh38
rs989973721 Chr.4: 98885148-98885148 on Build GRCh38
rs369244266 Chr.4: 98884899-98884899 on Build GRCh38

View Full Product Details