Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99202958-99203457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH6
Entrez Gene ID
130
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
ADH-5
UniGene
Hs.586161
Gene Chromosome Location
Chr.4: 99202638-99219246 on Build GRCh38
Transcripts
NM_001102470.1 NR_132990.1 NR_037884.1

Gene Symbol
LOC100507053
Entrez Gene ID
100507053
Gene Name
uncharacterized LOC100507053
Gene Aliases
-
UniGene
Hs.635084
Gene Chromosome Location
Chr.4: 99088857-99301356 on Build GRCh38
Transcripts
NM_001102470.1 NR_132990.1 NR_037884.1

SNP Detail

SNP ID Chromosome Location
rs922905920 Chr.4: 99203256-99203256 on Build GRCh38
rs758883797 Chr.4: 99203172-99203172 on Build GRCh38
rs990821491 Chr.4: 99203241-99203241 on Build GRCh38
rs1011973378 Chr.4: 99203177-99203177 on Build GRCh38
rs983950957 Chr.4: 99203410-99203410 on Build GRCh38
rs946853381 Chr.4: 99203315-99203315 on Build GRCh38
rs533512278 Chr.4: 99203367-99203367 on Build GRCh38
rs780654254 Chr.4: 99203184-99203184 on Build GRCh38
rs1024752168 Chr.4: 99203201-99203201 on Build GRCh38
rs577558185 Chr.4: 99203357-99203357 on Build GRCh38
rs184370277 Chr.4: 99203151-99203151 on Build GRCh38
rs746259104 Chr.4: 99203116-99203116 on Build GRCh38
rs768840906 Chr.4: 99203226-99203226 on Build GRCh38
rs1042791802 Chr.4: 99203341-99203341 on Build GRCh38
rs1035651421 Chr.4: 99203150-99203150 on Build GRCh38
rs1049789770 Chr.4: 99203296-99203296 on Build GRCh38
rs998439329 Chr.4: 99203119-99203119 on Build GRCh38
rs545389232 Chr.4: 99203016-99203016 on Build GRCh38
rs1015718746 Chr.4: 99203415-99203415 on Build GRCh38
rs779204979 Chr.4: 99203051-99203051 on Build GRCh38
rs931215799 Chr.4: 99203368-99203368 on Build GRCh38
rs569090921 Chr.4: 99203006-99203006 on Build GRCh38
rs1013261618 Chr.4: 99203388-99203388 on Build GRCh38
rs1049987050 Chr.4: 99203278-99203278 on Build GRCh38
rs543763861 Chr.4: 99203052-99203052 on Build GRCh38
rs977146991 Chr.4: 99203238-99203238 on Build GRCh38
rs1023238744 Chr.4: 99203209-99203209 on Build GRCh38
rs187916376 Chr.4: 99203224-99203224 on Build GRCh38
rs116429331 Chr.4: 99203216-99203216 on Build GRCh38
rs991360620 Chr.4: 99203197-99203197 on Build GRCh38
rs888897558 Chr.4: 99203056-99203056 on Build GRCh38
rs553079690 Chr.4: 99203017-99203017 on Build GRCh38
rs28720166 Chr.4: 99203013-99203013 on Build GRCh38
rs995829710 Chr.4: 99203078-99203079 on Build GRCh38
rs1130919 Chr.4: 99203392-99203392 on Build GRCh38
rs952524889 Chr.4: 99203396-99203396 on Build GRCh38
rs566317390 Chr.4: 99203386-99203386 on Build GRCh38
rs971543375 Chr.4: 99203214-99203214 on Build GRCh38
rs551791912 Chr.4: 99203427-99203427 on Build GRCh38
rs1022846435 Chr.4: 99203389-99203389 on Build GRCh38
rs551521624 Chr.4: 99203364-99203364 on Build GRCh38
rs576633024 Chr.4: 99203043-99203043 on Build GRCh38
rs1021302566 Chr.4: 99203069-99203069 on Build GRCh38
rs914556859 Chr.4: 99203244-99203244 on Build GRCh38
rs893991570 Chr.4: 99203308-99203308 on Build GRCh38
rs1011300840 Chr.4: 99203060-99203060 on Build GRCh38
rs902938970 Chr.4: 99203354-99203354 on Build GRCh38
rs952494344 Chr.4: 99203213-99203213 on Build GRCh38
rs932880130 Chr.4: 99203265-99203265 on Build GRCh38
rs756903692 Chr.4: 99203018-99203018 on Build GRCh38
rs895459029 Chr.4: 99203375-99203375 on Build GRCh38

View Full Product Details